Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60874068-60874611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC8
Entrez Gene ID
1161
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases
CKN1, CSA, UVSS2
UniGene
Hs.435237
Gene Chromosome Location
Chr.5: 60873832-60945078 on Build GRCh38
Transcripts
NM_000082.3 NM_001007233.2 NM_001290285.1

SNP Detail

SNP ID Chromosome Location
rs995948838 Chr.5: 60874581-60874581 on Build GRCh38
rs1008282178 Chr.5: 60874270-60874270 on Build GRCh38
rs917059743 Chr.5: 60874560-60874560 on Build GRCh38
rs4647152 Chr.5: 60874579-60874579 on Build GRCh38
rs958799964 Chr.5: 60874441-60874441 on Build GRCh38
rs925588173 Chr.5: 60874273-60874273 on Build GRCh38
rs927151457 Chr.5: 60874120-60874120 on Build GRCh38
rs991017332 Chr.5: 60874370-60874370 on Build GRCh38
rs1027690651 Chr.5: 60874353-60874353 on Build GRCh38
rs1005060800 Chr.5: 60874355-60874355 on Build GRCh38
rs4647156 Chr.5: 60874157-60874157 on Build GRCh38
rs778344145 Chr.5: 60874571-60874571 on Build GRCh38
rs147992873 Chr.5: 60874361-60874361 on Build GRCh38
rs971112935 Chr.5: 60874412-60874412 on Build GRCh38
rs559175991 Chr.5: 60874204-60874204 on Build GRCh38
rs1047032 Chr.5: 60874334-60874334 on Build GRCh38
rs4647155 Chr.5: 60874176-60874176 on Build GRCh38
rs1015565700 Chr.5: 60874305-60874305 on Build GRCh38
rs747271093 Chr.5: 60874553-60874554 on Build GRCh38
rs3117 Chr.5: 60874510-60874510 on Build GRCh38
rs944101229 Chr.5: 60874518-60874518 on Build GRCh38
rs772119030 Chr.5: 60874466-60874466 on Build GRCh38
rs935769389 Chr.5: 60874296-60874296 on Build GRCh38
rs924291689 Chr.5: 60874156-60874156 on Build GRCh38
rs577134348 Chr.5: 60874368-60874368 on Build GRCh38
rs891232602 Chr.5: 60874237-60874237 on Build GRCh38
rs1049786668 Chr.5: 60874225-60874225 on Build GRCh38
rs886060719 Chr.5: 60874153-60874153 on Build GRCh38
rs1049524495 Chr.5: 60874504-60874504 on Build GRCh38
rs994939890 Chr.5: 60874382-60874382 on Build GRCh38
rs879085837 Chr.5: 60874314-60874314 on Build GRCh38
rs4647157 Chr.5: 60874096-60874096 on Build GRCh38
rs924267001 Chr.5: 60874445-60874445 on Build GRCh38
rs112808360 Chr.5: 60874425-60874425 on Build GRCh38
rs886060720 Chr.5: 60874239-60874240 on Build GRCh38
rs990074545 Chr.5: 60874487-60874487 on Build GRCh38
rs537108408 Chr.5: 60874352-60874352 on Build GRCh38
rs1051859073 Chr.5: 60874330-60874330 on Build GRCh38
rs567684336 Chr.5: 60874502-60874502 on Build GRCh38
rs568206790 Chr.5: 60874338-60874338 on Build GRCh38
rs373740060 Chr.5: 60874350-60874352 on Build GRCh38
rs4647153 Chr.5: 60874378-60874378 on Build GRCh38
rs4647154 Chr.5: 60874319-60874319 on Build GRCh38
rs145230004 Chr.5: 60874279-60874279 on Build GRCh38
rs754513955 Chr.5: 60874568-60874568 on Build GRCh38
rs934359489 Chr.5: 60874473-60874473 on Build GRCh38
rs914518781 Chr.5: 60874497-60874497 on Build GRCh38
rs748843229 Chr.5: 60874110-60874110 on Build GRCh38
rs981613091 Chr.5: 60874440-60874440 on Build GRCh38
rs368665460 Chr.5: 60874565-60874565 on Build GRCh38
rs973871103 Chr.5: 60874107-60874107 on Build GRCh38
rs565773632 Chr.5: 60874211-60874211 on Build GRCh38

View Full Product Details