Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140245700-140246198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFDN1
Entrez Gene ID
5201
Gene Name
prefoldin subunit 1
Gene Aliases
PDF, PFD1
UniGene
Hs.483564
Gene Chromosome Location
Chr.5: 140245050-140303141 on Build GRCh38
Transcripts
NM_002622.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826889 c.307C>T p.R103* Chr.5: 140246036-140246036 on Build GRCh38
5820118 c.337C>T p.R113W Chr.5: 140246006-140246006 on Build GRCh38
246550 c.332delA p.N111fs*6 Chr.5: 140246011-140246011 on Build GRCh38
4442185 c.353C>A p.A118E Chr.5: 140245990-140245990 on Build GRCh38
1061586 c.313G>A p.V105I Chr.5: 140246030-140246030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376260726 Chr.5: 140246104-140246104 on Build GRCh38
rs370351660 Chr.5: 140246035-140246035 on Build GRCh38
rs1007051020 Chr.5: 140245882-140245882 on Build GRCh38
rs9042 Chr.5: 140245762-140245762 on Build GRCh38
rs920293094 Chr.5: 140245877-140245877 on Build GRCh38
rs11558411 Chr.5: 140245954-140245954 on Build GRCh38
rs994425181 Chr.5: 140245916-140245916 on Build GRCh38
rs562940788 Chr.5: 140245898-140245898 on Build GRCh38
rs201193165 Chr.5: 140245958-140245958 on Build GRCh38
rs946153555 Chr.5: 140245933-140245933 on Build GRCh38
rs757466775 Chr.5: 140246030-140246030 on Build GRCh38
rs181692055 Chr.5: 140245793-140245793 on Build GRCh38
rs954573189 Chr.5: 140245779-140245780 on Build GRCh38
rs778957646 Chr.5: 140245940-140245940 on Build GRCh38
rs373133651 Chr.5: 140245927-140245927 on Build GRCh38
rs376876150 Chr.5: 140246101-140246101 on Build GRCh38
rs910287509 Chr.5: 140246095-140246095 on Build GRCh38
rs935648704 Chr.5: 140246037-140246037 on Build GRCh38
rs888730937 Chr.5: 140246162-140246162 on Build GRCh38
rs1019830309 Chr.5: 140245892-140245892 on Build GRCh38
rs1050126891 Chr.5: 140246042-140246042 on Build GRCh38
rs753965486 Chr.5: 140246006-140246006 on Build GRCh38
rs752930110 Chr.5: 140245976-140245977 on Build GRCh38
rs570375556 Chr.5: 140245792-140245792 on Build GRCh38
rs767437405 Chr.5: 140245948-140245948 on Build GRCh38
rs888895905 Chr.5: 140245810-140245810 on Build GRCh38
rs1053800715 Chr.5: 140245822-140245833 on Build GRCh38
rs868570664 Chr.5: 140246003-140246003 on Build GRCh38
rs980380555 Chr.5: 140245890-140245890 on Build GRCh38
rs758841417 Chr.5: 140246052-140246052 on Build GRCh38
rs141382227 Chr.5: 140245782-140245782 on Build GRCh38
rs112538595 Chr.5: 140245749-140245749 on Build GRCh38
rs186032931 Chr.5: 140246130-140246130 on Build GRCh38
rs758695613 Chr.5: 140246049-140246050 on Build GRCh38
rs548688403 Chr.5: 140245736-140245736 on Build GRCh38
rs772679561 Chr.5: 140245802-140245802 on Build GRCh38
rs780162595 Chr.5: 140245930-140245930 on Build GRCh38
rs200309445 Chr.5: 140246048-140246048 on Build GRCh38
rs992615822 Chr.5: 140245964-140245964 on Build GRCh38
rs962162572 Chr.5: 140246136-140246136 on Build GRCh38
rs967233193 Chr.5: 140245988-140245988 on Build GRCh38
rs992646866 Chr.5: 140245756-140245756 on Build GRCh38
rs927123496 Chr.5: 140245747-140245747 on Build GRCh38
rs11558409 Chr.5: 140245752-140245752 on Build GRCh38
rs764254781 Chr.5: 140245995-140245995 on Build GRCh38
rs756127441 Chr.5: 140245977-140245977 on Build GRCh38
rs904546609 Chr.5: 140245957-140245957 on Build GRCh38
rs901351547 Chr.5: 140246113-140246113 on Build GRCh38
rs11558408 Chr.5: 140245748-140245748 on Build GRCh38
rs866791091 Chr.5: 140245996-140245996 on Build GRCh38
rs946005559 Chr.5: 140245777-140245777 on Build GRCh38
rs778178626 Chr.5: 140246067-140246068 on Build GRCh38
rs3822739 Chr.5: 140246158-140246158 on Build GRCh38
rs780653255 Chr.5: 140246060-140246060 on Build GRCh38
rs747963593 Chr.5: 140246105-140246105 on Build GRCh38
rs374519129 Chr.5: 140245932-140245932 on Build GRCh38
rs11558405 Chr.5: 140245874-140245874 on Build GRCh38
rs539373520 Chr.5: 140246161-140246161 on Build GRCh38
rs757369669 Chr.5: 140245800-140245800 on Build GRCh38
rs754825425 Chr.5: 140245993-140245993 on Build GRCh38
rs747502245 Chr.5: 140246102-140246103 on Build GRCh38
rs376066004 Chr.5: 140245956-140245956 on Build GRCh38
rs1028837136 Chr.5: 140245764-140245764 on Build GRCh38
rs779049738 Chr.5: 140246031-140246031 on Build GRCh38
rs866404347 Chr.5: 140245757-140245757 on Build GRCh38

View Full Product Details