Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43620729-43621234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP2
Entrez Gene ID
54676
Gene Name
GTP binding protein 2
Gene Aliases
-
UniGene
Hs.485449
Gene Chromosome Location
Chr.6: 43620481-43629246 on Build GRCh38
Transcripts
NM_001286216.1 NM_019096.4

SNP Detail

SNP ID Chromosome Location
rs532754140 Chr.6: 43620926-43620926 on Build GRCh38
rs907801921 Chr.6: 43621185-43621185 on Build GRCh38
rs967392243 Chr.6: 43620764-43620764 on Build GRCh38
rs113493779 Chr.6: 43621067-43621067 on Build GRCh38
rs540769356 Chr.6: 43620822-43620822 on Build GRCh38
rs916451127 Chr.6: 43620899-43620899 on Build GRCh38
rs76645046 Chr.6: 43621068-43621068 on Build GRCh38
rs1055965893 Chr.6: 43620776-43620776 on Build GRCh38
rs950201642 Chr.6: 43621052-43621052 on Build GRCh38
rs887351487 Chr.6: 43621069-43621069 on Build GRCh38
rs754711864 Chr.6: 43620991-43620991 on Build GRCh38
rs142764161 Chr.6: 43620969-43620969 on Build GRCh38
rs996952601 Chr.6: 43621141-43621141 on Build GRCh38
rs764607890 Chr.6: 43620800-43620800 on Build GRCh38
rs1040263734 Chr.6: 43621200-43621200 on Build GRCh38
rs528560193 Chr.6: 43620812-43620812 on Build GRCh38
rs975861782 Chr.6: 43620932-43620932 on Build GRCh38
rs1004207110 Chr.6: 43621092-43621092 on Build GRCh38
rs556809124 Chr.6: 43621175-43621175 on Build GRCh38
rs1056280332 Chr.6: 43620891-43620891 on Build GRCh38
rs138697361 Chr.6: 43620927-43620927 on Build GRCh38
rs45618540 Chr.6: 43620996-43620996 on Build GRCh38
rs565474403 Chr.6: 43620925-43620925 on Build GRCh38
rs775764551 Chr.6: 43621160-43621160 on Build GRCh38
rs1028163442 Chr.6: 43620936-43620936 on Build GRCh38
rs940571832 Chr.6: 43621192-43621192 on Build GRCh38
rs925065049 Chr.6: 43621025-43621025 on Build GRCh38
rs529316405 Chr.6: 43620980-43620980 on Build GRCh38
rs995976249 Chr.6: 43620975-43620975 on Build GRCh38
rs1016937877 Chr.6: 43621116-43621116 on Build GRCh38
rs898454426 Chr.6: 43621138-43621138 on Build GRCh38
rs147343298 Chr.6: 43620986-43620986 on Build GRCh38
rs981412383 Chr.6: 43620790-43620790 on Build GRCh38
rs919986016 Chr.6: 43621127-43621127 on Build GRCh38
rs568359730 Chr.6: 43621037-43621037 on Build GRCh38
rs530455578 Chr.6: 43620970-43620970 on Build GRCh38
rs886334070 Chr.6: 43620940-43620940 on Build GRCh38
rs561361142 Chr.6: 43620780-43620780 on Build GRCh38
rs1014472201 Chr.6: 43620786-43620786 on Build GRCh38
rs772023317 Chr.6: 43621056-43621056 on Build GRCh38
rs145977915 Chr.6: 43620832-43620832 on Build GRCh38
rs749531849 Chr.6: 43621176-43621176 on Build GRCh38
rs571069932 Chr.6: 43621106-43621106 on Build GRCh38
rs967588281 Chr.6: 43621046-43621046 on Build GRCh38
rs533462398 Chr.6: 43620756-43620756 on Build GRCh38
rs769827721 Chr.6: 43621154-43621154 on Build GRCh38
rs186614639 Chr.6: 43620869-43620869 on Build GRCh38
rs763401938 Chr.6: 43620782-43620782 on Build GRCh38
rs981444950 Chr.6: 43621049-43621049 on Build GRCh38
rs749970709 Chr.6: 43620948-43620948 on Build GRCh38
rs940372420 Chr.6: 43620943-43620943 on Build GRCh38
rs752051724 Chr.6: 43620828-43620828 on Build GRCh38
rs538461815 Chr.6: 43621136-43621136 on Build GRCh38

View Full Product Details