Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43622450-43622977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTPBP2
Entrez Gene ID
54676
Gene Name
GTP binding protein 2
Gene Aliases
-
UniGene
Hs.485449
Gene Chromosome Location
Chr.6: 43620481-43629246 on Build GRCh38
Transcripts
NM_001286216.1 NM_019096.4 XM_017010976.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5720657 c.14G>A p.R5H Chr.6: 43622791-43622791 on Build GRCh38
1734570 c.1388G>A p.R463H Chr.6: 43622712-43622712 on Build GRCh38
1079657 c.1409G>A p.R470Q Chr.6: 43622691-43622691 on Build GRCh38
6281123 c.1463G>A p.R488H Chr.6: 43622637-43622637 on Build GRCh38
6038947 c.1400G>A p.R467H Chr.6: 43622700-43622700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936757493 Chr.6: 43622573-43622573 on Build GRCh38
rs868521466 Chr.6: 43622889-43622889 on Build GRCh38
rs984221460 Chr.6: 43622561-43622561 on Build GRCh38
rs765762316 Chr.6: 43622649-43622649 on Build GRCh38
rs375126349 Chr.6: 43622601-43622601 on Build GRCh38
rs773711871 Chr.6: 43622724-43622724 on Build GRCh38
rs201929088 Chr.6: 43622623-43622623 on Build GRCh38
rs898798034 Chr.6: 43622687-43622687 on Build GRCh38
rs1050287632 Chr.6: 43622674-43622674 on Build GRCh38
rs753044684 Chr.6: 43622654-43622654 on Build GRCh38
rs776144133 Chr.6: 43622935-43622935 on Build GRCh38
rs904352228 Chr.6: 43622751-43622751 on Build GRCh38
rs772592993 Chr.6: 43622794-43622794 on Build GRCh38
rs749027158 Chr.6: 43622731-43622731 on Build GRCh38
rs16896609 Chr.6: 43622671-43622671 on Build GRCh38
rs373767620 Chr.6: 43622582-43622582 on Build GRCh38
rs375105778 Chr.6: 43622586-43622586 on Build GRCh38
rs373328154 Chr.6: 43622714-43622714 on Build GRCh38
rs748492429 Chr.6: 43622645-43622645 on Build GRCh38
rs868454616 Chr.6: 43622522-43622523 on Build GRCh38
rs372952724 Chr.6: 43622911-43622911 on Build GRCh38
rs534118235 Chr.6: 43622848-43622848 on Build GRCh38
rs764332785 Chr.6: 43622826-43622826 on Build GRCh38
rs760125182 Chr.6: 43622811-43622811 on Build GRCh38
rs768320832 Chr.6: 43622741-43622741 on Build GRCh38
rs763672576 Chr.6: 43622614-43622614 on Build GRCh38
rs983835255 Chr.6: 43622947-43622947 on Build GRCh38
rs774763253 Chr.6: 43622514-43622514 on Build GRCh38
rs1028719766 Chr.6: 43622706-43622706 on Build GRCh38
rs959723175 Chr.6: 43622490-43622490 on Build GRCh38
rs529907749 Chr.6: 43622820-43622820 on Build GRCh38
rs747087499 Chr.6: 43622699-43622699 on Build GRCh38
rs570907811 Chr.6: 43622951-43622951 on Build GRCh38
rs993014339 Chr.6: 43622518-43622518 on Build GRCh38
rs769723702 Chr.6: 43622718-43622718 on Build GRCh38
rs1031378318 Chr.6: 43622486-43622486 on Build GRCh38
rs755520205 Chr.6: 43622648-43622648 on Build GRCh38
rs767105036 Chr.6: 43622789-43622789 on Build GRCh38
rs189928431 Chr.6: 43622522-43622522 on Build GRCh38
rs745867430 Chr.6: 43622715-43622715 on Build GRCh38
rs139589078 Chr.6: 43622847-43622847 on Build GRCh38
rs998804907 Chr.6: 43622783-43622783 on Build GRCh38
rs371811079 Chr.6: 43622594-43622594 on Build GRCh38
rs185215360 Chr.6: 43622837-43622837 on Build GRCh38
rs753132957 Chr.6: 43622817-43622817 on Build GRCh38
rs1007812513 Chr.6: 43622692-43622692 on Build GRCh38
rs138150365 Chr.6: 43622703-43622703 on Build GRCh38
rs112297133 Chr.6: 43622526-43622527 on Build GRCh38
rs143756297 Chr.6: 43622493-43622493 on Build GRCh38
rs35758978 Chr.6: 43622696-43622696 on Build GRCh38
rs761341453 Chr.6: 43622786-43622786 on Build GRCh38
rs774017306 Chr.6: 43622756-43622756 on Build GRCh38
rs777360356 Chr.6: 43622624-43622625 on Build GRCh38
rs989993863 Chr.6: 43622913-43622913 on Build GRCh38
rs985617769 Chr.6: 43622763-43622763 on Build GRCh38
rs915644526 Chr.6: 43622519-43622519 on Build GRCh38
rs889848479 Chr.6: 43622691-43622691 on Build GRCh38
rs754409219 Chr.6: 43622647-43622647 on Build GRCh38
rs757404538 Chr.6: 43622831-43622831 on Build GRCh38
rs940317452 Chr.6: 43622619-43622619 on Build GRCh38
rs181366514 Chr.6: 43622558-43622558 on Build GRCh38
rs559128811 Chr.6: 43622893-43622893 on Build GRCh38
rs762664560 Chr.6: 43622602-43622602 on Build GRCh38
rs369604717 Chr.6: 43622665-43622665 on Build GRCh38
rs1004546621 Chr.6: 43622667-43622667 on Build GRCh38
rs765780417 Chr.6: 43622816-43622816 on Build GRCh38
rs969796967 Chr.6: 43622898-43622898 on Build GRCh38
rs528049978 Chr.6: 43622485-43622485 on Build GRCh38
rs775271288 Chr.6: 43622719-43622719 on Build GRCh38
rs766827821 Chr.6: 43622643-43622643 on Build GRCh38
rs910140705 Chr.6: 43622574-43622574 on Build GRCh38

View Full Product Details