Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96207508-96208014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT9
Entrez Gene ID
10690
Gene Name
fucosyltransferase 9
Gene Aliases
Fuc-TIX
UniGene
Hs.49117
Gene Chromosome Location
Chr.6: 96015969-96215612 on Build GRCh38
Transcripts
NM_006581.3 XM_011535383.2 XM_011535385.2 XM_017010188.1 XM_017010190.1

SNP Detail

SNP ID Chromosome Location
rs547233217 Chr.6: 96207684-96207684 on Build GRCh38
rs34525493 Chr.6: 96207985-96207986 on Build GRCh38
rs995518924 Chr.6: 96207918-96207918 on Build GRCh38
rs1021575451 Chr.6: 96207932-96207932 on Build GRCh38
rs982799582 Chr.6: 96207957-96207957 on Build GRCh38
rs866174577 Chr.6: 96207787-96207787 on Build GRCh38
rs186445016 Chr.6: 96207663-96207663 on Build GRCh38
rs975611728 Chr.6: 96207640-96207640 on Build GRCh38
rs1021480152 Chr.6: 96207536-96207536 on Build GRCh38
rs1027065688 Chr.6: 96207569-96207569 on Build GRCh38
rs4143938 Chr.6: 96207610-96207610 on Build GRCh38
rs189736854 Chr.6: 96207868-96207868 on Build GRCh38
rs912693791 Chr.6: 96207598-96207598 on Build GRCh38
rs762206652 Chr.6: 96207960-96207960 on Build GRCh38
rs1046193409 Chr.6: 96207837-96207837 on Build GRCh38
rs1027360019 Chr.6: 96207919-96207919 on Build GRCh38
rs1034182337 Chr.6: 96207976-96207976 on Build GRCh38
rs575466245 Chr.6: 96207565-96207565 on Build GRCh38
rs892437187 Chr.6: 96207895-96207895 on Build GRCh38
rs546878725 Chr.6: 96207576-96207576 on Build GRCh38
rs998279414 Chr.6: 96207882-96207882 on Build GRCh38
rs560291199 Chr.6: 96207693-96207693 on Build GRCh38
rs945074077 Chr.6: 96207803-96207803 on Build GRCh38
rs529262221 Chr.6: 96207757-96207757 on Build GRCh38
rs7746212 Chr.6: 96207589-96207589 on Build GRCh38
rs141572572 Chr.6: 96207893-96207893 on Build GRCh38
rs562209096 Chr.6: 96207543-96207543 on Build GRCh38
rs942895623 Chr.6: 96207836-96207836 on Build GRCh38
rs566678123 Chr.6: 96207741-96207741 on Build GRCh38
rs1039834875 Chr.6: 96207846-96207846 on Build GRCh38
rs865880300 Chr.6: 96207573-96207573 on Build GRCh38
rs868458921 Chr.6: 96207730-96207730 on Build GRCh38
rs551591171 Chr.6: 96207915-96207915 on Build GRCh38
rs571376764 Chr.6: 96207939-96207939 on Build GRCh38
rs936337083 Chr.6: 96207676-96207676 on Build GRCh38
rs536023022 Chr.6: 96207754-96207754 on Build GRCh38
rs774716275 Chr.6: 96207564-96207564 on Build GRCh38
rs549516949 Chr.6: 96207785-96207785 on Build GRCh38
rs892212958 Chr.6: 96207795-96207795 on Build GRCh38
rs899113100 Chr.6: 96207914-96207914 on Build GRCh38
rs888635636 Chr.6: 96207820-96207823 on Build GRCh38
rs1048601410 Chr.6: 96207807-96207807 on Build GRCh38
rs1001401645 Chr.6: 96207975-96207976 on Build GRCh38
rs951588126 Chr.6: 96207585-96207585 on Build GRCh38
rs938435678 Chr.6: 96207633-96207633 on Build GRCh38
rs944221961 Chr.6: 96207603-96207603 on Build GRCh38
rs930237548 Chr.6: 96207711-96207711 on Build GRCh38
rs906342192 Chr.6: 96207867-96207867 on Build GRCh38

View Full Product Details