Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125977539-125978080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT3
Entrez Gene ID
135114
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
HINT4
UniGene
Hs.72325
Gene Chromosome Location
Chr.6: 125956715-125980244 on Build GRCh38
Transcripts
NM_138571.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7448594 c.518C>T p.A173V Chr.6: 125977645-125977645 on Build GRCh38
391965 c.524delA p.H175fs*2 Chr.6: 125977651-125977651 on Build GRCh38
1072950 c.547T>C p.*183R Chr.6: 125977674-125977674 on Build GRCh38
7271038 c.517-2A>C p.? Chr.6: 125977642-125977642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903789864 Chr.6: 125977687-125977687 on Build GRCh38
rs1047069046 Chr.6: 125977629-125977629 on Build GRCh38
rs181030637 Chr.6: 125977601-125977601 on Build GRCh38
rs1043381506 Chr.6: 125977635-125977635 on Build GRCh38
rs1003897757 Chr.6: 125977766-125977766 on Build GRCh38
rs558109437 Chr.6: 125977797-125977797 on Build GRCh38
rs1056255026 Chr.6: 125977579-125977579 on Build GRCh38
rs985440453 Chr.6: 125977978-125977978 on Build GRCh38
rs1002271249 Chr.6: 125977691-125977691 on Build GRCh38
rs986545490 Chr.6: 125977966-125977966 on Build GRCh38
rs746810009 Chr.6: 125977612-125977612 on Build GRCh38
rs770588383 Chr.6: 125977703-125977703 on Build GRCh38
rs758724584 Chr.6: 125977593-125977593 on Build GRCh38
rs773941060 Chr.6: 125977659-125977660 on Build GRCh38
rs965169159 Chr.6: 125977792-125977792 on Build GRCh38
rs762528806 Chr.6: 125977683-125977683 on Build GRCh38
rs796866383 Chr.6: 125977583-125977583 on Build GRCh38
rs999409699 Chr.6: 125977741-125977741 on Build GRCh38
rs747709906 Chr.6: 125977709-125977710 on Build GRCh38
rs774049144 Chr.6: 125977709-125977709 on Build GRCh38
rs1026618800 Chr.6: 125977705-125977705 on Build GRCh38
rs1052606698 Chr.6: 125977589-125977589 on Build GRCh38
rs538139215 Chr.6: 125977809-125977810 on Build GRCh38
rs761025513 Chr.6: 125977710-125977710 on Build GRCh38
rs954115771 Chr.6: 125977904-125977904 on Build GRCh38
rs185302825 Chr.6: 125977852-125977852 on Build GRCh38
rs759601933 Chr.6: 125977647-125977647 on Build GRCh38
rs963294740 Chr.6: 125978041-125978041 on Build GRCh38
rs772812993 Chr.6: 125977678-125977678 on Build GRCh38
rs537656554 Chr.6: 125977842-125977842 on Build GRCh38
rs28394097 Chr.6: 125977891-125977891 on Build GRCh38
rs773053210 Chr.6: 125977996-125977999 on Build GRCh38
rs768627104 Chr.6: 125977617-125977617 on Build GRCh38
rs780984691 Chr.6: 125977650-125977650 on Build GRCh38
rs888197662 Chr.6: 125977755-125977755 on Build GRCh38
rs143604204 Chr.6: 125977980-125977980 on Build GRCh38
rs562396311 Chr.6: 125977882-125977882 on Build GRCh38
rs764561469 Chr.6: 125977724-125977724 on Build GRCh38
rs949645074 Chr.6: 125977611-125977611 on Build GRCh38
rs572371820 Chr.6: 125977572-125977572 on Build GRCh38
rs765379585 Chr.6: 125977902-125977902 on Build GRCh38
rs1015392052 Chr.6: 125977777-125977777 on Build GRCh38
rs775848205 Chr.6: 125977958-125977958 on Build GRCh38
rs376018013 Chr.6: 125977975-125977975 on Build GRCh38
rs576900168 Chr.6: 125977805-125977805 on Build GRCh38
rs748153797 Chr.6: 125977651-125977651 on Build GRCh38

View Full Product Details