Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23273430-23273938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312974 c.1466-2A>G p.? Chr.7: 23273519-23273519 on Build GRCh38
6109700 c.1525A>G p.I509V Chr.7: 23273580-23273580 on Build GRCh38
6338582 c.1475C>T p.S492L Chr.7: 23273530-23273530 on Build GRCh38
6673389 c.1510G>C p.V504L Chr.7: 23273565-23273565 on Build GRCh38
7198915 c.1466-1G>C p.? Chr.7: 23273520-23273520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772043664 Chr.7: 23273634-23273634 on Build GRCh38
rs754168711 Chr.7: 23273789-23273789 on Build GRCh38
rs901329430 Chr.7: 23273821-23273821 on Build GRCh38
rs796798742 Chr.7: 23273841-23273841 on Build GRCh38
rs371885339 Chr.7: 23273565-23273565 on Build GRCh38
rs763396154 Chr.7: 23273591-23273591 on Build GRCh38
rs778744522 Chr.7: 23273631-23273631 on Build GRCh38
rs200362530 Chr.7: 23273559-23273559 on Build GRCh38
rs981373132 Chr.7: 23273703-23273703 on Build GRCh38
rs756416732 Chr.7: 23273531-23273531 on Build GRCh38
rs142059223 Chr.7: 23273471-23273471 on Build GRCh38
rs780357717 Chr.7: 23273616-23273616 on Build GRCh38
rs191149673 Chr.7: 23273900-23273900 on Build GRCh38
rs997054023 Chr.7: 23273829-23273829 on Build GRCh38
rs531800146 Chr.7: 23273644-23273644 on Build GRCh38
rs1054057763 Chr.7: 23273731-23273731 on Build GRCh38
rs1011345089 Chr.7: 23273863-23273863 on Build GRCh38
rs371837989 Chr.7: 23273633-23273633 on Build GRCh38
rs156420 Chr.7: 23273649-23273649 on Build GRCh38
rs760219653 Chr.7: 23273572-23273572 on Build GRCh38
rs553863004 Chr.7: 23273823-23273823 on Build GRCh38
rs927252196 Chr.7: 23273715-23273715 on Build GRCh38
rs139732810 Chr.7: 23273561-23273561 on Build GRCh38
rs764691173 Chr.7: 23273502-23273502 on Build GRCh38
rs111824849 Chr.7: 23273518-23273518 on Build GRCh38
rs754803421 Chr.7: 23273620-23273620 on Build GRCh38
rs2268748 Chr.7: 23273552-23273552 on Build GRCh38
rs1021268497 Chr.7: 23273874-23273874 on Build GRCh38
rs767693130 Chr.7: 23273601-23273601 on Build GRCh38
rs144208310 Chr.7: 23273580-23273580 on Build GRCh38
rs753154220 Chr.7: 23273484-23273484 on Build GRCh38
rs962132156 Chr.7: 23273658-23273658 on Build GRCh38
rs762478565 Chr.7: 23273599-23273599 on Build GRCh38
rs1046082555 Chr.7: 23273462-23273462 on Build GRCh38
rs755446171 Chr.7: 23273830-23273830 on Build GRCh38
rs148779588 Chr.7: 23273530-23273530 on Build GRCh38
rs1048351936 Chr.7: 23273749-23273749 on Build GRCh38
rs940236506 Chr.7: 23273806-23273806 on Build GRCh38
rs763713616 Chr.7: 23273528-23273528 on Build GRCh38
rs146559546 Chr.7: 23273598-23273598 on Build GRCh38
rs752814162 Chr.7: 23273858-23273858 on Build GRCh38
rs967040760 Chr.7: 23273880-23273880 on Build GRCh38
rs141502819 Chr.7: 23273543-23273543 on Build GRCh38
rs746559479 Chr.7: 23273639-23273639 on Build GRCh38
rs998772124 Chr.7: 23273891-23273891 on Build GRCh38
rs533895787 Chr.7: 23273746-23273746 on Build GRCh38
rs771563559 Chr.7: 23273566-23273566 on Build GRCh38
rs1041320468 Chr.7: 23273812-23273812 on Build GRCh38
rs560128748 Chr.7: 23273595-23273595 on Build GRCh38
rs908556277 Chr.7: 23273778-23273778 on Build GRCh38
rs775677080 Chr.7: 23273587-23273587 on Build GRCh38
rs938569307 Chr.7: 23273687-23273687 on Build GRCh38
rs781499634 Chr.7: 23273522-23273522 on Build GRCh38
rs757426958 Chr.7: 23273503-23273503 on Build GRCh38
rs559944461 Chr.7: 23273586-23273586 on Build GRCh38
rs368070458 Chr.7: 23273619-23273619 on Build GRCh38
rs991977215 Chr.7: 23273742-23273742 on Build GRCh38
rs200910471 Chr.7: 23273608-23273608 on Build GRCh38
rs772762462 Chr.7: 23273567-23273567 on Build GRCh38
rs2268747 Chr.7: 23273843-23273843 on Build GRCh38
rs571426706 Chr.7: 23273743-23273743 on Build GRCh38
rs375167919 Chr.7: 23273650-23273650 on Build GRCh38
rs549699385 Chr.7: 23273611-23273611 on Build GRCh38
rs11537975 Chr.7: 23273606-23273606 on Build GRCh38
rs765503003 Chr.7: 23273474-23273474 on Build GRCh38
rs369707456 Chr.7: 23273657-23273657 on Build GRCh38
rs774203216 Chr.7: 23273910-23273910 on Build GRCh38
rs928420954 Chr.7: 23273670-23273670 on Build GRCh38
rs112596920 Chr.7: 23273615-23273615 on Build GRCh38
rs949950966 Chr.7: 23273695-23273695 on Build GRCh38
rs1041882331 Chr.7: 23273798-23273798 on Build GRCh38
rs763337542 Chr.7: 23273489-23273489 on Build GRCh38
rs749152145 Chr.7: 23273548-23273548 on Build GRCh38
rs373399241 Chr.7: 23273659-23273659 on Build GRCh38
rs936968815 Chr.7: 23273727-23273727 on Build GRCh38

View Full Product Details