Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157695950-157696460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL3
Entrez Gene ID
115352
Gene Name
Fc receptor like 3
Gene Aliases
CD307c, FCRH3, IFGP3, IRTA3, SPAP2
UniGene
Hs.292449
Gene Chromosome Location
Chr.1: 157676481-157700985 on Build GRCh38
Transcripts
NM_001320333.1 NM_052939.3 NR_135214.1 NR_135215.1 NR_135216.1 NR_135217.1 XM_006711145.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5716369 c.1050G>A p.E350E Chr.1: 157696122-157696122 on Build GRCh38
1985288 c.1054G>A p.D352N Chr.1: 157696118-157696118 on Build GRCh38
4861030 c.937C>G p.Q313E Chr.1: 157696235-157696235 on Build GRCh38
4023534 c.895C>T p.L299L Chr.1: 157696277-157696277 on Build GRCh38
897559 c.1095C>T p.S365S Chr.1: 157696077-157696077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769911717 Chr.1: 157696060-157696060 on Build GRCh38
rs563378114 Chr.1: 157695984-157695984 on Build GRCh38
rs115596233 Chr.1: 157696294-157696294 on Build GRCh38
rs375355913 Chr.1: 157696030-157696030 on Build GRCh38
rs749904431 Chr.1: 157696174-157696174 on Build GRCh38
rs149701662 Chr.1: 157696283-157696283 on Build GRCh38
rs762711979 Chr.1: 157696339-157696339 on Build GRCh38
rs777663586 Chr.1: 157696365-157696365 on Build GRCh38
rs899255335 Chr.1: 157696107-157696107 on Build GRCh38
rs115903952 Chr.1: 157696237-157696237 on Build GRCh38
rs915613498 Chr.1: 157696246-157696246 on Build GRCh38
rs376010044 Chr.1: 157696130-157696130 on Build GRCh38
rs765590832 Chr.1: 157696011-157696011 on Build GRCh38
rs781010547 Chr.1: 157696216-157696216 on Build GRCh38
rs145953660 Chr.1: 157696257-157696257 on Build GRCh38
rs1055164722 Chr.1: 157696110-157696110 on Build GRCh38
rs773644561 Chr.1: 157696397-157696397 on Build GRCh38
rs190625090 Chr.1: 157696227-157696227 on Build GRCh38
rs748705036 Chr.1: 157696383-157696383 on Build GRCh38
rs767917731 Chr.1: 157696156-157696156 on Build GRCh38
rs752692952 Chr.1: 157696355-157696355 on Build GRCh38
rs777131717 Chr.1: 157696034-157696037 on Build GRCh38
rs770174272 Chr.1: 157696391-157696391 on Build GRCh38
rs759624449 Chr.1: 157696151-157696151 on Build GRCh38
rs765896045 Chr.1: 157696342-157696342 on Build GRCh38
rs376853177 Chr.1: 157696327-157696327 on Build GRCh38
rs762200307 Chr.1: 157696268-157696268 on Build GRCh38
rs757021147 Chr.1: 157696167-157696167 on Build GRCh38
rs765449039 Chr.1: 157695995-157695995 on Build GRCh38
rs764941090 Chr.1: 157696173-157696173 on Build GRCh38
rs768496707 Chr.1: 157696138-157696138 on Build GRCh38
rs537235260 Chr.1: 157696319-157696319 on Build GRCh38
rs754226096 Chr.1: 157696163-157696163 on Build GRCh38
rs751110019 Chr.1: 157696348-157696348 on Build GRCh38
rs774439537 Chr.1: 157696146-157696146 on Build GRCh38
rs140279920 Chr.1: 157696040-157696040 on Build GRCh38
rs1052043785 Chr.1: 157696092-157696092 on Build GRCh38
rs775169447 Chr.1: 157696054-157696054 on Build GRCh38
rs146382073 Chr.1: 157696162-157696162 on Build GRCh38
rs781069840 Chr.1: 157696292-157696292 on Build GRCh38
rs774568399 Chr.1: 157696230-157696230 on Build GRCh38
rs747096549 Chr.1: 157696371-157696371 on Build GRCh38
rs918059670 Chr.1: 157696119-157696119 on Build GRCh38
rs761983243 Chr.1: 157695991-157695991 on Build GRCh38
rs753037286 Chr.1: 157696096-157696096 on Build GRCh38
rs763145695 Chr.1: 157696063-157696063 on Build GRCh38
rs367748106 Chr.1: 157696204-157696204 on Build GRCh38
rs747886104 Chr.1: 157696219-157696219 on Build GRCh38
rs1037988401 Chr.1: 157696411-157696411 on Build GRCh38
rs754875256 Chr.1: 157696214-157696214 on Build GRCh38
rs12041673 Chr.1: 157696253-157696253 on Build GRCh38
rs778607388 Chr.1: 157696121-157696121 on Build GRCh38
rs769289421 Chr.1: 157696321-157696321 on Build GRCh38
rs745869083 Chr.1: 157696051-157696051 on Build GRCh38
rs574986381 Chr.1: 157696368-157696368 on Build GRCh38
rs745363847 Chr.1: 157696123-157696123 on Build GRCh38
rs955354738 Chr.1: 157696226-157696226 on Build GRCh38
rs761285739 Chr.1: 157696243-157696243 on Build GRCh38
rs777586064 Chr.1: 157696104-157696104 on Build GRCh38
rs770508420 Chr.1: 157696046-157696046 on Build GRCh38
rs560196456 Chr.1: 157696085-157696085 on Build GRCh38
rs781415832 Chr.1: 157696381-157696381 on Build GRCh38
rs767459603 Chr.1: 157696352-157696352 on Build GRCh38
rs757375284 Chr.1: 157696255-157696255 on Build GRCh38
rs370492097 Chr.1: 157696021-157696021 on Build GRCh38
rs368325573 Chr.1: 157696086-157696086 on Build GRCh38
rs371001813 Chr.1: 157696370-157696370 on Build GRCh38
rs556446223 Chr.1: 157696349-157696349 on Build GRCh38
rs749548879 Chr.1: 157696145-157696145 on Build GRCh38
rs562810998 Chr.1: 157696013-157696013 on Build GRCh38
rs947122182 Chr.1: 157696073-157696073 on Build GRCh38
rs768023202 Chr.1: 157696090-157696090 on Build GRCh38
rs138219281 Chr.1: 157696062-157696062 on Build GRCh38
rs777531137 Chr.1: 157696031-157696031 on Build GRCh38
rs898146500 Chr.1: 157696412-157696412 on Build GRCh38
rs564259735 Chr.1: 157696141-157696141 on Build GRCh38
rs7517830 Chr.1: 157696015-157696015 on Build GRCh38
rs756045540 Chr.1: 157696364-157696364 on Build GRCh38
rs779416617 Chr.1: 157696203-157696203 on Build GRCh38
rs148422137 Chr.1: 157696286-157696286 on Build GRCh38
rs757865879 Chr.1: 157696180-157696180 on Build GRCh38
rs370033272 Chr.1: 157695989-157695989 on Build GRCh38
rs200782762 Chr.1: 157696099-157696099 on Build GRCh38
rs371651307 Chr.1: 157696331-157696331 on Build GRCh38
rs762394881 Chr.1: 157696328-157696328 on Build GRCh38
rs760476507 Chr.1: 157696055-157696055 on Build GRCh38
rs772016366 Chr.1: 157696052-157696052 on Build GRCh38
rs745951308 Chr.1: 157696231-157696231 on Build GRCh38
rs754929262 Chr.1: 157696287-157696287 on Build GRCh38
rs111735268 Chr.1: 157696017-157696017 on Build GRCh38
rs750531331 Chr.1: 157696001-157696001 on Build GRCh38
rs571480875 Chr.1: 157696298-157696298 on Build GRCh38
rs963474346 Chr.1: 157695997-157695997 on Build GRCh38
rs915646866 Chr.1: 157696070-157696070 on Build GRCh38
rs146153982 Chr.1: 157696295-157696295 on Build GRCh38
rs996525883 Chr.1: 157696421-157696421 on Build GRCh38
rs199931244 Chr.1: 157696301-157696301 on Build GRCh38
rs759951376 Chr.1: 157695999-157695999 on Build GRCh38
rs568327102 Chr.1: 157696238-157696238 on Build GRCh38
rs749294556 Chr.1: 157696388-157696388 on Build GRCh38
rs779972812 Chr.1: 157696131-157696131 on Build GRCh38
rs371587126 Chr.1: 157696106-157696106 on Build GRCh38
rs773413038 Chr.1: 157696332-157696332 on Build GRCh38
rs544479679 Chr.1: 157696010-157696010 on Build GRCh38
rs778553895 Chr.1: 157696047-157696047 on Build GRCh38
rs371493046 Chr.1: 157696284-157696284 on Build GRCh38
rs746793090 Chr.1: 157696132-157696132 on Build GRCh38
rs768913583 Chr.1: 157696377-157696377 on Build GRCh38
rs567003130 Chr.1: 157696279-157696279 on Build GRCh38

View Full Product Details