Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162384682-162385179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf226
Entrez Gene ID
400793
Gene Name
chromosome 1 open reading frame 226
Gene Aliases
-
UniGene
Hs.447011
Gene Chromosome Location
Chr.1: 162378906-162386818 on Build GRCh38
Transcripts
NM_001085375.1

SNP Detail

SNP ID Chromosome Location
rs888175997 Chr.1: 162384762-162384762 on Build GRCh38
rs1024919241 Chr.1: 162384772-162384772 on Build GRCh38
rs773070080 Chr.1: 162384928-162384928 on Build GRCh38
rs970064308 Chr.1: 162384885-162384885 on Build GRCh38
rs1043171992 Chr.1: 162385041-162385041 on Build GRCh38
rs1014180296 Chr.1: 162384850-162384850 on Build GRCh38
rs958107668 Chr.1: 162384991-162384991 on Build GRCh38
rs374733407 Chr.1: 162384983-162384983 on Build GRCh38
rs950704475 Chr.1: 162384820-162384820 on Build GRCh38
rs763883899 Chr.1: 162384949-162384949 on Build GRCh38
rs974532391 Chr.1: 162384923-162384923 on Build GRCh38
rs962274406 Chr.1: 162384770-162384770 on Build GRCh38
rs1015090748 Chr.1: 162384767-162384767 on Build GRCh38
rs536142743 Chr.1: 162384857-162384857 on Build GRCh38
rs752951784 Chr.1: 162384856-162384856 on Build GRCh38
rs943850872 Chr.1: 162385034-162385034 on Build GRCh38
rs541712292 Chr.1: 162385153-162385153 on Build GRCh38
rs1055827735 Chr.1: 162385132-162385132 on Build GRCh38
rs960104769 Chr.1: 162384724-162384724 on Build GRCh38
rs769258229 Chr.1: 162384873-162384873 on Build GRCh38
rs535149658 Chr.1: 162385076-162385076 on Build GRCh38
rs924832 Chr.1: 162385136-162385136 on Build GRCh38
rs186713921 Chr.1: 162385149-162385149 on Build GRCh38
rs539956274 Chr.1: 162385138-162385138 on Build GRCh38
rs576985349 Chr.1: 162385101-162385101 on Build GRCh38
rs770670359 Chr.1: 162385057-162385057 on Build GRCh38
rs963118475 Chr.1: 162384884-162384884 on Build GRCh38
rs921579836 Chr.1: 162384944-162384944 on Build GRCh38
rs16861569 Chr.1: 162384716-162384716 on Build GRCh38
rs867783661 Chr.1: 162385135-162385135 on Build GRCh38
rs946293362 Chr.1: 162385039-162385039 on Build GRCh38
rs774710757 Chr.1: 162385148-162385148 on Build GRCh38
rs980722942 Chr.1: 162384962-162384962 on Build GRCh38
rs164419 Chr.1: 162384735-162384735 on Build GRCh38
rs555836552 Chr.1: 162385012-162385012 on Build GRCh38
rs904733485 Chr.1: 162385099-162385099 on Build GRCh38
rs3738393 Chr.1: 162385013-162385013 on Build GRCh38
rs572744056 Chr.1: 162385100-162385100 on Build GRCh38

View Full Product Details