Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96160322-96160813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF6
Entrez Gene ID
392255
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1
UniGene
Hs.492277
Gene Chromosome Location
Chr.8: 96142330-96160792 on Build GRCh38
Transcripts
NM_001001557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311430 c.303C>A p.I101I Chr.8: 96160390-96160390 on Build GRCh38
1208063 c.269C>T p.P90L Chr.8: 96160424-96160424 on Build GRCh38
3902271 c.140A>C p.K47T Chr.8: 96160553-96160553 on Build GRCh38
5777355 c.64G>T p.G22C Chr.8: 96160629-96160629 on Build GRCh38
295654 c.273C>T p.R91R Chr.8: 96160420-96160420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779042060 Chr.8: 96160672-96160672 on Build GRCh38
rs773483807 Chr.8: 96160428-96160428 on Build GRCh38
rs966657559 Chr.8: 96160778-96160778 on Build GRCh38
rs779881000 Chr.8: 96160715-96160715 on Build GRCh38
rs921660513 Chr.8: 96160781-96160781 on Build GRCh38
rs866637539 Chr.8: 96160641-96160641 on Build GRCh38
rs370658707 Chr.8: 96160378-96160378 on Build GRCh38
rs770249145 Chr.8: 96160616-96160616 on Build GRCh38
rs755677702 Chr.8: 96160387-96160387 on Build GRCh38
rs776936551 Chr.8: 96160423-96160423 on Build GRCh38
rs772028403 Chr.8: 96160706-96160706 on Build GRCh38
rs778071820 Chr.8: 96160663-96160663 on Build GRCh38
rs760870324 Chr.8: 96160353-96160353 on Build GRCh38
rs760057321 Chr.8: 96160633-96160633 on Build GRCh38
rs766832433 Chr.8: 96160557-96160557 on Build GRCh38
rs768251290 Chr.8: 96160613-96160613 on Build GRCh38
rs990872341 Chr.8: 96160407-96160407 on Build GRCh38
rs773668867 Chr.8: 96160653-96160653 on Build GRCh38
rs768741148 Chr.8: 96160421-96160421 on Build GRCh38
rs139075817 Chr.8: 96160581-96160581 on Build GRCh38
rs574711442 Chr.8: 96160465-96160465 on Build GRCh38
rs930635526 Chr.8: 96160432-96160432 on Build GRCh38
rs761442119 Chr.8: 96160727-96160727 on Build GRCh38
rs121909354 Chr.8: 96160568-96160568 on Build GRCh38
rs372946584 Chr.8: 96160574-96160574 on Build GRCh38
rs143020575 Chr.8: 96160603-96160603 on Build GRCh38
rs368498747 Chr.8: 96160371-96160371 on Build GRCh38
rs776298591 Chr.8: 96160618-96160618 on Build GRCh38
rs774264491 Chr.8: 96160452-96160452 on Build GRCh38
rs753326851 Chr.8: 96160536-96160536 on Build GRCh38
rs200398313 Chr.8: 96160420-96160420 on Build GRCh38
rs755403035 Chr.8: 96160506-96160506 on Build GRCh38
rs779321709 Chr.8: 96160397-96160397 on Build GRCh38
rs543473222 Chr.8: 96160478-96160478 on Build GRCh38
rs745979763 Chr.8: 96160675-96160675 on Build GRCh38
rs988646683 Chr.8: 96160448-96160448 on Build GRCh38
rs369859861 Chr.8: 96160669-96160669 on Build GRCh38
rs368712395 Chr.8: 96160513-96160513 on Build GRCh38
rs907889569 Chr.8: 96160771-96160771 on Build GRCh38
rs759656984 Chr.8: 96160714-96160714 on Build GRCh38
rs751465124 Chr.8: 96160377-96160377 on Build GRCh38
rs768986192 Chr.8: 96160354-96160354 on Build GRCh38
rs748500252 Chr.8: 96160569-96160569 on Build GRCh38
rs922215713 Chr.8: 96160791-96160791 on Build GRCh38
rs976341314 Chr.8: 96160789-96160789 on Build GRCh38
rs750416057 Chr.8: 96160360-96160360 on Build GRCh38
rs1012074147 Chr.8: 96160685-96160685 on Build GRCh38
rs760516509 Chr.8: 96160611-96160611 on Build GRCh38
rs781205930 Chr.8: 96160507-96160507 on Build GRCh38
rs767552315 Chr.8: 96160444-96160458 on Build GRCh38
rs397514725 Chr.8: 96160524-96160524 on Build GRCh38
rs762390186 Chr.8: 96160744-96160744 on Build GRCh38
rs778654954 Chr.8: 96160594-96160594 on Build GRCh38
rs575981211 Chr.8: 96160358-96160358 on Build GRCh38
rs769923080 Chr.8: 96160426-96160426 on Build GRCh38
rs753416812 Chr.8: 96160474-96160474 on Build GRCh38
rs375724225 Chr.8: 96160632-96160632 on Build GRCh38
rs35554903 Chr.8: 96160463-96160463 on Build GRCh38
rs756577201 Chr.8: 96160660-96160660 on Build GRCh38
rs367883918 Chr.8: 96160412-96160412 on Build GRCh38
rs375108591 Chr.8: 96160526-96160526 on Build GRCh38
rs536459986 Chr.8: 96160736-96160736 on Build GRCh38
rs978501950 Chr.8: 96160770-96160770 on Build GRCh38
rs980017956 Chr.8: 96160392-96160392 on Build GRCh38
rs767034161 Chr.8: 96160460-96160460 on Build GRCh38
rs770207817 Chr.8: 96160511-96160511 on Build GRCh38
rs770941597 Chr.8: 96160520-96160520 on Build GRCh38
rs112296824 Chr.8: 96160438-96160438 on Build GRCh38
rs900181760 Chr.8: 96160527-96160527 on Build GRCh38
rs766458579 Chr.8: 96160440-96160440 on Build GRCh38
rs756784754 Chr.8: 96160543-96160543 on Build GRCh38
rs983830536 Chr.8: 96160760-96160760 on Build GRCh38
rs563637313 Chr.8: 96160484-96160484 on Build GRCh38
rs947153209 Chr.8: 96160456-96160456 on Build GRCh38
rs773393980 Chr.8: 96160512-96160512 on Build GRCh38
rs913074522 Chr.8: 96160752-96160752 on Build GRCh38
rs770548453 Chr.8: 96160447-96160447 on Build GRCh38
rs141468446 Chr.8: 96160592-96160594 on Build GRCh38
rs769371013 Chr.8: 96160734-96160734 on Build GRCh38
rs200796417 Chr.8: 96160719-96160719 on Build GRCh38
rs752395845 Chr.8: 96160464-96160464 on Build GRCh38
rs746768308 Chr.8: 96160718-96160718 on Build GRCh38
rs367719359 Chr.8: 96160621-96160621 on Build GRCh38
rs764817524 Chr.8: 96160630-96160630 on Build GRCh38
rs527612795 Chr.8: 96160598-96160598 on Build GRCh38
rs947171573 Chr.8: 96160761-96160761 on Build GRCh38
rs369474176 Chr.8: 96160534-96160534 on Build GRCh38
rs143941048 Chr.8: 96160532-96160532 on Build GRCh38
rs776208705 Chr.8: 96160723-96160723 on Build GRCh38
rs1040541530 Chr.8: 96160470-96160470 on Build GRCh38
rs756381156 Chr.8: 96160570-96160570 on Build GRCh38
rs752112002 Chr.8: 96160563-96160563 on Build GRCh38
rs762770107 Chr.8: 96160364-96160364 on Build GRCh38
rs371969433 Chr.8: 96160525-96160525 on Build GRCh38
rs148321868 Chr.8: 96160443-96160443 on Build GRCh38
rs866512501 Chr.8: 96160665-96160665 on Build GRCh38
rs374850065 Chr.8: 96160384-96160384 on Build GRCh38
rs878853102 Chr.8: 96160620-96160620 on Build GRCh38
rs763584976 Chr.8: 96160468-96160468 on Build GRCh38
rs749337343 Chr.8: 96160578-96160578 on Build GRCh38
rs759395579 Chr.8: 96160455-96160455 on Build GRCh38
rs746383854 Chr.8: 96160398-96160398 on Build GRCh38

View Full Product Details