Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18474001-18474496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1569332 c.27T>A p.P9P Chr.9: 18474259-18474259 on Build GRCh38
98323 c.14G>A p.R5H Chr.9: 18474246-18474246 on Build GRCh38
1244414 c.58C>A p.L20I Chr.9: 18474290-18474290 on Build GRCh38
5077404 c.27T>A p.P9P Chr.9: 18474259-18474259 on Build GRCh38
1648144 c.48G>T p.L16L Chr.9: 18474280-18474280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931231763 Chr.9: 18474353-18474353 on Build GRCh38
rs922482212 Chr.9: 18474171-18474171 on Build GRCh38
rs146667173 Chr.9: 18474429-18474429 on Build GRCh38
rs751081301 Chr.9: 18474377-18474378 on Build GRCh38
rs569428018 Chr.9: 18474206-18474206 on Build GRCh38
rs1014169723 Chr.9: 18474281-18474281 on Build GRCh38
rs758597483 Chr.9: 18474460-18474460 on Build GRCh38
rs1034538358 Chr.9: 18474407-18474407 on Build GRCh38
rs573128155 Chr.9: 18474393-18474393 on Build GRCh38
rs1039613722 Chr.9: 18474340-18474340 on Build GRCh38
rs371444541 Chr.9: 18474045-18474045 on Build GRCh38
rs533986798 Chr.9: 18474415-18474415 on Build GRCh38
rs912446536 Chr.9: 18474048-18474048 on Build GRCh38
rs1004852986 Chr.9: 18474080-18474080 on Build GRCh38
rs555573459 Chr.9: 18474424-18474424 on Build GRCh38
rs865940342 Chr.9: 18474152-18474152 on Build GRCh38
rs146877500 Chr.9: 18474376-18474377 on Build GRCh38
rs1049151697 Chr.9: 18474069-18474069 on Build GRCh38
rs374239134 Chr.9: 18474252-18474252 on Build GRCh38
rs942585439 Chr.9: 18474052-18474052 on Build GRCh38
rs537184899 Chr.9: 18474320-18474320 on Build GRCh38
rs1005067778 Chr.9: 18474354-18474355 on Build GRCh38
rs976533943 Chr.9: 18474367-18474367 on Build GRCh38
rs780100352 Chr.9: 18474190-18474190 on Build GRCh38
rs754441702 Chr.9: 18474199-18474199 on Build GRCh38
rs1024488680 Chr.9: 18474059-18474059 on Build GRCh38
rs771747649 Chr.9: 18474215-18474215 on Build GRCh38
rs756090965 Chr.9: 18474060-18474060 on Build GRCh38
rs373998511 Chr.9: 18474279-18474279 on Build GRCh38
rs535381575 Chr.9: 18474067-18474067 on Build GRCh38
rs552875484 Chr.9: 18474066-18474066 on Build GRCh38
rs546753448 Chr.9: 18474218-18474218 on Build GRCh38
rs754954111 Chr.9: 18474287-18474287 on Build GRCh38
rs530798439 Chr.9: 18474035-18474035 on Build GRCh38
rs547752588 Chr.9: 18474043-18474043 on Build GRCh38
rs528666817 Chr.9: 18474141-18474141 on Build GRCh38
rs151261509 Chr.9: 18474355-18474356 on Build GRCh38
rs752229352 Chr.9: 18474113-18474113 on Build GRCh38
rs770158093 Chr.9: 18474324-18474324 on Build GRCh38
rs774622455 Chr.9: 18474249-18474249 on Build GRCh38
rs886526833 Chr.9: 18474079-18474079 on Build GRCh38
rs10963614 Chr.9: 18474359-18474359 on Build GRCh38
rs182464276 Chr.9: 18474365-18474365 on Build GRCh38
rs535845515 Chr.9: 18474041-18474041 on Build GRCh38
rs534479187 Chr.9: 18474375-18474375 on Build GRCh38
rs971464127 Chr.9: 18474092-18474092 on Build GRCh38
rs377361900 Chr.9: 18474391-18474392 on Build GRCh38
rs771087258 Chr.9: 18474328-18474329 on Build GRCh38
rs746301527 Chr.9: 18474229-18474229 on Build GRCh38
rs1020863094 Chr.9: 18474445-18474445 on Build GRCh38
rs952268765 Chr.9: 18474111-18474111 on Build GRCh38
rs765959133 Chr.9: 18474027-18474027 on Build GRCh38
rs111868972 Chr.9: 18474187-18474187 on Build GRCh38
rs752271549 Chr.9: 18474294-18474294 on Build GRCh38
rs368969738 Chr.9: 18474289-18474289 on Build GRCh38
rs185805899 Chr.9: 18474385-18474385 on Build GRCh38
rs904392927 Chr.9: 18474023-18474023 on Build GRCh38
rs573859470 Chr.9: 18474425-18474425 on Build GRCh38
rs976416215 Chr.9: 18474155-18474155 on Build GRCh38
rs990294526 Chr.9: 18474181-18474181 on Build GRCh38
rs1011665244 Chr.9: 18474436-18474436 on Build GRCh38
rs767851550 Chr.9: 18474255-18474255 on Build GRCh38
rs750277177 Chr.9: 18474186-18474186 on Build GRCh38
rs756005447 Chr.9: 18474189-18474189 on Build GRCh38
rs567827769 Chr.9: 18474243-18474243 on Build GRCh38
rs749240332 Chr.9: 18474194-18474194 on Build GRCh38
rs201675556 Chr.9: 18474278-18474278 on Build GRCh38
rs891785697 Chr.9: 18474042-18474042 on Build GRCh38
rs766271459 Chr.9: 18474182-18474182 on Build GRCh38
rs371051447 Chr.9: 18474318-18474318 on Build GRCh38
rs778444440 Chr.9: 18474436-18474437 on Build GRCh38
rs980413934 Chr.9: 18474072-18474072 on Build GRCh38
rs1001726104 Chr.9: 18474103-18474103 on Build GRCh38
rs777786817 Chr.9: 18474186-18474187 on Build GRCh38
rs952281024 Chr.9: 18474354-18474354 on Build GRCh38
rs747258344 Chr.9: 18474227-18474228 on Build GRCh38
rs780306246 Chr.9: 18474326-18474326 on Build GRCh38
rs907202749 Chr.9: 18474387-18474387 on Build GRCh38
rs143550681 Chr.9: 18474286-18474286 on Build GRCh38
rs942366744 Chr.9: 18474336-18474336 on Build GRCh38
rs957503954 Chr.9: 18474174-18474174 on Build GRCh38
rs41305317 Chr.9: 18474356-18474356 on Build GRCh38
rs13286264 Chr.9: 18474282-18474282 on Build GRCh38
rs770065912 Chr.9: 18474230-18474230 on Build GRCh38
rs201842948 Chr.9: 18474266-18474266 on Build GRCh38
rs750175515 Chr.9: 18474256-18474256 on Build GRCh38
rs140413273 Chr.9: 18474248-18474248 on Build GRCh38
rs757973400 Chr.9: 18474301-18474301 on Build GRCh38
rs898313331 Chr.9: 18474064-18474064 on Build GRCh38
rs908304194 Chr.9: 18474126-18474126 on Build GRCh38
rs775997285 Chr.9: 18474239-18474239 on Build GRCh38
rs935879505 Chr.9: 18474223-18474223 on Build GRCh38
rs747773383 Chr.9: 18474211-18474211 on Build GRCh38
rs931096181 Chr.9: 18474350-18474350 on Build GRCh38
rs776988093 Chr.9: 18474329-18474330 on Build GRCh38
rs750706593 Chr.9: 18474433-18474433 on Build GRCh38
rs1024721339 Chr.9: 18474090-18474090 on Build GRCh38
rs769326548 Chr.9: 18474346-18474346 on Build GRCh38
rs77805914 Chr.9: 18474342-18474342 on Build GRCh38
rs961006551 Chr.9: 18474137-18474137 on Build GRCh38

View Full Product Details