Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35814763-35815300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HINT2
Entrez Gene ID
84681
Gene Name
histidine triad nucleotide binding protein 2
Gene Aliases
HIT-17
UniGene
Hs.70573
Gene Chromosome Location
Chr.9: 35812960-35815045 on Build GRCh38
Transcripts
NM_032593.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4801136 c.72C>A p.R24R Chr.9: 35814908-35814908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770125020 Chr.9: 35814797-35814797 on Build GRCh38
rs867048211 Chr.9: 35815032-35815032 on Build GRCh38
rs755091743 Chr.9: 35815021-35815021 on Build GRCh38
rs776216921 Chr.9: 35814939-35814939 on Build GRCh38
rs761886127 Chr.9: 35815058-35815058 on Build GRCh38
rs1028617349 Chr.9: 35814990-35814990 on Build GRCh38
rs768337196 Chr.9: 35814924-35814924 on Build GRCh38
rs1054979969 Chr.9: 35815101-35815101 on Build GRCh38
rs767574948 Chr.9: 35815188-35815188 on Build GRCh38
rs932764661 Chr.9: 35815140-35815140 on Build GRCh38
rs768889507 Chr.9: 35814859-35814859 on Build GRCh38
rs1035472447 Chr.9: 35815189-35815189 on Build GRCh38
rs903240674 Chr.9: 35814951-35814951 on Build GRCh38
rs990731885 Chr.9: 35814908-35814908 on Build GRCh38
rs990615744 Chr.9: 35815056-35815056 on Build GRCh38
rs997091467 Chr.9: 35814977-35814977 on Build GRCh38
rs749688731 Chr.9: 35814860-35814860 on Build GRCh38
rs771623710 Chr.9: 35814869-35814869 on Build GRCh38
rs893299819 Chr.9: 35815109-35815109 on Build GRCh38
rs893321891 Chr.9: 35814969-35814969 on Build GRCh38
rs1023613529 Chr.9: 35815089-35815089 on Build GRCh38
rs910564552 Chr.9: 35815037-35815037 on Build GRCh38
rs760934399 Chr.9: 35815197-35815197 on Build GRCh38
rs919874483 Chr.9: 35814963-35814963 on Build GRCh38
rs533904224 Chr.9: 35815006-35815006 on Build GRCh38
rs981205403 Chr.9: 35814958-35814958 on Build GRCh38
rs1011682441 Chr.9: 35814888-35814888 on Build GRCh38
rs1049929428 Chr.9: 35815175-35815175 on Build GRCh38
rs1011845694 Chr.9: 35815081-35815081 on Build GRCh38
rs931225472 Chr.9: 35815007-35815007 on Build GRCh38
rs934871532 Chr.9: 35815064-35815064 on Build GRCh38
rs750384187 Chr.9: 35815228-35815228 on Build GRCh38
rs961426237 Chr.9: 35814839-35814839 on Build GRCh38
rs563336642 Chr.9: 35814831-35814831 on Build GRCh38
rs143962419 Chr.9: 35814818-35814818 on Build GRCh38
rs556860912 Chr.9: 35815179-35815179 on Build GRCh38
rs959237958 Chr.9: 35815046-35815046 on Build GRCh38
rs766338678 Chr.9: 35814998-35814998 on Build GRCh38
rs570547188 Chr.9: 35815068-35815068 on Build GRCh38
rs919807208 Chr.9: 35814784-35814784 on Build GRCh38
rs1049054619 Chr.9: 35814795-35814795 on Build GRCh38
rs1045719262 Chr.9: 35814941-35814941 on Build GRCh38
rs1053198415 Chr.9: 35815066-35815066 on Build GRCh38
rs774920159 Chr.9: 35814893-35814893 on Build GRCh38
rs961207236 Chr.9: 35814960-35814960 on Build GRCh38
rs973904531 Chr.9: 35814843-35814843 on Build GRCh38
rs1040414642 Chr.9: 35814806-35814806 on Build GRCh38
rs866974601 Chr.9: 35814955-35814955 on Build GRCh38
rs915910864 Chr.9: 35814929-35814929 on Build GRCh38
rs372932465 Chr.9: 35814804-35814804 on Build GRCh38
rs1022617357 Chr.9: 35814904-35814904 on Build GRCh38
rs961128449 Chr.9: 35815022-35815022 on Build GRCh38
rs1040445448 Chr.9: 35815055-35815055 on Build GRCh38
rs150674252 Chr.9: 35815225-35815225 on Build GRCh38
rs768013756 Chr.9: 35814902-35814902 on Build GRCh38
rs1015687408 Chr.9: 35815010-35815010 on Build GRCh38
rs775895197 Chr.9: 35814798-35814798 on Build GRCh38
rs985224047 Chr.9: 35815009-35815009 on Build GRCh38
rs961111536 Chr.9: 35815193-35815193 on Build GRCh38
rs375867774 Chr.9: 35814959-35814959 on Build GRCh38
rs952623858 Chr.9: 35815276-35815276 on Build GRCh38
rs902024870 Chr.9: 35814814-35814814 on Build GRCh38
rs773216742 Chr.9: 35814961-35814961 on Build GRCh38
rs893408470 Chr.9: 35815072-35815072 on Build GRCh38
rs746524360 Chr.9: 35814914-35814914 on Build GRCh38
rs529090876 Chr.9: 35814846-35814846 on Build GRCh38
rs867310069 Chr.9: 35815207-35815207 on Build GRCh38
rs762859975 Chr.9: 35814996-35814996 on Build GRCh38
rs539736529 Chr.9: 35815114-35815114 on Build GRCh38
rs893386428 Chr.9: 35814870-35814870 on Build GRCh38
rs981386779 Chr.9: 35815084-35815084 on Build GRCh38
rs761628658 Chr.9: 35814957-35814957 on Build GRCh38
rs930669873 Chr.9: 35814785-35814785 on Build GRCh38
rs934747557 Chr.9: 35815092-35815099 on Build GRCh38
rs972583073 Chr.9: 35815214-35815214 on Build GRCh38
rs950015418 Chr.9: 35814933-35814933 on Build GRCh38
rs192948763 Chr.9: 35815045-35815045 on Build GRCh38
rs905529676 Chr.9: 35815120-35815120 on Build GRCh38
rs568233543 Chr.9: 35814994-35814994 on Build GRCh38
rs901930043 Chr.9: 35815060-35815060 on Build GRCh38
rs774516417 Chr.9: 35815016-35815016 on Build GRCh38
rs937931361 Chr.9: 35814900-35814900 on Build GRCh38

View Full Product Details