Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215005457-215005960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK2
Entrez Gene ID
3776
Gene Name
potassium two pore domain channel subfamily K member 2
Gene Aliases
K2p2.1, TPKC1, TREK, TREK-1, TREK1, hTREK-1c, hTREK-1e
UniGene
Hs.497745
Gene Chromosome Location
Chr.1: 215002920-215237093 on Build GRCh38
Transcripts
NM_001017424.2 XM_017001248.1 XM_017001249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4028033 c.27T>C p.D9D Chr.1: 215005948-215005948 on Build GRCh38
7373148 c.25G>T p.D9Y Chr.1: 215005946-215005946 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761578460 Chr.1: 215005628-215005628 on Build GRCh38
rs1027308102 Chr.1: 215005498-215005498 on Build GRCh38
rs981321479 Chr.1: 215005574-215005574 on Build GRCh38
rs750854866 Chr.1: 215005892-215005892 on Build GRCh38
rs556853615 Chr.1: 215005511-215005511 on Build GRCh38
rs559792291 Chr.1: 215005566-215005566 on Build GRCh38
rs1005946127 Chr.1: 215005770-215005770 on Build GRCh38
rs34015427 Chr.1: 215005512-215005512 on Build GRCh38
rs765014119 Chr.1: 215005675-215005675 on Build GRCh38
rs756693706 Chr.1: 215005907-215005907 on Build GRCh38
rs139798249 Chr.1: 215005897-215005897 on Build GRCh38
rs7538655 Chr.1: 215005481-215005481 on Build GRCh38
rs895288637 Chr.1: 215005823-215005823 on Build GRCh38
rs989771087 Chr.1: 215005565-215005565 on Build GRCh38
rs780223600 Chr.1: 215005918-215005918 on Build GRCh38
rs775768312 Chr.1: 215005567-215005567 on Build GRCh38
rs1006737529 Chr.1: 215005773-215005773 on Build GRCh38
rs886125397 Chr.1: 215005755-215005755 on Build GRCh38
rs933234394 Chr.1: 215005752-215005752 on Build GRCh38
rs149894109 Chr.1: 215005594-215005594 on Build GRCh38
rs1005978832 Chr.1: 215005489-215005489 on Build GRCh38
rs928091740 Chr.1: 215005614-215005614 on Build GRCh38
rs551202985 Chr.1: 215005800-215005800 on Build GRCh38
rs187235423 Chr.1: 215005849-215005849 on Build GRCh38
rs1018052067 Chr.1: 215005515-215005515 on Build GRCh38
rs755245091 Chr.1: 215005932-215005932 on Build GRCh38
rs562747900 Chr.1: 215005719-215005719 on Build GRCh38
rs749435660 Chr.1: 215005921-215005921 on Build GRCh38
rs943245271 Chr.1: 215005687-215005687 on Build GRCh38
rs989595371 Chr.1: 215005673-215005673 on Build GRCh38
rs145974725 Chr.1: 215005887-215005887 on Build GRCh38
rs542933442 Chr.1: 215005551-215005551 on Build GRCh38
rs936717194 Chr.1: 215005661-215005661 on Build GRCh38
rs565413423 Chr.1: 215005707-215005707 on Build GRCh38
rs888346040 Chr.1: 215005765-215005765 on Build GRCh38
rs1048327344 Chr.1: 215005744-215005744 on Build GRCh38
rs965096015 Chr.1: 215005523-215005523 on Build GRCh38
rs567512690 Chr.1: 215005816-215005816 on Build GRCh38
rs1039918679 Chr.1: 215005835-215005835 on Build GRCh38
rs1039344018 Chr.1: 215005725-215005725 on Build GRCh38
rs1016047248 Chr.1: 215005777-215005777 on Build GRCh38
rs542960984 Chr.1: 215005627-215005627 on Build GRCh38
rs767900074 Chr.1: 215005544-215005544 on Build GRCh38
rs918200311 Chr.1: 215005564-215005564 on Build GRCh38
rs978313885 Chr.1: 215005898-215005898 on Build GRCh38
rs757603577 Chr.1: 215005877-215005877 on Build GRCh38
rs530888335 Chr.1: 215005742-215005742 on Build GRCh38
rs78980985 Chr.1: 215005874-215005874 on Build GRCh38
rs956352631 Chr.1: 215005557-215005557 on Build GRCh38
rs76740997 Chr.1: 215005591-215005591 on Build GRCh38
rs530797734 Chr.1: 215005788-215005788 on Build GRCh38
rs747949311 Chr.1: 215005935-215005935 on Build GRCh38
rs528446708 Chr.1: 215005570-215005570 on Build GRCh38
rs752635434 Chr.1: 215005635-215005635 on Build GRCh38

View Full Product Details