Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236686211-236686748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN2
Entrez Gene ID
88
Gene Name
actinin alpha 2
Gene Aliases
CMD1AA, CMH23
UniGene
Hs.498178
Gene Chromosome Location
Chr.1: 236686454-236764631 on Build GRCh38
Transcripts
NM_001103.3 NM_001278343.1 NM_001278344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7110531 c.63C>G p.I21M Chr.1: 236686736-236686736 on Build GRCh38
6429997 c.26A>G p.Q9R Chr.1: 236686699-236686699 on Build GRCh38
4659140 c.45C>T p.D15D Chr.1: 236686718-236686718 on Build GRCh38
4659137 c.43G>A p.D15N Chr.1: 236686716-236686716 on Build GRCh38
6456913 c.61A>C p.I21L Chr.1: 236686734-236686734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025891738 Chr.1: 236686443-236686443 on Build GRCh38
rs561114298 Chr.1: 236686505-236686505 on Build GRCh38
rs1025690790 Chr.1: 236686526-236686526 on Build GRCh38
rs373785727 Chr.1: 236686653-236686653 on Build GRCh38
rs377343516 Chr.1: 236686641-236686641 on Build GRCh38
rs775052416 Chr.1: 236686719-236686719 on Build GRCh38
rs886046205 Chr.1: 236686704-236686704 on Build GRCh38
rs867323461 Chr.1: 236686426-236686426 on Build GRCh38
rs771424965 Chr.1: 236686718-236686718 on Build GRCh38
rs935647251 Chr.1: 236686478-236686478 on Build GRCh38
rs775224980 Chr.1: 236686650-236686650 on Build GRCh38
rs781720338 Chr.1: 236686685-236686685 on Build GRCh38
rs979876881 Chr.1: 236686374-236686374 on Build GRCh38
rs758365969 Chr.1: 236686666-236686666 on Build GRCh38
rs980228032 Chr.1: 236686477-236686477 on Build GRCh38
rs867320952 Chr.1: 236686354-236686354 on Build GRCh38
rs896984229 Chr.1: 236686307-236686307 on Build GRCh38
rs891736706 Chr.1: 236686404-236686404 on Build GRCh38
rs951644250 Chr.1: 236686451-236686451 on Build GRCh38
rs901340700 Chr.1: 236686571-236686572 on Build GRCh38
rs762061889 Chr.1: 236686658-236686658 on Build GRCh38
rs1054274216 Chr.1: 236686386-236686386 on Build GRCh38
rs374500536 Chr.1: 236686640-236686640 on Build GRCh38
rs886046201 Chr.1: 236686515-236686515 on Build GRCh38
rs898467020 Chr.1: 236686441-236686441 on Build GRCh38
rs1018374700 Chr.1: 236686429-236686429 on Build GRCh38
rs749017788 Chr.1: 236686623-236686623 on Build GRCh38
rs1036856545 Chr.1: 236686303-236686303 on Build GRCh38
rs370646112 Chr.1: 236686667-236686667 on Build GRCh38
rs121434525 Chr.1: 236686699-236686699 on Build GRCh38
rs995582459 Chr.1: 236686312-236686312 on Build GRCh38
rs969935283 Chr.1: 236686559-236686559 on Build GRCh38
rs935690753 Chr.1: 236686381-236686381 on Build GRCh38
rs942568236 Chr.1: 236686501-236686501 on Build GRCh38
rs397516580 Chr.1: 236686721-236686721 on Build GRCh38
rs552471202 Chr.1: 236686572-236686578 on Build GRCh38
rs1018653161 Chr.1: 236686270-236686271 on Build GRCh38
rs969590464 Chr.1: 236686470-236686470 on Build GRCh38
rs887110909 Chr.1: 236686530-236686530 on Build GRCh38
rs527465278 Chr.1: 236686293-236686294 on Build GRCh38
rs755508640 Chr.1: 236686679-236686679 on Build GRCh38
rs532698332 Chr.1: 236686665-236686665 on Build GRCh38
rs201920417 Chr.1: 236686671-236686671 on Build GRCh38
rs545730306 Chr.1: 236686255-236686255 on Build GRCh38
rs1007353035 Chr.1: 236686428-236686428 on Build GRCh38
rs1025106342 Chr.1: 236686467-236686467 on Build GRCh38
rs886046204 Chr.1: 236686590-236686596 on Build GRCh38
rs896386651 Chr.1: 236686263-236686263 on Build GRCh38
rs1025921138 Chr.1: 236686332-236686332 on Build GRCh38
rs1025237694 Chr.1: 236686556-236686556 on Build GRCh38
rs995501541 Chr.1: 236686519-236686519 on Build GRCh38
rs962750932 Chr.1: 236686284-236686285 on Build GRCh38
rs551141480 Chr.1: 236686695-236686695 on Build GRCh38
rs995532810 Chr.1: 236686442-236686442 on Build GRCh38
rs1002398063 Chr.1: 236686571-236686571 on Build GRCh38
rs989770685 Chr.1: 236686588-236686588 on Build GRCh38
rs200003968 Chr.1: 236686631-236686631 on Build GRCh38
rs543054770 Chr.1: 236686498-236686498 on Build GRCh38
rs957177731 Chr.1: 236686580-236686580 on Build GRCh38
rs937691869 Chr.1: 236686258-236686258 on Build GRCh38
rs974701527 Chr.1: 236686325-236686325 on Build GRCh38
rs886046203 Chr.1: 236686596-236686597 on Build GRCh38
rs753623943 Chr.1: 236686657-236686657 on Build GRCh38
rs949879767 Chr.1: 236686370-236686370 on Build GRCh38
rs924925281 Chr.1: 236686257-236686257 on Build GRCh38
rs528593690 Chr.1: 236686508-236686508 on Build GRCh38
rs886046202 Chr.1: 236686570-236686570 on Build GRCh38
rs575984942 Chr.1: 236686356-236686356 on Build GRCh38
rs946005661 Chr.1: 236686497-236686497 on Build GRCh38
rs763476785 Chr.1: 236686630-236686635 on Build GRCh38
rs557720359 Chr.1: 236686316-236686316 on Build GRCh38
rs750521308 Chr.1: 236686659-236686659 on Build GRCh38
rs992327109 Chr.1: 236686466-236686466 on Build GRCh38
rs916995764 Chr.1: 236686367-236686367 on Build GRCh38
rs1050918506 Chr.1: 236686418-236686423 on Build GRCh38
rs949366353 Chr.1: 236686278-236686278 on Build GRCh38
rs368367224 Chr.1: 236686691-236686691 on Build GRCh38
rs1040174989 Chr.1: 236686503-236686503 on Build GRCh38
rs1013809530 Chr.1: 236686541-236686541 on Build GRCh38
rs989803409 Chr.1: 236686490-236686490 on Build GRCh38
rs367979371 Chr.1: 236686656-236686656 on Build GRCh38
rs771008376 Chr.1: 236686714-236686716 on Build GRCh38
rs945870177 Chr.1: 236686417-236686417 on Build GRCh38
rs912859442 Chr.1: 236686634-236686634 on Build GRCh38
rs1054747023 Chr.1: 236686261-236686261 on Build GRCh38
rs979592721 Chr.1: 236686236-236686236 on Build GRCh38
rs138279482 Chr.1: 236686652-236686652 on Build GRCh38
rs531612546 Chr.1: 236686300-236686300 on Build GRCh38
rs34326848 Chr.1: 236686314-236686314 on Build GRCh38
rs926916683 Chr.1: 236686381-236686381 on Build GRCh38
rs772077859 Chr.1: 236686645-236686645 on Build GRCh38
rs913142084 Chr.1: 236686493-236686493 on Build GRCh38

View Full Product Details