Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236731145-236731659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTN2
Entrez Gene ID
88
Gene Name
actinin alpha 2
Gene Aliases
CMD1AA, CMH23
UniGene
Hs.498178
Gene Chromosome Location
Chr.1: 236686454-236764631 on Build GRCh38
Transcripts
NM_001103.3 NM_001278343.1 NM_001278344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5832527 c.697G>T p.D233Y Chr.1: 236731314-236731314 on Build GRCh38
6062092 c.664C>A p.H222N Chr.1: 236731281-236731281 on Build GRCh38
3485646 c.616G>A p.D206N Chr.1: 236731233-236731233 on Build GRCh38
6209973 c.655G>A p.A219T Chr.1: 236731272-236731272 on Build GRCh38
680004 c.669G>T p.L223L Chr.1: 236731286-236731286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773185359 Chr.1: 236731354-236731354 on Build GRCh38
rs750480942 Chr.1: 236731219-236731219 on Build GRCh38
rs776401740 Chr.1: 236731343-236731343 on Build GRCh38
rs892152228 Chr.1: 236731366-236731366 on Build GRCh38
rs563385787 Chr.1: 236731607-236731607 on Build GRCh38
rs1016533556 Chr.1: 236731451-236731451 on Build GRCh38
rs184754556 Chr.1: 236731417-236731417 on Build GRCh38
rs192122725 Chr.1: 236731515-236731515 on Build GRCh38
rs370725711 Chr.1: 236731182-236731182 on Build GRCh38
rs565935053 Chr.1: 236731211-236731211 on Build GRCh38
rs139489232 Chr.1: 236731307-236731307 on Build GRCh38
rs819640 Chr.1: 236731347-236731347 on Build GRCh38
rs939646272 Chr.1: 236731196-236731196 on Build GRCh38
rs758419978 Chr.1: 236731255-236731255 on Build GRCh38
rs778336485 Chr.1: 236731284-236731284 on Build GRCh38
rs141793230 Chr.1: 236731303-236731303 on Build GRCh38
rs1008850231 Chr.1: 236731413-236731413 on Build GRCh38
rs963230056 Chr.1: 236731557-236731557 on Build GRCh38
rs112324898 Chr.1: 236731316-236731316 on Build GRCh38
rs972155342 Chr.1: 236731563-236731563 on Build GRCh38
rs751979954 Chr.1: 236731246-236731246 on Build GRCh38
rs760420624 Chr.1: 236731186-236731186 on Build GRCh38
rs749019847 Chr.1: 236731263-236731263 on Build GRCh38
rs919315956 Chr.1: 236731566-236731566 on Build GRCh38
rs756903825 Chr.1: 236731268-236731268 on Build GRCh38
rs762058124 Chr.1: 236731344-236731344 on Build GRCh38
rs766810456 Chr.1: 236731365-236731365 on Build GRCh38
rs111464645 Chr.1: 236731230-236731230 on Build GRCh38
rs794728962 Chr.1: 236731237-236731237 on Build GRCh38
rs116526937 Chr.1: 236731426-236731426 on Build GRCh38
rs377331254 Chr.1: 236731367-236731367 on Build GRCh38
rs377629538 Chr.1: 236731405-236731405 on Build GRCh38
rs889032976 Chr.1: 236731406-236731406 on Build GRCh38
rs758272111 Chr.1: 236731213-236731216 on Build GRCh38
rs765856578 Chr.1: 236731218-236731218 on Build GRCh38
rs369064035 Chr.1: 236731362-236731362 on Build GRCh38
rs904672923 Chr.1: 236731314-236731314 on Build GRCh38
rs764302979 Chr.1: 236731198-236731198 on Build GRCh38
rs768496700 Chr.1: 236731327-236731327 on Build GRCh38
rs763025229 Chr.1: 236731359-236731359 on Build GRCh38
rs148048699 Chr.1: 236731234-236731234 on Build GRCh38
rs786205144 Chr.1: 236731300-236731300 on Build GRCh38
rs992792617 Chr.1: 236731607-236731608 on Build GRCh38
rs781683654 Chr.1: 236731262-236731262 on Build GRCh38
rs1036498027 Chr.1: 236731334-236731334 on Build GRCh38
rs139694189 Chr.1: 236731633-236731633 on Build GRCh38
rs113669376 Chr.1: 236731315-236731315 on Build GRCh38
rs754001787 Chr.1: 236731202-236731202 on Build GRCh38
rs545283568 Chr.1: 236731581-236731581 on Build GRCh38
rs926838777 Chr.1: 236731189-236731189 on Build GRCh38
rs558330657 Chr.1: 236731259-236731259 on Build GRCh38
rs746819290 Chr.1: 236731319-236731319 on Build GRCh38
rs980737536 Chr.1: 236731620-236731620 on Build GRCh38
rs949012591 Chr.1: 236731616-236731616 on Build GRCh38
rs189638338 Chr.1: 236731418-236731418 on Build GRCh38
rs12723093 Chr.1: 236731612-236731612 on Build GRCh38
rs539077634 Chr.1: 236731227-236731227 on Build GRCh38
rs369957871 Chr.1: 236731209-236731209 on Build GRCh38

View Full Product Details