Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72101874-72102421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC1
Entrez Gene ID
51008
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
ASC1p50, CGI-18, SMABF2, p50
UniGene
Hs.500007
Gene Chromosome Location
Chr.10: 72096032-72217134 on Build GRCh38
Transcripts
NM_001198798.2 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016296.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1 XM_017016303.1

SNP Detail

SNP ID Chromosome Location
rs1007872711 Chr.10: 72102009-72102009 on Build GRCh38
rs1011345373 Chr.10: 72102291-72102291 on Build GRCh38
rs1022983752 Chr.10: 72102392-72102392 on Build GRCh38
rs541291811 Chr.10: 72102092-72102092 on Build GRCh38
rs756798269 Chr.10: 72102304-72102304 on Build GRCh38
rs368099344 Chr.10: 72101922-72101922 on Build GRCh38
rs748058740 Chr.10: 72102038-72102038 on Build GRCh38
rs991879863 Chr.10: 72102364-72102364 on Build GRCh38
rs922544901 Chr.10: 72101958-72101958 on Build GRCh38
rs565879750 Chr.10: 72102297-72102297 on Build GRCh38
rs530948266 Chr.10: 72102199-72102199 on Build GRCh38
rs1037880893 Chr.10: 72101984-72101984 on Build GRCh38
rs964456277 Chr.10: 72102171-72102171 on Build GRCh38
rs551086250 Chr.10: 72102359-72102359 on Build GRCh38
rs926417713 Chr.10: 72102344-72102344 on Build GRCh38
rs535567132 Chr.10: 72102017-72102017 on Build GRCh38
rs902921511 Chr.10: 72102356-72102356 on Build GRCh38
rs539290594 Chr.10: 72102107-72102126 on Build GRCh38
rs946502628 Chr.10: 72102063-72102063 on Build GRCh38
rs555690069 Chr.10: 72102083-72102083 on Build GRCh38
rs756289547 Chr.10: 72102164-72102164 on Build GRCh38
rs943617691 Chr.10: 72102234-72102234 on Build GRCh38
rs769805829 Chr.10: 72102365-72102365 on Build GRCh38
rs998198772 Chr.10: 72102182-72102182 on Build GRCh38
rs144994153 Chr.10: 72101945-72101945 on Build GRCh38
rs1035629398 Chr.10: 72102134-72102134 on Build GRCh38
rs1051935042 Chr.10: 72102002-72102002 on Build GRCh38
rs1017893425 Chr.10: 72102049-72102049 on Build GRCh38
rs187745294 Chr.10: 72102229-72102229 on Build GRCh38
rs563623963 Chr.10: 72102188-72102188 on Build GRCh38
rs545525504 Chr.10: 72102157-72102157 on Build GRCh38
rs994434940 Chr.10: 72102146-72102146 on Build GRCh38
rs987987145 Chr.10: 72101987-72101987 on Build GRCh38
rs142218806 Chr.10: 72102320-72102320 on Build GRCh38
rs565388793 Chr.10: 72102211-72102211 on Build GRCh38
rs529084715 Chr.10: 72102250-72102250 on Build GRCh38
rs1044858924 Chr.10: 72102078-72102078 on Build GRCh38
rs915877870 Chr.10: 72102149-72102149 on Build GRCh38
rs1006133333 Chr.10: 72102158-72102158 on Build GRCh38
rs991364199 Chr.10: 72102373-72102373 on Build GRCh38
rs149093279 Chr.10: 72102123-72102123 on Build GRCh38
rs775450027 Chr.10: 72102289-72102291 on Build GRCh38
rs561864333 Chr.10: 72102248-72102248 on Build GRCh38
rs1021464018 Chr.10: 72102115-72102115 on Build GRCh38
rs991468695 Chr.10: 72101935-72101935 on Build GRCh38
rs942100667 Chr.10: 72101954-72101954 on Build GRCh38
rs1018737885 Chr.10: 72101923-72101923 on Build GRCh38
rs956671014 Chr.10: 72102221-72102221 on Build GRCh38
rs939028829 Chr.10: 72102084-72102084 on Build GRCh38
rs183151036 Chr.10: 72102151-72102151 on Build GRCh38
rs76563850 Chr.10: 72101901-72101901 on Build GRCh38
rs367640316 Chr.10: 72102251-72102255 on Build GRCh38
rs1668154 Chr.10: 72102274-72102274 on Build GRCh38
rs547036128 Chr.10: 72102003-72102003 on Build GRCh38
rs778815472 Chr.10: 72101985-72101985 on Build GRCh38
rs964118862 Chr.10: 72101932-72101932 on Build GRCh38
rs929591625 Chr.10: 72101993-72101993 on Build GRCh38
rs991519166 Chr.10: 72102148-72102148 on Build GRCh38
rs1018386485 Chr.10: 72102165-72102165 on Build GRCh38
rs772026308 Chr.10: 72102117-72102117 on Build GRCh38

View Full Product Details