Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31433373-31433871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297955 c.389G>T p.G130V Chr.11: 31433503-31433503 on Build GRCh38
5372340 c.325C>T p.P109S Chr.11: 31433567-31433567 on Build GRCh38
1604479 c.334C>T p.H112Y Chr.11: 31433558-31433558 on Build GRCh38
4571314 c.429T>C p.F143F Chr.11: 31433463-31433463 on Build GRCh38
3787044 c.420A>G p.R140R Chr.11: 31433472-31433472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768405132 Chr.11: 31433549-31433549 on Build GRCh38
rs146915142 Chr.11: 31433546-31433546 on Build GRCh38
rs764159033 Chr.11: 31433560-31433560 on Build GRCh38
rs749673092 Chr.11: 31433733-31433733 on Build GRCh38
rs769628686 Chr.11: 31433410-31433410 on Build GRCh38
rs3741028 Chr.11: 31433428-31433428 on Build GRCh38
rs927510118 Chr.11: 31433775-31433775 on Build GRCh38
rs143220698 Chr.11: 31433778-31433780 on Build GRCh38
rs989016651 Chr.11: 31433568-31433568 on Build GRCh38
rs773956162 Chr.11: 31433411-31433412 on Build GRCh38
rs755830243 Chr.11: 31433622-31433622 on Build GRCh38
rs981676944 Chr.11: 31433769-31433769 on Build GRCh38
rs769377770 Chr.11: 31433497-31433497 on Build GRCh38
rs758364525 Chr.11: 31433439-31433439 on Build GRCh38
rs201249598 Chr.11: 31433605-31433605 on Build GRCh38
rs747631328 Chr.11: 31433425-31433425 on Build GRCh38
rs1050927606 Chr.11: 31433453-31433453 on Build GRCh38
rs930039693 Chr.11: 31433828-31433828 on Build GRCh38
rs920899074 Chr.11: 31433743-31433743 on Build GRCh38
rs374392413 Chr.11: 31433603-31433608 on Build GRCh38
rs1047150165 Chr.11: 31433836-31433836 on Build GRCh38
rs956204126 Chr.11: 31433677-31433678 on Build GRCh38
rs939829446 Chr.11: 31433781-31433781 on Build GRCh38
rs1018852066 Chr.11: 31433466-31433466 on Build GRCh38
rs898561348 Chr.11: 31433789-31433789 on Build GRCh38
rs762365367 Chr.11: 31433527-31433527 on Build GRCh38
rs937365616 Chr.11: 31433786-31433786 on Build GRCh38
rs577768226 Chr.11: 31433820-31433820 on Build GRCh38
rs996214263 Chr.11: 31433474-31433474 on Build GRCh38
rs779854656 Chr.11: 31433463-31433463 on Build GRCh38
rs536008398 Chr.11: 31433492-31433492 on Build GRCh38
rs187371367 Chr.11: 31433419-31433419 on Build GRCh38
rs920047409 Chr.11: 31433825-31433825 on Build GRCh38
rs768404781 Chr.11: 31433490-31433490 on Build GRCh38
rs747869422 Chr.11: 31433620-31433620 on Build GRCh38
rs773462108 Chr.11: 31433759-31433759 on Build GRCh38
rs554529749 Chr.11: 31433494-31433494 on Build GRCh38
rs184787746 Chr.11: 31433842-31433842 on Build GRCh38
rs761593248 Chr.11: 31433661-31433661 on Build GRCh38
rs757274950 Chr.11: 31433566-31433566 on Build GRCh38
rs1038774261 Chr.11: 31433797-31433797 on Build GRCh38
rs575990820 Chr.11: 31433512-31433512 on Build GRCh38
rs1022030048 Chr.11: 31433731-31433731 on Build GRCh38
rs754857082 Chr.11: 31433595-31433595 on Build GRCh38
rs975122702 Chr.11: 31433738-31433739 on Build GRCh38
rs1038890625 Chr.11: 31433783-31433783 on Build GRCh38
rs774699376 Chr.11: 31433537-31433537 on Build GRCh38
rs777040082 Chr.11: 31433543-31433545 on Build GRCh38
rs537104972 Chr.11: 31433599-31433604 on Build GRCh38
rs369134104 Chr.11: 31433417-31433417 on Build GRCh38
rs760019177 Chr.11: 31433540-31433540 on Build GRCh38
rs754020699 Chr.11: 31433564-31433564 on Build GRCh38
rs771443829 Chr.11: 31433463-31433465 on Build GRCh38
rs375864882 Chr.11: 31433571-31433571 on Build GRCh38
rs1031802631 Chr.11: 31433625-31433625 on Build GRCh38
rs771525870 Chr.11: 31433529-31433529 on Build GRCh38
rs987884076 Chr.11: 31433683-31433683 on Build GRCh38
rs751524852 Chr.11: 31433579-31433579 on Build GRCh38
rs753805691 Chr.11: 31433438-31433438 on Build GRCh38
rs908417139 Chr.11: 31433778-31433778 on Build GRCh38
rs746866007 Chr.11: 31433473-31433473 on Build GRCh38

View Full Product Details