Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31460406-31460903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMP1L
Entrez Gene ID
196294
Gene Name
inner mitochondrial membrane peptidase subunit 1
Gene Aliases
IMMP1, IMP1, IMP1-LIKE
UniGene
Hs.502223
Gene Chromosome Location
Chr.11: 31432399-31509644 on Build GRCh38
Transcripts
NM_001304274.1 NM_144981.2 XM_005252812.3 XM_011519942.1 XM_011519943.2 XM_011519944.2 XM_011519945.2 XM_011519946.2 XM_011519947.2 XM_017017305.1 XM_017017306.1 XM_017017307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5477550 c.110C>T p.S37F Chr.11: 31460710-31460710 on Build GRCh38
6487694 c.145G>T p.D49Y Chr.11: 31460675-31460675 on Build GRCh38
1604480 c.185G>A p.G62D Chr.11: 31460635-31460635 on Build GRCh38
3986124 c.176A>G p.H59R Chr.11: 31460644-31460644 on Build GRCh38
214215 c.158C>G p.A53G Chr.11: 31460662-31460662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374694863 Chr.11: 31460722-31460722 on Build GRCh38
rs754481017 Chr.11: 31460726-31460726 on Build GRCh38
rs745957995 Chr.11: 31460671-31460671 on Build GRCh38
rs768536844 Chr.11: 31460688-31460688 on Build GRCh38
rs753183948 Chr.11: 31460601-31460601 on Build GRCh38
rs753270607 Chr.11: 31460741-31460741 on Build GRCh38
rs574196289 Chr.11: 31460723-31460723 on Build GRCh38
rs140884488 Chr.11: 31460717-31460717 on Build GRCh38
rs1002280520 Chr.11: 31460767-31460767 on Build GRCh38
rs1054470435 Chr.11: 31460438-31460438 on Build GRCh38
rs200077336 Chr.11: 31460471-31460471 on Build GRCh38
rs758071114 Chr.11: 31460598-31460598 on Build GRCh38
rs974333842 Chr.11: 31460867-31460867 on Build GRCh38
rs765035013 Chr.11: 31460592-31460592 on Build GRCh38
rs757330958 Chr.11: 31460696-31460696 on Build GRCh38
rs765159617 Chr.11: 31460711-31460711 on Build GRCh38
rs749534500 Chr.11: 31460648-31460648 on Build GRCh38
rs971440248 Chr.11: 31460768-31460768 on Build GRCh38
rs934500765 Chr.11: 31460838-31460838 on Build GRCh38
rs750074290 Chr.11: 31460596-31460596 on Build GRCh38
rs370083551 Chr.11: 31460699-31460699 on Build GRCh38
rs751609546 Chr.11: 31460615-31460615 on Build GRCh38
rs1046774395 Chr.11: 31460443-31460443 on Build GRCh38
rs201106760 Chr.11: 31460476-31460477 on Build GRCh38
rs778072033 Chr.11: 31460647-31460647 on Build GRCh38
rs762723126 Chr.11: 31460572-31460574 on Build GRCh38
rs761558164 Chr.11: 31460710-31460710 on Build GRCh38
rs191752636 Chr.11: 31460552-31460552 on Build GRCh38
rs183886081 Chr.11: 31460845-31460845 on Build GRCh38
rs1039468338 Chr.11: 31460635-31460635 on Build GRCh38
rs943396206 Chr.11: 31460487-31460487 on Build GRCh38
rs767958655 Chr.11: 31460736-31460736 on Build GRCh38
rs934469872 Chr.11: 31460434-31460434 on Build GRCh38
rs997767519 Chr.11: 31460615-31460615 on Build GRCh38
rs751011374 Chr.11: 31460629-31460629 on Build GRCh38
rs11031383 Chr.11: 31460634-31460634 on Build GRCh38
rs753645594 Chr.11: 31460575-31460575 on Build GRCh38
rs779720688 Chr.11: 31460602-31460602 on Build GRCh38
rs761658032 Chr.11: 31460585-31460585 on Build GRCh38
rs771085131 Chr.11: 31460667-31460667 on Build GRCh38
rs981342269 Chr.11: 31460788-31460788 on Build GRCh38
rs770112359 Chr.11: 31460645-31460645 on Build GRCh38
rs374734567 Chr.11: 31460639-31460639 on Build GRCh38
rs756572968 Chr.11: 31460763-31460763 on Build GRCh38
rs537903910 Chr.11: 31460798-31460798 on Build GRCh38
rs774301812 Chr.11: 31460670-31460670 on Build GRCh38
rs970700710 Chr.11: 31460750-31460750 on Build GRCh38
rs367907378 Chr.11: 31460724-31460724 on Build GRCh38
rs755501262 Chr.11: 31460638-31460638 on Build GRCh38
rs765725389 Chr.11: 31460522-31460522 on Build GRCh38

View Full Product Details