Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46358870-46359440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

SNP Detail

SNP ID Chromosome Location
rs766030977 Chr.20: 46359296-46359296 on Build GRCh38
rs987817821 Chr.20: 46358987-46358987 on Build GRCh38
rs1030355637 Chr.20: 46359373-46359373 on Build GRCh38
rs1006687343 Chr.20: 46358911-46358911 on Build GRCh38
rs1018143391 Chr.20: 46359167-46359167 on Build GRCh38
rs887981659 Chr.20: 46359326-46359326 on Build GRCh38
rs756138409 Chr.20: 46359229-46359229 on Build GRCh38
rs76610198 Chr.20: 46359250-46359250 on Build GRCh38
rs972710213 Chr.20: 46358944-46358944 on Build GRCh38
rs560507590 Chr.20: 46359380-46359380 on Build GRCh38
rs149619366 Chr.20: 46358900-46358900 on Build GRCh38
rs934752808 Chr.20: 46359254-46359254 on Build GRCh38
rs114143690 Chr.20: 46359308-46359308 on Build GRCh38
rs942192295 Chr.20: 46359330-46359330 on Build GRCh38
rs867413428 Chr.20: 46359001-46359001 on Build GRCh38
rs989327715 Chr.20: 46359300-46359300 on Build GRCh38
rs545391578 Chr.20: 46358904-46358904 on Build GRCh38
rs941585731 Chr.20: 46359381-46359381 on Build GRCh38
rs997928678 Chr.20: 46359129-46359129 on Build GRCh38
rs897912906 Chr.20: 46359348-46359348 on Build GRCh38
rs989948587 Chr.20: 46359218-46359218 on Build GRCh38
rs577860001 Chr.20: 46359018-46359018 on Build GRCh38
rs1046018779 Chr.20: 46359060-46359060 on Build GRCh38
rs554364720 Chr.20: 46358913-46358913 on Build GRCh38
rs955288064 Chr.20: 46359156-46359156 on Build GRCh38
rs1017979047 Chr.20: 46359214-46359214 on Build GRCh38
rs867561180 Chr.20: 46359202-46359202 on Build GRCh38
rs950065985 Chr.20: 46359083-46359083 on Build GRCh38
rs937619965 Chr.20: 46359275-46359275 on Build GRCh38
rs185833253 Chr.20: 46359210-46359210 on Build GRCh38
rs934700356 Chr.20: 46359069-46359069 on Build GRCh38
rs922511707 Chr.20: 46358950-46358950 on Build GRCh38
rs562148227 Chr.20: 46359080-46359080 on Build GRCh38
rs910456947 Chr.20: 46359006-46359006 on Build GRCh38
rs1036493569 Chr.20: 46359334-46359334 on Build GRCh38
rs2771285 Chr.20: 46359415-46359415 on Build GRCh38
rs75008282 Chr.20: 46359051-46359051 on Build GRCh38
rs527663286 Chr.20: 46359384-46359384 on Build GRCh38
rs112377987 Chr.20: 46359189-46359189 on Build GRCh38
rs954556128 Chr.20: 46359168-46359168 on Build GRCh38
rs576913060 Chr.20: 46359052-46359052 on Build GRCh38
rs540459127 Chr.20: 46359385-46359385 on Build GRCh38
rs180791124 Chr.20: 46359130-46359130 on Build GRCh38
rs997581294 Chr.20: 46359349-46359349 on Build GRCh38
rs551925625 Chr.20: 46358901-46358901 on Build GRCh38
rs572691718 Chr.20: 46359010-46359010 on Build GRCh38
rs529899104 Chr.20: 46359124-46359124 on Build GRCh38
rs985298754 Chr.20: 46358992-46358992 on Build GRCh38
rs1006337442 Chr.20: 46359116-46359116 on Build GRCh38
rs1005666098 Chr.20: 46358903-46358903 on Build GRCh38
rs543862041 Chr.20: 46359075-46359075 on Build GRCh38
rs907464899 Chr.20: 46359070-46359070 on Build GRCh38
rs911028856 Chr.20: 46358996-46358996 on Build GRCh38
rs898931040 Chr.20: 46359154-46359154 on Build GRCh38
rs962342094 Chr.20: 46358921-46358921 on Build GRCh38
rs73303977 Chr.20: 46359166-46359166 on Build GRCh38
rs887930742 Chr.20: 46359108-46359108 on Build GRCh38
rs1054593615 Chr.20: 46359021-46359021 on Build GRCh38
rs955236273 Chr.20: 46358975-46358975 on Build GRCh38
rs1016004430 Chr.20: 46358908-46358908 on Build GRCh38
rs945774831 Chr.20: 46359244-46359244 on Build GRCh38
rs962108893 Chr.20: 46359125-46359125 on Build GRCh38
rs928282479 Chr.20: 46359263-46359263 on Build GRCh38

View Full Product Details