Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16026394-16026894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKA
Entrez Gene ID
1187
Gene Name
chloride voltage-gated channel Ka
Gene Aliases
CLCK1, ClC-K1, hClC-Ka
UniGene
Hs.591533
Gene Chromosome Location
Chr.1: 16021991-16034050 on Build GRCh38
Transcripts
NM_001042704.1 NM_001257139.1 NM_004070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463323 c.551G>A p.R184H Chr.1: 16026588-16026588 on Build GRCh38
7361437 c.507C>G p.F169L Chr.1: 16026544-16026544 on Build GRCh38
1335857 c.655+1G>A p.? Chr.1: 16026776-16026776 on Build GRCh38
6215701 c.614C>T p.A205V Chr.1: 16026734-16026734 on Build GRCh38
5674995 c.655+3G>T p.? Chr.1: 16026778-16026778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776631698 Chr.1: 16026549-16026549 on Build GRCh38
rs112900744 Chr.1: 16026461-16026461 on Build GRCh38
rs759964930 Chr.1: 16026752-16026752 on Build GRCh38
rs570937615 Chr.1: 16026436-16026436 on Build GRCh38
rs781309400 Chr.1: 16026710-16026710 on Build GRCh38
rs775251887 Chr.1: 16026566-16026566 on Build GRCh38
rs944652877 Chr.1: 16026656-16026656 on Build GRCh38
rs757434753 Chr.1: 16026768-16026768 on Build GRCh38
rs763464090 Chr.1: 16026581-16026581 on Build GRCh38
rs1010069 Chr.1: 16026442-16026442 on Build GRCh38
rs747105403 Chr.1: 16026637-16026637 on Build GRCh38
rs555359365 Chr.1: 16026613-16026613 on Build GRCh38
rs959052356 Chr.1: 16026819-16026819 on Build GRCh38
rs753955664 Chr.1: 16026761-16026761 on Build GRCh38
rs374987775 Chr.1: 16026579-16026579 on Build GRCh38
rs531064376 Chr.1: 16026850-16026850 on Build GRCh38
rs557960613 Chr.1: 16026694-16026694 on Build GRCh38
rs754261632 Chr.1: 16026610-16026610 on Build GRCh38
rs370650380 Chr.1: 16026539-16026539 on Build GRCh38
rs775413977 Chr.1: 16026757-16026758 on Build GRCh38
rs372470559 Chr.1: 16026586-16026586 on Build GRCh38
rs370155493 Chr.1: 16026480-16026495 on Build GRCh38
rs567139740 Chr.1: 16026521-16026521 on Build GRCh38
rs766870843 Chr.1: 16026608-16026608 on Build GRCh38
rs769670534 Chr.1: 16026657-16026657 on Build GRCh38
rs777133212 Chr.1: 16026735-16026735 on Build GRCh38
rs768109067 Chr.1: 16026498-16026498 on Build GRCh38
rs756524581 Chr.1: 16026514-16026514 on Build GRCh38
rs529112534 Chr.1: 16026820-16026820 on Build GRCh38
rs769617228 Chr.1: 16026565-16026565 on Build GRCh38
rs759390883 Chr.1: 16026559-16026559 on Build GRCh38
rs952329167 Chr.1: 16026619-16026619 on Build GRCh38
rs745765503 Chr.1: 16026665-16026665 on Build GRCh38
rs752964218 Chr.1: 16026692-16026692 on Build GRCh38
rs548769869 Chr.1: 16026766-16026799 on Build GRCh38
rs750856639 Chr.1: 16026504-16026504 on Build GRCh38
rs200009004 Chr.1: 16026592-16026592 on Build GRCh38
rs751638580 Chr.1: 16026704-16026704 on Build GRCh38
rs750391236 Chr.1: 16026775-16026775 on Build GRCh38
rs755985942 Chr.1: 16026715-16026715 on Build GRCh38
rs749089655 Chr.1: 16026792-16026792 on Build GRCh38
rs771534800 Chr.1: 16026733-16026733 on Build GRCh38
rs769682824 Chr.1: 16026714-16026714 on Build GRCh38
rs374410422 Chr.1: 16026625-16026625 on Build GRCh38
rs1021142890 Chr.1: 16026499-16026499 on Build GRCh38
rs930730530 Chr.1: 16026435-16026435 on Build GRCh38
rs146015976 Chr.1: 16026477-16026492 on Build GRCh38
rs141655219 Chr.1: 16026562-16026562 on Build GRCh38
rs138548956 Chr.1: 16026699-16026699 on Build GRCh38
rs757379581 Chr.1: 16026706-16026706 on Build GRCh38
rs755262766 Chr.1: 16026527-16026527 on Build GRCh38
rs773801220 Chr.1: 16026666-16026666 on Build GRCh38
rs779648302 Chr.1: 16026718-16026718 on Build GRCh38
rs1028152061 Chr.1: 16026473-16026473 on Build GRCh38
rs768254686 Chr.1: 16026725-16026725 on Build GRCh38
rs920753103 Chr.1: 16026609-16026609 on Build GRCh38
rs766594254 Chr.1: 16026464-16026464 on Build GRCh38
rs752311189 Chr.1: 16026486-16026486 on Build GRCh38
rs755317852 Chr.1: 16026622-16026622 on Build GRCh38
rs535444141 Chr.1: 16026543-16026543 on Build GRCh38
rs762455464 Chr.1: 16026663-16026663 on Build GRCh38
rs966367718 Chr.1: 16026505-16026505 on Build GRCh38
rs779150006 Chr.1: 16026624-16026624 on Build GRCh38
rs778465270 Chr.1: 16026795-16026795 on Build GRCh38
rs768429352 Chr.1: 16026781-16026782 on Build GRCh38
rs767648310 Chr.1: 16026771-16026771 on Build GRCh38
rs371660947 Chr.1: 16026762-16026762 on Build GRCh38
rs902267709 Chr.1: 16026787-16026787 on Build GRCh38
rs747753167 Chr.1: 16026796-16026796 on Build GRCh38
rs575335621 Chr.1: 16026617-16026617 on Build GRCh38
rs766923963 Chr.1: 16026682-16026682 on Build GRCh38
rs376882398 Chr.1: 16026640-16026640 on Build GRCh38
rs750503425 Chr.1: 16026458-16026458 on Build GRCh38
rs774035133 Chr.1: 16026730-16026730 on Build GRCh38
rs377164446 Chr.1: 16026494-16026494 on Build GRCh38
rs748280702 Chr.1: 16026540-16026540 on Build GRCh38
rs759928289 Chr.1: 16026690-16026690 on Build GRCh38
rs540395494 Chr.1: 16026782-16026782 on Build GRCh38
rs776032487 Chr.1: 16026822-16026822 on Build GRCh38
rs745752380 Chr.1: 16026711-16026711 on Build GRCh38
rs560019715 Chr.1: 16026808-16026808 on Build GRCh38
rs908076005 Chr.1: 16026852-16026852 on Build GRCh38
rs747655936 Chr.1: 16026731-16026731 on Build GRCh38
rs768217760 Chr.1: 16026664-16026664 on Build GRCh38
rs775305478 Chr.1: 16026658-16026658 on Build GRCh38
rs761375883 Chr.1: 16026670-16026670 on Build GRCh38
rs369567599 Chr.1: 16026824-16026824 on Build GRCh38
rs765524557 Chr.1: 16026691-16026691 on Build GRCh38
rs762556682 Chr.1: 16026580-16026580 on Build GRCh38
rs760874889 Chr.1: 16026462-16026462 on Build GRCh38
rs561745386 Chr.1: 16026841-16026841 on Build GRCh38
rs781250795 Chr.1: 16026644-16026644 on Build GRCh38
rs777644672 Chr.1: 16026635-16026635 on Build GRCh38
rs530343797 Chr.1: 16026525-16026526 on Build GRCh38
rs371805800 Chr.1: 16026633-16026633 on Build GRCh38
rs771571706 Chr.1: 16026806-16026806 on Build GRCh38
rs1042098784 Chr.1: 16026782-16026782 on Build GRCh38
rs749105337 Chr.1: 16026724-16026724 on Build GRCh38
rs367613620 Chr.1: 16026686-16026686 on Build GRCh38
rs765726723 Chr.1: 16026757-16026757 on Build GRCh38
rs745697389 Chr.1: 16026651-16026651 on Build GRCh38
rs746506653 Chr.1: 16026817-16026817 on Build GRCh38
rs754774466 Chr.1: 16026794-16026794 on Build GRCh38

View Full Product Details