Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72544732-72545237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARIH1
Entrez Gene ID
25820
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Gene Aliases
ARI, HARI, HHARI, UBCH7BP
UniGene
Hs.268787
Gene Chromosome Location
Chr.15: 72474282-72586563 on Build GRCh38
Transcripts
NM_005744.3 XM_011521438.2 XM_011521439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6244715 c.444G>T p.R148S Chr.15: 72544820-72544820 on Build GRCh38
6864195 c.587C>T p.S196L Chr.15: 72544963-72544963 on Build GRCh38
3748608 c.507G>A p.K169K Chr.15: 72544883-72544883 on Build GRCh38
4663317 c.519C>T p.R173R Chr.15: 72544895-72544895 on Build GRCh38
7148140 c.559C>G p.Q187E Chr.15: 72544935-72544935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770948239 Chr.15: 72544774-72544774 on Build GRCh38
rs376187664 Chr.15: 72544978-72544978 on Build GRCh38
rs901739742 Chr.15: 72545144-72545144 on Build GRCh38
rs911349831 Chr.15: 72545099-72545099 on Build GRCh38
rs1041080867 Chr.15: 72545117-72545117 on Build GRCh38
rs748816194 Chr.15: 72545054-72545054 on Build GRCh38
rs781271565 Chr.15: 72544775-72544775 on Build GRCh38
rs1025353869 Chr.15: 72545178-72545178 on Build GRCh38
rs567631177 Chr.15: 72545159-72545160 on Build GRCh38
rs573613293 Chr.15: 72545038-72545038 on Build GRCh38
rs745378017 Chr.15: 72544784-72544786 on Build GRCh38
rs749780125 Chr.15: 72544862-72544862 on Build GRCh38
rs779301712 Chr.15: 72545003-72545003 on Build GRCh38
rs756567197 Chr.15: 72544797-72544797 on Build GRCh38
rs746505361 Chr.15: 72544887-72544887 on Build GRCh38
rs780665447 Chr.15: 72544805-72544805 on Build GRCh38
rs534511248 Chr.15: 72545074-72545074 on Build GRCh38
rs556067386 Chr.15: 72545163-72545163 on Build GRCh38
rs770559164 Chr.15: 72544888-72544888 on Build GRCh38
rs774271876 Chr.15: 72545193-72545193 on Build GRCh38
rs779405166 Chr.15: 72544952-72544952 on Build GRCh38
rs933413352 Chr.15: 72544861-72544861 on Build GRCh38
rs769548634 Chr.15: 72544913-72544913 on Build GRCh38
rs200248700 Chr.15: 72544993-72544993 on Build GRCh38
rs771679181 Chr.15: 72544838-72544840 on Build GRCh38
rs186887765 Chr.15: 72544919-72544919 on Build GRCh38
rs759288383 Chr.15: 72544905-72544905 on Build GRCh38
rs750016031 Chr.15: 72545072-72545072 on Build GRCh38
rs750737971 Chr.15: 72544778-72544778 on Build GRCh38
rs764009252 Chr.15: 72544923-72544923 on Build GRCh38
rs1005885775 Chr.15: 72545180-72545180 on Build GRCh38
rs780542854 Chr.15: 72544992-72544992 on Build GRCh38
rs755536712 Chr.15: 72544995-72544995 on Build GRCh38
rs769334669 Chr.15: 72544876-72544876 on Build GRCh38
rs969696237 Chr.15: 72544894-72544894 on Build GRCh38
rs574377204 Chr.15: 72545206-72545206 on Build GRCh38
rs756442722 Chr.15: 72544985-72544985 on Build GRCh38
rs979829613 Chr.15: 72544898-72544898 on Build GRCh38
rs772492316 Chr.15: 72545150-72545150 on Build GRCh38
rs886280234 Chr.15: 72545176-72545176 on Build GRCh38
rs148941976 Chr.15: 72544893-72544893 on Build GRCh38
rs942670569 Chr.15: 72545101-72545101 on Build GRCh38
rs775316269 Chr.15: 72544914-72544914 on Build GRCh38
rs997460312 Chr.15: 72545145-72545145 on Build GRCh38
rs1000868639 Chr.15: 72544947-72544947 on Build GRCh38
rs761809403 Chr.15: 72544975-72544975 on Build GRCh38
rs775705634 Chr.15: 72545204-72545204 on Build GRCh38
rs372914669 Chr.15: 72544982-72544982 on Build GRCh38
rs746306589 Chr.15: 72545027-72545027 on Build GRCh38
rs145634735 Chr.15: 72544973-72544973 on Build GRCh38

View Full Product Details