Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75201816-75202345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

SNP Detail

SNP ID Chromosome Location
rs1024149546 Chr.15: 75201924-75201924 on Build GRCh38
rs951006315 Chr.15: 75201849-75201849 on Build GRCh38
rs948188330 Chr.15: 75202244-75202244 on Build GRCh38
rs951290260 Chr.15: 75202229-75202229 on Build GRCh38
rs980727646 Chr.15: 75202249-75202249 on Build GRCh38
rs1003661873 Chr.15: 75202082-75202082 on Build GRCh38
rs553070870 Chr.15: 75202296-75202296 on Build GRCh38
rs918401356 Chr.15: 75202225-75202225 on Build GRCh38
rs984186267 Chr.15: 75202232-75202232 on Build GRCh38
rs1017976195 Chr.15: 75202171-75202171 on Build GRCh38
rs376764100 Chr.15: 75202311-75202311 on Build GRCh38
rs955805953 Chr.15: 75201926-75201926 on Build GRCh38
rs930046977 Chr.15: 75201966-75201966 on Build GRCh38
rs1009949507 Chr.15: 75202143-75202143 on Build GRCh38
rs1048377305 Chr.15: 75201891-75201891 on Build GRCh38
rs1029358822 Chr.15: 75202222-75202222 on Build GRCh38
rs1048973158 Chr.15: 75201991-75201991 on Build GRCh38
rs948072131 Chr.15: 75202064-75202064 on Build GRCh38
rs1010417637 Chr.15: 75201927-75201927 on Build GRCh38
rs955293796 Chr.15: 75202224-75202224 on Build GRCh38
rs1053967186 Chr.15: 75202006-75202006 on Build GRCh38
rs1006560049 Chr.15: 75202168-75202168 on Build GRCh38
rs893848278 Chr.15: 75201919-75201919 on Build GRCh38
rs965275466 Chr.15: 75202200-75202200 on Build GRCh38
rs988031857 Chr.15: 75201965-75201965 on Build GRCh38
rs976677927 Chr.15: 75202221-75202221 on Build GRCh38
rs918452314 Chr.15: 75201954-75201954 on Build GRCh38
rs1045291811 Chr.15: 75201918-75201918 on Build GRCh38
rs1045133673 Chr.15: 75202068-75202068 on Build GRCh38
rs1051997792 Chr.15: 75201913-75201913 on Build GRCh38
rs963713206 Chr.15: 75201934-75201934 on Build GRCh38
rs983771062 Chr.15: 75201870-75201870 on Build GRCh38
rs985342607 Chr.15: 75201937-75201937 on Build GRCh38
rs1047333106 Chr.15: 75202089-75202089 on Build GRCh38
rs930074418 Chr.15: 75201886-75201886 on Build GRCh38
rs866001607 Chr.15: 75201967-75201967 on Build GRCh38
rs968792363 Chr.15: 75201931-75201931 on Build GRCh38
rs541137348 Chr.15: 75202237-75202237 on Build GRCh38
rs915514959 Chr.15: 75202277-75202277 on Build GRCh38
rs569884649 Chr.15: 75201908-75201908 on Build GRCh38
rs909842209 Chr.15: 75202016-75202016 on Build GRCh38
rs577065182 Chr.15: 75201952-75201952 on Build GRCh38
rs948055591 Chr.15: 75202025-75202025 on Build GRCh38
rs955173321 Chr.15: 75201956-75201956 on Build GRCh38
rs1003665707 Chr.15: 75201921-75201921 on Build GRCh38
rs372162245 Chr.15: 75201942-75201942 on Build GRCh38
rs924146644 Chr.15: 75201972-75201972 on Build GRCh38
rs918671946 Chr.15: 75201859-75201859 on Build GRCh38
rs956877673 Chr.15: 75202238-75202238 on Build GRCh38
rs935615053 Chr.15: 75201917-75201917 on Build GRCh38
rs927984605 Chr.15: 75202274-75202274 on Build GRCh38
rs989664730 Chr.15: 75202250-75202250 on Build GRCh38
rs1003781589 Chr.15: 75202108-75202108 on Build GRCh38
rs888456679 Chr.15: 75201899-75201899 on Build GRCh38
rs982019719 Chr.15: 75201839-75201839 on Build GRCh38
rs906594611 Chr.15: 75202080-75202080 on Build GRCh38
rs965327521 Chr.15: 75201943-75201943 on Build GRCh38
rs906777714 Chr.15: 75202099-75202099 on Build GRCh38
rs886941643 Chr.15: 75202100-75202100 on Build GRCh38
rs970969912 Chr.15: 75201837-75201847 on Build GRCh38
rs902745553 Chr.15: 75201915-75201915 on Build GRCh38
rs968532845 Chr.15: 75202163-75202163 on Build GRCh38
rs868167844 Chr.15: 75202101-75202101 on Build GRCh38
rs1052526105 Chr.15: 75202111-75202111 on Build GRCh38
rs996897764 Chr.15: 75202157-75202157 on Build GRCh38
rs1047160406 Chr.15: 75202133-75202133 on Build GRCh38
rs143188727 Chr.15: 75201920-75201920 on Build GRCh38
rs1031154182 Chr.15: 75201925-75201925 on Build GRCh38
rs975568938 Chr.15: 75201896-75201896 on Build GRCh38
rs1036976271 Chr.15: 75201846-75201861 on Build GRCh38
rs1021267826 Chr.15: 75201928-75201928 on Build GRCh38
rs892648596 Chr.15: 75202126-75202126 on Build GRCh38
rs939584200 Chr.15: 75202288-75202288 on Build GRCh38
rs909582429 Chr.15: 75201879-75201879 on Build GRCh38
rs931422283 Chr.15: 75201843-75201843 on Build GRCh38
rs537341127 Chr.15: 75201916-75201916 on Build GRCh38
rs1045077164 Chr.15: 75202059-75202059 on Build GRCh38
rs1021801758 Chr.15: 75202146-75202146 on Build GRCh38
rs928024759 Chr.15: 75202306-75202306 on Build GRCh38

View Full Product Details