Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69330377-69330874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG8
Entrez Gene ID
84342
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
CDG2H, DOR1
UniGene
Hs.130849
Gene Chromosome Location
Chr.16: 69328621-69339623 on Build GRCh38
Transcripts
NM_032382.4 NM_022341.1

Gene Symbol
PDF
Entrez Gene ID
64146
Gene Name
peptide deformylase (mitochondrial)
Gene Aliases
-
UniGene
Hs.130849
Gene Chromosome Location
Chr.16: 69328621-69330595 on Build GRCh38
Transcripts
NM_032382.4 NM_022341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5957678 c.32G>T p.W11L Chr.16: 69330539-69330539 on Build GRCh38
3765922 c.31T>C p.W11R Chr.16: 69330540-69330540 on Build GRCh38
7323627 c.138C>T p.R46R Chr.16: 69330433-69330433 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775558858 Chr.16: 69330416-69330416 on Build GRCh38
rs920668791 Chr.16: 69330658-69330658 on Build GRCh38
rs1023610412 Chr.16: 69330806-69330806 on Build GRCh38
rs978025503 Chr.16: 69330526-69330526 on Build GRCh38
rs550201036 Chr.16: 69330720-69330720 on Build GRCh38
rs1039691491 Chr.16: 69330693-69330693 on Build GRCh38
rs1016189204 Chr.16: 69330592-69330592 on Build GRCh38
rs922190390 Chr.16: 69330839-69330839 on Build GRCh38
rs920637612 Chr.16: 69330569-69330569 on Build GRCh38
rs989213496 Chr.16: 69330743-69330743 on Build GRCh38
rs1023956784 Chr.16: 69330765-69330765 on Build GRCh38
rs528288920 Chr.16: 69330701-69330701 on Build GRCh38
rs79876002 Chr.16: 69330642-69330642 on Build GRCh38
rs747145620 Chr.16: 69330579-69330579 on Build GRCh38
rs983419812 Chr.16: 69330589-69330589 on Build GRCh38
rs559003792 Chr.16: 69330577-69330577 on Build GRCh38
rs35955795 Chr.16: 69330600-69330600 on Build GRCh38
rs991119747 Chr.16: 69330699-69330699 on Build GRCh38
rs960880193 Chr.16: 69330536-69330536 on Build GRCh38
rs962605560 Chr.16: 69330769-69330769 on Build GRCh38
rs942353936 Chr.16: 69330751-69330751 on Build GRCh38
rs763767741 Chr.16: 69330463-69330463 on Build GRCh38
rs962862115 Chr.16: 69330653-69330653 on Build GRCh38
rs550213086 Chr.16: 69330515-69330515 on Build GRCh38
rs1018502478 Chr.16: 69330408-69330408 on Build GRCh38
rs1009521681 Chr.16: 69330586-69330586 on Build GRCh38
rs1012207264 Chr.16: 69330733-69330733 on Build GRCh38
rs933901463 Chr.16: 69330767-69330767 on Build GRCh38
rs1026522924 Chr.16: 69330499-69330499 on Build GRCh38
rs1039208226 Chr.16: 69330612-69330612 on Build GRCh38
rs761394854 Chr.16: 69330495-69330495 on Build GRCh38
rs902650775 Chr.16: 69330849-69330849 on Build GRCh38
rs927820215 Chr.16: 69330402-69330402 on Build GRCh38
rs894574168 Chr.16: 69330422-69330422 on Build GRCh38
rs1043920616 Chr.16: 69330846-69330846 on Build GRCh38
rs949574022 Chr.16: 69330628-69330628 on Build GRCh38
rs971399035 Chr.16: 69330816-69330816 on Build GRCh38
rs916453075 Chr.16: 69330652-69330652 on Build GRCh38
rs973483905 Chr.16: 69330654-69330654 on Build GRCh38
rs922515718 Chr.16: 69330762-69330762 on Build GRCh38
rs1003785897 Chr.16: 69330825-69330825 on Build GRCh38
rs762874045 Chr.16: 69330446-69330446 on Build GRCh38
rs758556703 Chr.16: 69330568-69330568 on Build GRCh38
rs554777002 Chr.16: 69330548-69330548 on Build GRCh38
rs991110846 Chr.16: 69330649-69330649 on Build GRCh38
rs8057004 Chr.16: 69330540-69330540 on Build GRCh38
rs1039338061 Chr.16: 69330754-69330754 on Build GRCh38
rs1034700320 Chr.16: 69330716-69330716 on Build GRCh38
rs529767569 Chr.16: 69330798-69330798 on Build GRCh38
rs1037669411 Chr.16: 69330454-69330454 on Build GRCh38
rs774102989 Chr.16: 69330473-69330473 on Build GRCh38
rs532018045 Chr.16: 69330506-69330506 on Build GRCh38
rs950717327 Chr.16: 69330441-69330441 on Build GRCh38
rs950809983 Chr.16: 69330792-69330792 on Build GRCh38
rs1018165322 Chr.16: 69330508-69330508 on Build GRCh38
rs540477623 Chr.16: 69330702-69330702 on Build GRCh38
rs747022716 Chr.16: 69330395-69330395 on Build GRCh38
rs1022034146 Chr.16: 69330621-69330621 on Build GRCh38
rs1043889611 Chr.16: 69330704-69330704 on Build GRCh38
rs188405840 Chr.16: 69330563-69330563 on Build GRCh38
rs112744495 Chr.16: 69330664-69330664 on Build GRCh38
rs750410882 Chr.16: 69330827-69330828 on Build GRCh38
rs779162844 Chr.16: 69330554-69330554 on Build GRCh38
rs999205445 Chr.16: 69330638-69330638 on Build GRCh38
rs577130537 Chr.16: 69330619-69330619 on Build GRCh38
rs960659660 Chr.16: 69330643-69330643 on Build GRCh38
rs946928494 Chr.16: 69330845-69330845 on Build GRCh38
rs111394740 Chr.16: 69330706-69330706 on Build GRCh38
rs765380108 Chr.16: 69330502-69330503 on Build GRCh38
rs916721773 Chr.16: 69330505-69330505 on Build GRCh38
rs890649794 Chr.16: 69330502-69330502 on Build GRCh38
rs542819052 Chr.16: 69330675-69330675 on Build GRCh38
rs755417363 Chr.16: 69330544-69330544 on Build GRCh38
rs769366329 Chr.16: 69330773-69330773 on Build GRCh38
rs944684906 Chr.16: 69330823-69330823 on Build GRCh38
rs914951877 Chr.16: 69330747-69330747 on Build GRCh38
rs562227918 Chr.16: 69330731-69330731 on Build GRCh38
rs534480677 Chr.16: 69330566-69330566 on Build GRCh38
rs1055310381 Chr.16: 69330504-69330504 on Build GRCh38
rs184453457 Chr.16: 69330534-69330534 on Build GRCh38
rs983704231 Chr.16: 69330448-69330448 on Build GRCh38
rs1012662040 Chr.16: 69330798-69330798 on Build GRCh38
rs1018138824 Chr.16: 69330562-69330562 on Build GRCh38
rs899139532 Chr.16: 69330475-69330475 on Build GRCh38
rs1010699876 Chr.16: 69330617-69330617 on Build GRCh38
rs775033549 Chr.16: 69330784-69330784 on Build GRCh38
rs775560162 Chr.16: 69330549-69330549 on Build GRCh38
rs916361525 Chr.16: 69330546-69330546 on Build GRCh38
rs948841620 Chr.16: 69330551-69330551 on Build GRCh38
rs767089608 Chr.16: 69330496-69330496 on Build GRCh38
rs749161216 Chr.16: 69330847-69330847 on Build GRCh38
rs964493606 Chr.16: 69330510-69330510 on Build GRCh38
rs935098456 Chr.16: 69330725-69330725 on Build GRCh38
rs184252224 Chr.16: 69330713-69330713 on Build GRCh38
rs900659129 Chr.16: 69330616-69330616 on Build GRCh38
rs939179092 Chr.16: 69330403-69330403 on Build GRCh38
rs963626385 Chr.16: 69330594-69330594 on Build GRCh38
rs575189480 Chr.16: 69330432-69330439 on Build GRCh38
rs929367245 Chr.16: 69330660-69330660 on Build GRCh38
rs1057470076 Chr.16: 69330406-69330406 on Build GRCh38
rs924079768 Chr.16: 69330723-69330723 on Build GRCh38
rs1016242964 Chr.16: 69330669-69330669 on Build GRCh38
rs966394597 Chr.16: 69330636-69330636 on Build GRCh38
rs900692880 Chr.16: 69330695-69330695 on Build GRCh38
rs771133753 Chr.16: 69330842-69330842 on Build GRCh38
rs148264366 Chr.16: 69330661-69330661 on Build GRCh38
rs890730439 Chr.16: 69330585-69330585 on Build GRCh38
rs1022004544 Chr.16: 69330518-69330518 on Build GRCh38
rs3811347 Chr.16: 69330774-69330774 on Build GRCh38
rs1004909612 Chr.16: 69330666-69330666 on Build GRCh38
rs561105224 Chr.16: 69330683-69330683 on Build GRCh38
rs886923112 Chr.16: 69330779-69330779 on Build GRCh38
rs769869670 Chr.16: 69330400-69330400 on Build GRCh38
rs935103296 Chr.16: 69330850-69330850 on Build GRCh38
rs1025389420 Chr.16: 69330775-69330775 on Build GRCh38
rs893465234 Chr.16: 69330656-69330656 on Build GRCh38
rs778087920 Chr.16: 69330578-69330578 on Build GRCh38
rs934990473 Chr.16: 69330527-69330527 on Build GRCh38
rs975050877 Chr.16: 69330835-69330835 on Build GRCh38
rs780182921 Chr.16: 69330844-69330844 on Build GRCh38
rs886052250 Chr.16: 69330803-69330803 on Build GRCh38
rs868110177 Chr.16: 69330788-69330788 on Build GRCh38
rs911936186 Chr.16: 69330802-69330802 on Build GRCh38
rs755605675 Chr.16: 69330523-69330523 on Build GRCh38
rs948791022 Chr.16: 69330428-69330428 on Build GRCh38
rs911845749 Chr.16: 69330662-69330662 on Build GRCh38

View Full Product Details