Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75762922-75763423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK1
Entrez Gene ID
2584
Gene Name
galactokinase 1
Gene Aliases
GALK, GK1, HEL-S-19
UniGene
Hs.407966
Gene Chromosome Location
Chr.17: 75757937-75765199 on Build GRCh38
Transcripts
NM_000154.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250828 c.519C>A p.A173A Chr.17: 75763106-75763106 on Build GRCh38
5387323 c.472C>T p.P158S Chr.17: 75763323-75763323 on Build GRCh38
3522311 c.588C>T p.G196G Chr.17: 75763037-75763037 on Build GRCh38
5055781 c.497G>A p.R166H Chr.17: 75763128-75763128 on Build GRCh38
2742026 c.410delG p.G137fs*27 Chr.17: 75763385-75763385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140148059 Chr.17: 75763353-75763353 on Build GRCh38
rs760305997 Chr.17: 75763118-75763118 on Build GRCh38
rs145732791 Chr.17: 75763128-75763128 on Build GRCh38
rs773432859 Chr.17: 75762995-75762995 on Build GRCh38
rs994937759 Chr.17: 75763232-75763232 on Build GRCh38
rs767799179 Chr.17: 75763111-75763111 on Build GRCh38
rs770881427 Chr.17: 75763069-75763069 on Build GRCh38
rs775843885 Chr.17: 75763115-75763115 on Build GRCh38
rs780587160 Chr.17: 75763354-75763354 on Build GRCh38
rs899276169 Chr.17: 75763230-75763230 on Build GRCh38
rs200350673 Chr.17: 75762986-75762986 on Build GRCh38
rs1034818496 Chr.17: 75763054-75763054 on Build GRCh38
rs147739431 Chr.17: 75763094-75763094 on Build GRCh38
rs774392909 Chr.17: 75763205-75763206 on Build GRCh38
rs367550108 Chr.17: 75763129-75763129 on Build GRCh38
rs76743658 Chr.17: 75762980-75762980 on Build GRCh38
rs773416476 Chr.17: 75763073-75763073 on Build GRCh38
rs369410892 Chr.17: 75763290-75763290 on Build GRCh38
rs758398846 Chr.17: 75763286-75763286 on Build GRCh38
rs370204015 Chr.17: 75763005-75763005 on Build GRCh38
rs915070690 Chr.17: 75763385-75763385 on Build GRCh38
rs398031630 Chr.17: 75762960-75762960 on Build GRCh38
rs753104550 Chr.17: 75763349-75763349 on Build GRCh38
rs944834759 Chr.17: 75763277-75763277 on Build GRCh38
rs111788658 Chr.17: 75762963-75762963 on Build GRCh38
rs549737244 Chr.17: 75762960-75762960 on Build GRCh38
rs755764957 Chr.17: 75763031-75763031 on Build GRCh38
rs765888121 Chr.17: 75763224-75763224 on Build GRCh38
rs765744088 Chr.17: 75763348-75763348 on Build GRCh38
rs749544838 Chr.17: 75763148-75763148 on Build GRCh38
rs1009910947 Chr.17: 75763090-75763090 on Build GRCh38
rs142992759 Chr.17: 75763034-75763034 on Build GRCh38
rs373022508 Chr.17: 75762988-75762988 on Build GRCh38
rs141983398 Chr.17: 75763106-75763106 on Build GRCh38
rs188851202 Chr.17: 75762996-75762996 on Build GRCh38
rs988018523 Chr.17: 75763368-75763368 on Build GRCh38
rs996395516 Chr.17: 75762951-75762951 on Build GRCh38
rs774490771 Chr.17: 75763158-75763158 on Build GRCh38
rs371680833 Chr.17: 75763172-75763172 on Build GRCh38
rs374127081 Chr.17: 75763306-75763306 on Build GRCh38
rs749398448 Chr.17: 75763089-75763089 on Build GRCh38
rs1052029690 Chr.17: 75763155-75763155 on Build GRCh38
rs915037769 Chr.17: 75763055-75763055 on Build GRCh38
rs767329054 Chr.17: 75763384-75763385 on Build GRCh38
rs763715411 Chr.17: 75763204-75763204 on Build GRCh38
rs776098323 Chr.17: 75763206-75763206 on Build GRCh38
rs749966530 Chr.17: 75763231-75763231 on Build GRCh38
rs770871380 Chr.17: 75763151-75763151 on Build GRCh38
rs963791589 Chr.17: 75762989-75762989 on Build GRCh38
rs377498337 Chr.17: 75762985-75762985 on Build GRCh38
rs555279924 Chr.17: 75763140-75763140 on Build GRCh38
rs756405600 Chr.17: 75763145-75763145 on Build GRCh38
rs368634148 Chr.17: 75762939-75762939 on Build GRCh38
rs982043960 Chr.17: 75763013-75763013 on Build GRCh38
rs367594410 Chr.17: 75763051-75763051 on Build GRCh38
rs761316738 Chr.17: 75763378-75763378 on Build GRCh38
rs769550271 Chr.17: 75763379-75763379 on Build GRCh38
rs747536956 Chr.17: 75763355-75763355 on Build GRCh38
rs557302605 Chr.17: 75763234-75763234 on Build GRCh38
rs780027352 Chr.17: 75763039-75763039 on Build GRCh38
rs758416839 Chr.17: 75763139-75763139 on Build GRCh38
rs890693618 Chr.17: 75763179-75763179 on Build GRCh38
rs569457915 Chr.17: 75762974-75762974 on Build GRCh38
rs544089175 Chr.17: 75763159-75763159 on Build GRCh38
rs820386 Chr.17: 75763223-75763223 on Build GRCh38
rs763803267 Chr.17: 75763124-75763124 on Build GRCh38
rs757213297 Chr.17: 75763033-75763033 on Build GRCh38
rs759340781 Chr.17: 75763213-75763213 on Build GRCh38
rs750290029 Chr.17: 75763130-75763130 on Build GRCh38
rs778755807 Chr.17: 75762969-75762969 on Build GRCh38
rs375660566 Chr.17: 75763313-75763313 on Build GRCh38
rs372078515 Chr.17: 75763126-75763126 on Build GRCh38
rs770785288 Chr.17: 75762964-75762964 on Build GRCh38
rs905165111 Chr.17: 75763347-75763347 on Build GRCh38
rs752304350 Chr.17: 75763017-75763017 on Build GRCh38
rs80084721 Chr.17: 75763032-75763032 on Build GRCh38
rs777059513 Chr.17: 75763370-75763370 on Build GRCh38
rs746861299 Chr.17: 75763143-75763143 on Build GRCh38
rs35008831 Chr.17: 75762958-75762958 on Build GRCh38
rs147800408 Chr.17: 75763336-75763336 on Build GRCh38
rs772623590 Chr.17: 75763166-75763166 on Build GRCh38
rs759864030 Chr.17: 75763008-75763008 on Build GRCh38
rs745653883 Chr.17: 75763037-75763037 on Build GRCh38
rs1003512759 Chr.17: 75763334-75763334 on Build GRCh38
rs766494612 Chr.17: 75763293-75763293 on Build GRCh38
rs193213618 Chr.17: 75763209-75763209 on Build GRCh38
rs766401384 Chr.17: 75763399-75763399 on Build GRCh38
rs771717336 Chr.17: 75763038-75763038 on Build GRCh38
rs201110055 Chr.17: 75763396-75763396 on Build GRCh38
rs912070478 Chr.17: 75763249-75763249 on Build GRCh38
rs748316786 Chr.17: 75763044-75763044 on Build GRCh38
rs763880577 Chr.17: 75763127-75763127 on Build GRCh38
rs767887241 Chr.17: 75763010-75763010 on Build GRCh38
rs376725988 Chr.17: 75763183-75763183 on Build GRCh38
rs749400702 Chr.17: 75763345-75763345 on Build GRCh38
rs866703938 Chr.17: 75763084-75763084 on Build GRCh38
rs766858133 Chr.17: 75763004-75763004 on Build GRCh38
rs398079177 Chr.17: 75762959-75762959 on Build GRCh38
rs575077063 Chr.17: 75763146-75763146 on Build GRCh38
rs150323432 Chr.17: 75763363-75763363 on Build GRCh38
rs771443754 Chr.17: 75763091-75763091 on Build GRCh38
rs138057626 Chr.17: 75763387-75763387 on Build GRCh38
rs754767873 Chr.17: 75763321-75763321 on Build GRCh38
rs577510039 Chr.17: 75763022-75763022 on Build GRCh38

View Full Product Details