Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108748793-108749280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_006267.4 XM_005264002.2 XM_005264003.2 XM_005264004.2 XM_005264005.4 XM_005264007.2 XM_011511575.2 XM_011511576.2 XM_011511578.2 XM_017004623.1 XM_017004624.1 XM_017004625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4720234 c.1210G>A p.D404N Chr.2: 108749066-108749066 on Build GRCh38
4720235 c.1267G>A p.D423N Chr.2: 108749123-108749123 on Build GRCh38
5731697 c.1172C>G p.S391C Chr.2: 108749028-108749028 on Build GRCh38
7386830 c.1087C>T p.R363C Chr.2: 108748943-108748943 on Build GRCh38
3894496 c.1102T>C p.F368L Chr.2: 108748958-108748958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562171866 Chr.2: 108749251-108749251 on Build GRCh38
rs750123699 Chr.2: 108749069-108749069 on Build GRCh38
rs779663612 Chr.2: 108749016-108749016 on Build GRCh38
rs765812140 Chr.2: 108749077-108749077 on Build GRCh38
rs778919296 Chr.2: 108749066-108749068 on Build GRCh38
rs748650151 Chr.2: 108748974-108748974 on Build GRCh38
rs769873183 Chr.2: 108749048-108749048 on Build GRCh38
rs2693113 Chr.2: 108748968-108748968 on Build GRCh38
rs188392381 Chr.2: 108748861-108748861 on Build GRCh38
rs539296863 Chr.2: 108749026-108749026 on Build GRCh38
rs377754100 Chr.2: 108748904-108748904 on Build GRCh38
rs770828218 Chr.2: 108748907-108748908 on Build GRCh38
rs533671657 Chr.2: 108749061-108749061 on Build GRCh38
rs760348187 Chr.2: 108749073-108749073 on Build GRCh38
rs757639418 Chr.2: 108749090-108749090 on Build GRCh38
rs955460041 Chr.2: 108748900-108748900 on Build GRCh38
rs139206564 Chr.2: 108749060-108749060 on Build GRCh38
rs761793907 Chr.2: 108749126-108749126 on Build GRCh38
rs746093104 Chr.2: 108749099-108749099 on Build GRCh38
rs370375563 Chr.2: 108748989-108748989 on Build GRCh38
rs192629843 Chr.2: 108748907-108748907 on Build GRCh38
rs758155568 Chr.2: 108748935-108748935 on Build GRCh38
rs115292586 Chr.2: 108748876-108748876 on Build GRCh38
rs753329326 Chr.2: 108749171-108749171 on Build GRCh38
rs766031531 Chr.2: 108749162-108749162 on Build GRCh38
rs928757106 Chr.2: 108749199-108749199 on Build GRCh38
rs773074673 Chr.2: 108749152-108749152 on Build GRCh38
rs768518457 Chr.2: 108749122-108749122 on Build GRCh38
rs149207118 Chr.2: 108748943-108748943 on Build GRCh38
rs540387051 Chr.2: 108749235-108749235 on Build GRCh38
rs371319181 Chr.2: 108749066-108749066 on Build GRCh38
rs776867851 Chr.2: 108749039-108749039 on Build GRCh38
rs563927547 Chr.2: 108748825-108748825 on Build GRCh38
rs564987324 Chr.2: 108748899-108748903 on Build GRCh38
rs749369348 Chr.2: 108749121-108749121 on Build GRCh38
rs201087513 Chr.2: 108749081-108749081 on Build GRCh38
rs760472593 Chr.2: 108749161-108749161 on Build GRCh38
rs765260146 Chr.2: 108748992-108748992 on Build GRCh38
rs368553521 Chr.2: 108749125-108749125 on Build GRCh38
rs753909167 Chr.2: 108748921-108748921 on Build GRCh38
rs775570061 Chr.2: 108749059-108749059 on Build GRCh38
rs139239955 Chr.2: 108748953-108748953 on Build GRCh38
rs544178123 Chr.2: 108749091-108749091 on Build GRCh38
rs764602455 Chr.2: 108749181-108749181 on Build GRCh38
rs756155473 Chr.2: 108748856-108748858 on Build GRCh38
rs568766008 Chr.2: 108748979-108748979 on Build GRCh38
rs1003233688 Chr.2: 108748843-108748843 on Build GRCh38
rs779534152 Chr.2: 108748973-108748973 on Build GRCh38
rs886898523 Chr.2: 108748859-108748859 on Build GRCh38
rs1044622451 Chr.2: 108748819-108748819 on Build GRCh38
rs755717584 Chr.2: 108749013-108749013 on Build GRCh38
rs373103556 Chr.2: 108748912-108748912 on Build GRCh38
rs748616624 Chr.2: 108749018-108749018 on Build GRCh38
rs781289705 Chr.2: 108748899-108748899 on Build GRCh38
rs752527357 Chr.2: 108748926-108748926 on Build GRCh38
rs374334459 Chr.2: 108749074-108749074 on Build GRCh38
rs938955977 Chr.2: 108749200-108749200 on Build GRCh38
rs550471130 Chr.2: 108748944-108748944 on Build GRCh38
rs749996744 Chr.2: 108749008-108749008 on Build GRCh38
rs924601497 Chr.2: 108748924-108748924 on Build GRCh38
rs759527479 Chr.2: 108748925-108748925 on Build GRCh38
rs773568585 Chr.2: 108748985-108748985 on Build GRCh38
rs771128137 Chr.2: 108748987-108748987 on Build GRCh38
rs139907041 Chr.2: 108749087-108749087 on Build GRCh38
rs747377255 Chr.2: 108749031-108749031 on Build GRCh38
rs758874401 Chr.2: 108749024-108749024 on Build GRCh38
rs2693112 Chr.2: 108748970-108748970 on Build GRCh38
rs759115189 Chr.2: 108749176-108749176 on Build GRCh38
rs771181154 Chr.2: 108749038-108749038 on Build GRCh38
rs751369630 Chr.2: 108748999-108748999 on Build GRCh38
rs761672897 Chr.2: 108749064-108749064 on Build GRCh38
rs374936566 Chr.2: 108749035-108749035 on Build GRCh38
rs747201977 Chr.2: 108748986-108748986 on Build GRCh38
rs767259689 Chr.2: 108749005-108749005 on Build GRCh38
rs143137111 Chr.2: 108748964-108748964 on Build GRCh38
rs140275032 Chr.2: 108749065-108749065 on Build GRCh38
rs774067116 Chr.2: 108749062-108749062 on Build GRCh38
rs948957560 Chr.2: 108748814-108748814 on Build GRCh38
rs756921581 Chr.2: 108749001-108749001 on Build GRCh38
rs776335238 Chr.2: 108749076-108749076 on Build GRCh38
rs184221435 Chr.2: 108748860-108748860 on Build GRCh38
rs555090749 Chr.2: 108748997-108748997 on Build GRCh38
rs753385182 Chr.2: 108749080-108749080 on Build GRCh38
rs200373569 Chr.2: 108749114-108749114 on Build GRCh38
rs376733981 Chr.2: 108749046-108749046 on Build GRCh38
rs769995347 Chr.2: 108749108-108749108 on Build GRCh38
rs564502843 Chr.2: 108749194-108749194 on Build GRCh38
rs778312205 Chr.2: 108749082-108749082 on Build GRCh38
rs546414115 Chr.2: 108749190-108749190 on Build GRCh38
rs572187533 Chr.2: 108749252-108749252 on Build GRCh38
rs772008468 Chr.2: 108749127-108749127 on Build GRCh38
rs775309139 Chr.2: 108748995-108748995 on Build GRCh38
rs780788037 Chr.2: 108748881-108748881 on Build GRCh38
rs374341209 Chr.2: 108749138-108749138 on Build GRCh38
rs11885340 Chr.2: 108748848-108748848 on Build GRCh38
rs962492422 Chr.2: 108748872-108748872 on Build GRCh38
rs759578724 Chr.2: 108748990-108748990 on Build GRCh38
rs576001655 Chr.2: 108749182-108749182 on Build GRCh38
rs773339378 Chr.2: 108748910-108748910 on Build GRCh38
rs528659970 Chr.2: 108749208-108749208 on Build GRCh38
rs547003464 Chr.2: 108748893-108748893 on Build GRCh38
rs762886340 Chr.2: 108748996-108748996 on Build GRCh38

View Full Product Details