Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80288785-80289329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_001291812.1 NM_004464.3 NM_033143.2

SNP Detail

SNP ID Chromosome Location
rs34954478 Chr.4: 80288958-80288959 on Build GRCh38
rs573059936 Chr.4: 80288822-80288822 on Build GRCh38
rs918123673 Chr.4: 80289167-80289167 on Build GRCh38
rs889877665 Chr.4: 80288906-80288906 on Build GRCh38
rs1031976190 Chr.4: 80289147-80289147 on Build GRCh38
rs878886742 Chr.4: 80288997-80288997 on Build GRCh38
rs961534696 Chr.4: 80289097-80289097 on Build GRCh38
rs1001463427 Chr.4: 80288997-80288997 on Build GRCh38
rs867670683 Chr.4: 80288828-80288828 on Build GRCh38
rs34997154 Chr.4: 80289040-80289040 on Build GRCh38
rs543083583 Chr.4: 80289146-80289146 on Build GRCh38
rs931286462 Chr.4: 80288868-80288868 on Build GRCh38
rs35837608 Chr.4: 80289258-80289258 on Build GRCh38
rs891612354 Chr.4: 80288892-80288892 on Build GRCh38
rs35583660 Chr.4: 80288996-80288997 on Build GRCh38
rs1000878861 Chr.4: 80289013-80289013 on Build GRCh38
rs11937422 Chr.4: 80289185-80289185 on Build GRCh38
rs1006892544 Chr.4: 80288929-80288929 on Build GRCh38
rs950147761 Chr.4: 80289284-80289284 on Build GRCh38
rs543462070 Chr.4: 80289045-80289045 on Build GRCh38
rs147824048 Chr.4: 80288886-80288886 on Build GRCh38
rs901679685 Chr.4: 80289041-80289041 on Build GRCh38
rs957391799 Chr.4: 80289256-80289256 on Build GRCh38
rs772635563 Chr.4: 80288986-80288998 on Build GRCh38
rs772474992 Chr.4: 80288890-80288890 on Build GRCh38
rs775826914 Chr.4: 80289181-80289181 on Build GRCh38
rs563691572 Chr.4: 80289062-80289062 on Build GRCh38
rs574044270 Chr.4: 80289269-80289269 on Build GRCh38
rs922885122 Chr.4: 80288859-80288859 on Build GRCh38
rs949929047 Chr.4: 80288958-80288958 on Build GRCh38
rs966630929 Chr.4: 80289113-80289113 on Build GRCh38
rs973030741 Chr.4: 80289182-80289182 on Build GRCh38
rs34117650 Chr.4: 80288885-80288885 on Build GRCh38
rs1048880093 Chr.4: 80289224-80289224 on Build GRCh38
rs931552299 Chr.4: 80289192-80289192 on Build GRCh38
rs193012243 Chr.4: 80289199-80289199 on Build GRCh38
rs889783928 Chr.4: 80289242-80289242 on Build GRCh38
rs905742919 Chr.4: 80288988-80288988 on Build GRCh38
rs985829576 Chr.4: 80288861-80288861 on Build GRCh38
rs1045645270 Chr.4: 80288981-80288981 on Build GRCh38
rs1027708698 Chr.4: 80289018-80289019 on Build GRCh38
rs111605934 Chr.4: 80289066-80289066 on Build GRCh38
rs865951432 Chr.4: 80288995-80288995 on Build GRCh38
rs560933969 Chr.4: 80289151-80289151 on Build GRCh38
rs976984378 Chr.4: 80288850-80288850 on Build GRCh38
rs757293316 Chr.4: 80288878-80288878 on Build GRCh38
rs925934001 Chr.4: 80289252-80289252 on Build GRCh38
rs778980806 Chr.4: 80288882-80288882 on Build GRCh38
rs529411269 Chr.4: 80289072-80289072 on Build GRCh38
rs1017432321 Chr.4: 80288948-80288948 on Build GRCh38
rs145966495 Chr.4: 80288860-80288860 on Build GRCh38
rs983216384 Chr.4: 80288810-80288810 on Build GRCh38
rs979522350 Chr.4: 80289119-80289119 on Build GRCh38
rs903422586 Chr.4: 80288818-80288818 on Build GRCh38
rs1045936046 Chr.4: 80289299-80289299 on Build GRCh38
rs554490763 Chr.4: 80289150-80289150 on Build GRCh38

View Full Product Details