Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80289693-80290201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_001291812.1 NM_004464.3 NM_033143.2

SNP Detail

SNP ID Chromosome Location
rs993930892 Chr.4: 80289862-80289862 on Build GRCh38
rs911493236 Chr.4: 80289929-80289929 on Build GRCh38
rs879211662 Chr.4: 80290045-80290045 on Build GRCh38
rs764457205 Chr.4: 80290002-80290002 on Build GRCh38
rs567830864 Chr.4: 80290074-80290074 on Build GRCh38
rs375815495 Chr.4: 80289946-80289946 on Build GRCh38
rs529716811 Chr.4: 80290075-80290075 on Build GRCh38
rs922590042 Chr.4: 80289959-80289959 on Build GRCh38
rs1049446176 Chr.4: 80289807-80289807 on Build GRCh38
rs566578971 Chr.4: 80290089-80290089 on Build GRCh38
rs371374685 Chr.4: 80289840-80289840 on Build GRCh38
rs756762806 Chr.4: 80289857-80289857 on Build GRCh38
rs1032676878 Chr.4: 80289798-80289798 on Build GRCh38
rs4690149 Chr.4: 80289759-80289759 on Build GRCh38
rs142199666 Chr.4: 80290170-80290170 on Build GRCh38
rs1053690021 Chr.4: 80289791-80289791 on Build GRCh38
rs754301636 Chr.4: 80290083-80290083 on Build GRCh38
rs1053592859 Chr.4: 80290017-80290017 on Build GRCh38
rs540464512 Chr.4: 80289755-80289755 on Build GRCh38
rs970889707 Chr.4: 80289860-80289860 on Build GRCh38
rs902341441 Chr.4: 80289747-80289747 on Build GRCh38
rs997663468 Chr.4: 80289905-80289905 on Build GRCh38
rs143008601 Chr.4: 80289814-80289814 on Build GRCh38
rs751170831 Chr.4: 80289818-80289818 on Build GRCh38
rs191359720 Chr.4: 80289867-80289867 on Build GRCh38
rs1010464694 Chr.4: 80289963-80289963 on Build GRCh38
rs964268121 Chr.4: 80290128-80290128 on Build GRCh38
rs1046136748 Chr.4: 80290050-80290050 on Build GRCh38
rs889950308 Chr.4: 80289942-80289942 on Build GRCh38
rs369761424 Chr.4: 80289724-80289724 on Build GRCh38
rs893321294 Chr.4: 80290158-80290158 on Build GRCh38
rs565134375 Chr.4: 80290025-80290025 on Build GRCh38
rs958776672 Chr.4: 80290020-80290020 on Build GRCh38
rs1018365483 Chr.4: 80290112-80290112 on Build GRCh38
rs923799276 Chr.4: 80289983-80289983 on Build GRCh38
rs952898665 Chr.4: 80289887-80289887 on Build GRCh38
rs940595843 Chr.4: 80289824-80289824 on Build GRCh38
rs773728689 Chr.4: 80289974-80289974 on Build GRCh38
rs538725733 Chr.4: 80290136-80290136 on Build GRCh38
rs544312552 Chr.4: 80290022-80290022 on Build GRCh38
rs1007094091 Chr.4: 80289952-80289952 on Build GRCh38
rs936471030 Chr.4: 80289989-80289989 on Build GRCh38
rs964235624 Chr.4: 80289945-80289945 on Build GRCh38
rs35194320 Chr.4: 80290026-80290026 on Build GRCh38
rs866722954 Chr.4: 80290134-80290134 on Build GRCh38
rs545238400 Chr.4: 80289893-80289893 on Build GRCh38
rs1041796236 Chr.4: 80289971-80289971 on Build GRCh38
rs1034475285 Chr.4: 80290011-80290011 on Build GRCh38
rs565232020 Chr.4: 80289825-80289825 on Build GRCh38
rs17004872 Chr.4: 80290037-80290037 on Build GRCh38
rs1005690549 Chr.4: 80290090-80290090 on Build GRCh38
rs36028972 Chr.4: 80290082-80290082 on Build GRCh38
rs946161388 Chr.4: 80289961-80289961 on Build GRCh38
rs967389432 Chr.4: 80290149-80290149 on Build GRCh38
rs977931525 Chr.4: 80290152-80290152 on Build GRCh38

View Full Product Details