Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6588890-6589399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf26
Entrez Gene ID
79034
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
UniGene
Hs.487511
Gene Chromosome Location
Chr.7: 6589368-6608726 on Build GRCh38
Transcripts
XM_017012614.1 NM_001134387.1 NM_001134388.1 NM_001134389.1 NM_018106.3 XM_005249793.2 XM_005249794.3 XM_005249795.2 XM_005249796.3 XM_011515442.2 XM_017012391.1

Gene Symbol
ZDHHC4
Entrez Gene ID
55146
Gene Name
zinc finger DHHC-type containing 4
Gene Aliases
ZNF374
UniGene
Hs.5268
Gene Chromosome Location
Chr.7: 6577413-6588996 on Build GRCh38
Transcripts
XM_017012614.1 NM_001134387.1 NM_001134388.1 NM_001134389.1 NM_018106.3 XM_005249793.2 XM_005249794.3 XM_005249795.2 XM_005249796.3 XM_011515442.2 XM_017012391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390295 c.1017G>T p.E339D Chr.7: 6588892-6588892 on Build GRCh38
747272 c.1019G>T p.R340M Chr.7: 6588894-6588894 on Build GRCh38
7418321 c.1019G>A p.R340K Chr.7: 6588894-6588894 on Build GRCh38
4519643 c.1020G>T p.R340S Chr.7: 6588895-6588895 on Build GRCh38
4606733 c.1015G>C p.E339Q Chr.7: 6588890-6588890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998366383 Chr.7: 6589046-6589046 on Build GRCh38
rs773042493 Chr.7: 6589345-6589345 on Build GRCh38
rs929733274 Chr.7: 6589184-6589184 on Build GRCh38
rs370270228 Chr.7: 6588947-6588947 on Build GRCh38
rs766090238 Chr.7: 6589358-6589358 on Build GRCh38
rs530191628 Chr.7: 6589311-6589311 on Build GRCh38
rs544466054 Chr.7: 6589298-6589298 on Build GRCh38
rs906688810 Chr.7: 6589354-6589354 on Build GRCh38
rs970739894 Chr.7: 6588955-6588955 on Build GRCh38
rs916506565 Chr.7: 6589130-6589130 on Build GRCh38
rs7805414 Chr.7: 6589032-6589032 on Build GRCh38
rs998831672 Chr.7: 6589260-6589260 on Build GRCh38
rs927767834 Chr.7: 6589147-6589147 on Build GRCh38
rs945217735 Chr.7: 6589022-6589022 on Build GRCh38
rs915073972 Chr.7: 6589012-6589012 on Build GRCh38
rs909634910 Chr.7: 6589372-6589372 on Build GRCh38
rs1049322544 Chr.7: 6589308-6589310 on Build GRCh38
rs778465895 Chr.7: 6588915-6588915 on Build GRCh38
rs187446501 Chr.7: 6589136-6589136 on Build GRCh38
rs888634619 Chr.7: 6589101-6589101 on Build GRCh38
rs1003765295 Chr.7: 6589370-6589370 on Build GRCh38
rs1038940202 Chr.7: 6589289-6589290 on Build GRCh38
rs1033880966 Chr.7: 6589374-6589374 on Build GRCh38
rs191074188 Chr.7: 6589210-6589210 on Build GRCh38
rs977458929 Chr.7: 6589059-6589059 on Build GRCh38
rs569242853 Chr.7: 6588950-6588950 on Build GRCh38
rs774721746 Chr.7: 6589090-6589090 on Build GRCh38
rs1046791323 Chr.7: 6588973-6588973 on Build GRCh38
rs944623755 Chr.7: 6589285-6589286 on Build GRCh38
rs886180637 Chr.7: 6589334-6589334 on Build GRCh38
rs540875951 Chr.7: 6589251-6589251 on Build GRCh38
rs113477204 Chr.7: 6589266-6589266 on Build GRCh38
rs1023377562 Chr.7: 6589305-6589305 on Build GRCh38
rs142334144 Chr.7: 6589343-6589343 on Build GRCh38
rs947960345 Chr.7: 6589132-6589132 on Build GRCh38
rs771224663 Chr.7: 6589019-6589019 on Build GRCh38
rs929806795 Chr.7: 6589335-6589335 on Build GRCh38
rs542334530 Chr.7: 6589205-6589205 on Build GRCh38
rs969359384 Chr.7: 6589306-6589306 on Build GRCh38
rs572515342 Chr.7: 6589222-6589222 on Build GRCh38
rs547619012 Chr.7: 6589094-6589094 on Build GRCh38
rs1033066977 Chr.7: 6589297-6589297 on Build GRCh38
rs991867470 Chr.7: 6589109-6589109 on Build GRCh38
rs982004738 Chr.7: 6589308-6589308 on Build GRCh38
rs574971728 Chr.7: 6589186-6589186 on Build GRCh38
rs989557632 Chr.7: 6588980-6588980 on Build GRCh38
rs866818319 Chr.7: 6588968-6588968 on Build GRCh38
rs3813514 Chr.7: 6588956-6588956 on Build GRCh38
rs563105290 Chr.7: 6589301-6589301 on Build GRCh38
rs1004380074 Chr.7: 6589129-6589129 on Build GRCh38
rs577913682 Chr.7: 6589072-6589072 on Build GRCh38
rs995007430 Chr.7: 6589097-6589097 on Build GRCh38
rs970964369 Chr.7: 6589173-6589173 on Build GRCh38
rs538444008 Chr.7: 6589105-6589105 on Build GRCh38
rs764462585 Chr.7: 6588944-6588944 on Build GRCh38
rs762075658 Chr.7: 6588948-6588948 on Build GRCh38
rs1015720670 Chr.7: 6589346-6589346 on Build GRCh38
rs760674781 Chr.7: 6588921-6588921 on Build GRCh38
rs998951534 Chr.7: 6589272-6589272 on Build GRCh38
rs982578242 Chr.7: 6589352-6589352 on Build GRCh38
rs369818962 Chr.7: 6588957-6588957 on Build GRCh38
rs1005605242 Chr.7: 6589023-6589023 on Build GRCh38
rs957382120 Chr.7: 6589093-6589093 on Build GRCh38
rs550001004 Chr.7: 6589050-6589050 on Build GRCh38
rs552993025 Chr.7: 6589069-6589069 on Build GRCh38
rs150061103 Chr.7: 6589261-6589261 on Build GRCh38
rs112386861 Chr.7: 6588967-6588967 on Build GRCh38
rs879037705 Chr.7: 6588997-6588997 on Build GRCh38
rs775709383 Chr.7: 6588943-6588943 on Build GRCh38
rs938098783 Chr.7: 6588978-6588978 on Build GRCh38
rs768367859 Chr.7: 6589214-6589214 on Build GRCh38
rs766166073 Chr.7: 6588938-6588938 on Build GRCh38
rs749764669 Chr.7: 6589018-6589018 on Build GRCh38
rs900144499 Chr.7: 6589051-6589051 on Build GRCh38
rs1027391995 Chr.7: 6589098-6589098 on Build GRCh38
rs927841370 Chr.7: 6589323-6589323 on Build GRCh38
rs548717057 Chr.7: 6589317-6589317 on Build GRCh38
rs964609828 Chr.7: 6589045-6589045 on Build GRCh38
rs967465435 Chr.7: 6589056-6589056 on Build GRCh38

View Full Product Details