Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 92489113-92489613 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
UniGene
Hs.164682
Gene Chromosome Location
Chr.7: 92487023-92528531 on Build GRCh38
Transcripts
NM_000466.2 NM_001282677.1 NM_001282678.1 XM_017012319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778688 c.3683C>T p.A1228V Chr.7: 92489377-92489377 on Build GRCh38
1683639 c.3731_3732insCATTA p.S1246fs*37 Chr.7: 92489328-92489329 on Build GRCh38
3883365 c.3739G>A p.E1247K Chr.7: 92489321-92489321 on Build GRCh38
5873366 c.3705G>T p.M1235I Chr.7: 92489355-92489355 on Build GRCh38
6111429 c.3743A>G p.D1248G Chr.7: 92489317-92489317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34825053 Chr.7: 92489340-92489340 on Build GRCh38
rs769844398 Chr.7: 92489316-92489316 on Build GRCh38
rs184938084 Chr.7: 92489451-92489451 on Build GRCh38
rs999217020 Chr.7: 92489411-92489411 on Build GRCh38
rs781728903 Chr.7: 92489256-92489256 on Build GRCh38
rs745692619 Chr.7: 92489291-92489291 on Build GRCh38
rs928197279 Chr.7: 92489485-92489485 on Build GRCh38
rs750604651 Chr.7: 92489583-92489583 on Build GRCh38
rs552665097 Chr.7: 92489199-92489199 on Build GRCh38
rs770743353 Chr.7: 92489281-92489281 on Build GRCh38
rs766682184 Chr.7: 92489368-92489368 on Build GRCh38
rs765998871 Chr.7: 92489345-92489345 on Build GRCh38
rs758670883 Chr.7: 92489243-92489243 on Build GRCh38
rs773322114 Chr.7: 92489326-92489326 on Build GRCh38
rs769737360 Chr.7: 92489191-92489191 on Build GRCh38
rs571336413 Chr.7: 92489215-92489215 on Build GRCh38
rs35892336 Chr.7: 92489180-92489180 on Build GRCh38
rs752029825 Chr.7: 92489252-92489252 on Build GRCh38
rs182526018 Chr.7: 92489160-92489160 on Build GRCh38
rs982775512 Chr.7: 92489483-92489483 on Build GRCh38
rs751190818 Chr.7: 92489551-92489551 on Build GRCh38
rs999329609 Chr.7: 92489391-92489391 on Build GRCh38
rs189902683 Chr.7: 92489244-92489244 on Build GRCh38
rs931089270 Chr.7: 92489582-92489582 on Build GRCh38
rs746001220 Chr.7: 92489331-92489331 on Build GRCh38
rs761835242 Chr.7: 92489313-92489313 on Build GRCh38
rs190190301 Chr.7: 92489535-92489535 on Build GRCh38
rs367814599 Chr.7: 92489442-92489442 on Build GRCh38
rs536213690 Chr.7: 92489277-92489277 on Build GRCh38
rs756953510 Chr.7: 92489421-92489421 on Build GRCh38
rs777384403 Chr.7: 92489418-92489418 on Build GRCh38
rs863225085 Chr.7: 92489310-92489310 on Build GRCh38
rs773087914 Chr.7: 92489468-92489468 on Build GRCh38
rs1048574377 Chr.7: 92489337-92489337 on Build GRCh38
rs200312888 Chr.7: 92489467-92489468 on Build GRCh38
rs142422529 Chr.7: 92489226-92489226 on Build GRCh38
rs778565066 Chr.7: 92489282-92489282 on Build GRCh38
rs372959546 Chr.7: 92489254-92489254 on Build GRCh38
rs200306465 Chr.7: 92489410-92489410 on Build GRCh38
rs969384021 Chr.7: 92489481-92489481 on Build GRCh38
rs916565234 Chr.7: 92489189-92489189 on Build GRCh38
rs368274049 Chr.7: 92489373-92489373 on Build GRCh38
rs769836601 Chr.7: 92489364-92489367 on Build GRCh38
rs561270415 Chr.7: 92489159-92489159 on Build GRCh38
rs1019900850 Chr.7: 92489139-92489139 on Build GRCh38
rs775743560 Chr.7: 92489430-92489431 on Build GRCh38
rs778635449 Chr.7: 92489431-92489431 on Build GRCh38
rs755715309 Chr.7: 92489273-92489273 on Build GRCh38
rs483352694 Chr.7: 92489415-92489415 on Build GRCh38
rs781629883 Chr.7: 92489277-92489277 on Build GRCh38
rs912545547 Chr.7: 92489545-92489545 on Build GRCh38
rs901023979 Chr.7: 92489379-92489379 on Build GRCh38
rs751910989 Chr.7: 92489496-92489498 on Build GRCh38
rs375966274 Chr.7: 92489260-92489260 on Build GRCh38
rs745612436 Chr.7: 92489437-92489437 on Build GRCh38
rs920760510 Chr.7: 92489580-92489580 on Build GRCh38
rs1025543038 Chr.7: 92489528-92489528 on Build GRCh38
rs371575516 Chr.7: 92489395-92489395 on Build GRCh38
rs753461425 Chr.7: 92489420-92489420 on Build GRCh38
rs749534850 Chr.7: 92489442-92489442 on Build GRCh38
rs989580336 Chr.7: 92489426-92489426 on Build GRCh38
rs376375286 Chr.7: 92489462-92489462 on Build GRCh38
rs912670552 Chr.7: 92489219-92489219 on Build GRCh38
rs768692127 Chr.7: 92489457-92489458 on Build GRCh38
rs951282371 Chr.7: 92489532-92489532 on Build GRCh38
rs112833664 Chr.7: 92489183-92489183 on Build GRCh38
rs553001596 Chr.7: 92489304-92489304 on Build GRCh38
rs575723134 Chr.7: 92489497-92489497 on Build GRCh38
rs180871355 Chr.7: 92489446-92489446 on Build GRCh38
rs746919622 Chr.7: 92489450-92489450 on Build GRCh38
rs116367709 Chr.7: 92489166-92489166 on Build GRCh38
rs398123410 Chr.7: 92489346-92489346 on Build GRCh38
rs375213175 Chr.7: 92489298-92489298 on Build GRCh38
rs370369794 Chr.7: 92489280-92489280 on Build GRCh38
rs767870400 Chr.7: 92489396-92489396 on Build GRCh38

View Full Product Details