Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11493826-11494326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543827.1 XM_011543828.2 XM_011543829.2 XM_017013730.1

SNP Detail

SNP ID Chromosome Location
rs948954100 Chr.8: 11494210-11494210 on Build GRCh38
rs943083102 Chr.8: 11493858-11493858 on Build GRCh38
rs527736826 Chr.8: 11493998-11493998 on Build GRCh38
rs907225253 Chr.8: 11494218-11494218 on Build GRCh38
rs549066952 Chr.8: 11493999-11493999 on Build GRCh38
rs917474440 Chr.8: 11494156-11494156 on Build GRCh38
rs907296829 Chr.8: 11493980-11493980 on Build GRCh38
rs965254358 Chr.8: 11494284-11494284 on Build GRCh38
rs945099715 Chr.8: 11493903-11493903 on Build GRCh38
rs936170074 Chr.8: 11493939-11493939 on Build GRCh38
rs567463835 Chr.8: 11494049-11494049 on Build GRCh38
rs975383886 Chr.8: 11494075-11494075 on Build GRCh38
rs143339410 Chr.8: 11493923-11493923 on Build GRCh38
rs533285601 Chr.8: 11493943-11493943 on Build GRCh38
rs1017832341 Chr.8: 11494282-11494282 on Build GRCh38
rs538706235 Chr.8: 11494062-11494062 on Build GRCh38
rs975787320 Chr.8: 11494144-11494144 on Build GRCh38
rs984614223 Chr.8: 11494067-11494067 on Build GRCh38
rs142686759 Chr.8: 11494152-11494152 on Build GRCh38
rs964947096 Chr.8: 11494133-11494133 on Build GRCh38
rs766384358 Chr.8: 11493935-11493935 on Build GRCh38
rs544845115 Chr.8: 11493952-11493952 on Build GRCh38
rs896140070 Chr.8: 11493975-11493975 on Build GRCh38
rs886062589 Chr.8: 11494118-11494118 on Build GRCh38
rs571822414 Chr.8: 11494149-11494149 on Build GRCh38
rs921327617 Chr.8: 11493935-11493935 on Build GRCh38
rs148891021 Chr.8: 11494145-11494145 on Build GRCh38
rs1017527986 Chr.8: 11494077-11494077 on Build GRCh38
rs1000625079 Chr.8: 11494019-11494019 on Build GRCh38
rs145636076 Chr.8: 11493925-11493925 on Build GRCh38
rs952865943 Chr.8: 11494125-11494125 on Build GRCh38
rs3021519 Chr.8: 11494083-11494083 on Build GRCh38
rs936181501 Chr.8: 11494196-11494196 on Build GRCh38
rs1032589828 Chr.8: 11494027-11494027 on Build GRCh38
rs984727380 Chr.8: 11493856-11493856 on Build GRCh38
rs755284236 Chr.8: 11494227-11494232 on Build GRCh38
rs1009380343 Chr.8: 11494251-11494251 on Build GRCh38
rs772594702 Chr.8: 11494167-11494167 on Build GRCh38
rs1040409487 Chr.8: 11493875-11493875 on Build GRCh38
rs543839949 Chr.8: 11494270-11494270 on Build GRCh38
rs979648422 Chr.8: 11493870-11493870 on Build GRCh38
rs1026433588 Chr.8: 11494257-11494257 on Build GRCh38
rs894686074 Chr.8: 11493956-11493956 on Build GRCh38
rs566280995 Chr.8: 11494165-11494165 on Build GRCh38
rs1001017033 Chr.8: 11494187-11494187 on Build GRCh38
rs988116681 Chr.8: 11493892-11493892 on Build GRCh38
rs984261346 Chr.8: 11494126-11494126 on Build GRCh38
rs1028739613 Chr.8: 11494299-11494299 on Build GRCh38
rs142333989 Chr.8: 11494302-11494302 on Build GRCh38
rs1009474958 Chr.8: 11494034-11494034 on Build GRCh38
rs777933050 Chr.8: 11494199-11494199 on Build GRCh38
rs976428961 Chr.8: 11494292-11494292 on Build GRCh38
rs540630245 Chr.8: 11494215-11494215 on Build GRCh38
rs139930773 Chr.8: 11493891-11493891 on Build GRCh38
rs535949326 Chr.8: 11493851-11493851 on Build GRCh38
rs904909204 Chr.8: 11494013-11494014 on Build GRCh38
rs1054994732 Chr.8: 11494204-11494204 on Build GRCh38
rs573814484 Chr.8: 11494240-11494240 on Build GRCh38
rs554033678 Chr.8: 11494153-11494153 on Build GRCh38
rs765870935 Chr.8: 11494261-11494261 on Build GRCh38
rs558341178 Chr.8: 11494129-11494129 on Build GRCh38
rs975478356 Chr.8: 11493928-11493928 on Build GRCh38
rs151046937 Chr.8: 11494289-11494289 on Build GRCh38
rs536895669 Chr.8: 11494178-11494178 on Build GRCh38
rs925028300 Chr.8: 11494176-11494176 on Build GRCh38
rs886062590 Chr.8: 11494218-11494218 on Build GRCh38
rs956602115 Chr.8: 11494032-11494032 on Build GRCh38
rs892276111 Chr.8: 11494223-11494223 on Build GRCh38
rs761744676 Chr.8: 11494109-11494109 on Build GRCh38
rs965308447 Chr.8: 11494074-11494074 on Build GRCh38
rs910457352 Chr.8: 11493857-11493857 on Build GRCh38
rs183594315 Chr.8: 11493985-11493985 on Build GRCh38
rs1030246926 Chr.8: 11494294-11494294 on Build GRCh38
rs555491986 Chr.8: 11494207-11494207 on Build GRCh38
rs1024706443 Chr.8: 11493974-11493974 on Build GRCh38
rs1012924306 Chr.8: 11493963-11493963 on Build GRCh38
rs115316286 Chr.8: 11494063-11494063 on Build GRCh38
rs894747651 Chr.8: 11494208-11494208 on Build GRCh38

View Full Product Details