Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92474475-92474939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASPN
Entrez Gene ID
54829
Gene Name
asporin
Gene Aliases
OS3, PLAP-1, PLAP1, SLRR1C
UniGene
Hs.435655
Gene Chromosome Location
Chr.9: 92456205-92482562 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Gene Symbol
CENPP
Entrez Gene ID
401541
Gene Name
centromere protein P
Gene Aliases
CENP-P
UniGene
Hs.713775
Gene Chromosome Location
Chr.9: 92325457-92615164 on Build GRCh38
Transcripts
NM_001193335.1 NM_017680.4 NM_001012267.2 NM_001286969.1 XM_011518689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5987132 c.150T>G p.D50E Chr.9: 92474748-92474748 on Build GRCh38
3664469 c.77T>G p.L26R Chr.9: 92474821-92474821 on Build GRCh38
5625873 c.110C>A p.T37K Chr.9: 92474788-92474788 on Build GRCh38
3736573 c.155delA p.E52fs*40 Chr.9: 92474743-92474743 on Build GRCh38
5045703 c.117T>A p.D39E Chr.9: 92474781-92474781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144125864 Chr.9: 92474809-92474809 on Build GRCh38
rs766925567 Chr.9: 92474790-92474790 on Build GRCh38
rs368037584 Chr.9: 92474516-92474517 on Build GRCh38
rs763982158 Chr.9: 92474801-92474801 on Build GRCh38
rs757077133 Chr.9: 92474904-92474904 on Build GRCh38
rs774262129 Chr.9: 92474845-92474845 on Build GRCh38
rs140557213 Chr.9: 92474859-92474859 on Build GRCh38
rs796670703 Chr.9: 92474745-92474746 on Build GRCh38
rs377008610 Chr.9: 92474737-92474737 on Build GRCh38
rs766571754 Chr.9: 92474908-92474908 on Build GRCh38
rs867474586 Chr.9: 92474750-92474751 on Build GRCh38
rs370182715 Chr.9: 92474738-92474738 on Build GRCh38
rs76454027 Chr.9: 92474713-92474713 on Build GRCh38
rs760328846 Chr.9: 92474768-92474768 on Build GRCh38
rs749263045 Chr.9: 92474831-92474831 on Build GRCh38
rs762740047 Chr.9: 92474596-92474596 on Build GRCh38
rs148382623 Chr.9: 92474648-92474648 on Build GRCh38
rs3078372 Chr.9: 92474742-92474743 on Build GRCh38
rs373082643 Chr.9: 92474889-92474889 on Build GRCh38
rs71362392 Chr.9: 92474786-92474787 on Build GRCh38
rs113747060 Chr.9: 92474774-92474775 on Build GRCh38
rs387906276 Chr.9: 92474748-92474786 on Build GRCh38
rs139505803 Chr.9: 92474767-92474767 on Build GRCh38
rs756130551 Chr.9: 92474678-92474678 on Build GRCh38
rs147354437 Chr.9: 92474757-92474757 on Build GRCh38
rs749033215 Chr.9: 92474759-92474759 on Build GRCh38
rs150545989 Chr.9: 92474766-92474766 on Build GRCh38
rs145549450 Chr.9: 92474782-92474782 on Build GRCh38
rs1025635648 Chr.9: 92474799-92474799 on Build GRCh38
rs769561020 Chr.9: 92474577-92474577 on Build GRCh38
rs535881047 Chr.9: 92474521-92474521 on Build GRCh38
rs375376579 Chr.9: 92474797-92474797 on Build GRCh38
rs72537392 Chr.9: 92474766-92474767 on Build GRCh38
rs772484367 Chr.9: 92474758-92474758 on Build GRCh38
rs952838000 Chr.9: 92474548-92474548 on Build GRCh38
rs759019590 Chr.9: 92474787-92474789 on Build GRCh38
rs143279922 Chr.9: 92474748-92474748 on Build GRCh38
rs780141507 Chr.9: 92474821-92474821 on Build GRCh38
rs758206044 Chr.9: 92474799-92474807 on Build GRCh38
rs577647123 Chr.9: 92474514-92474514 on Build GRCh38
rs774762901 Chr.9: 92474755-92474756 on Build GRCh38
rs866462451 Chr.9: 92474783-92474786 on Build GRCh38
rs377478533 Chr.9: 92474830-92474830 on Build GRCh38
rs758945968 Chr.9: 92474639-92474639 on Build GRCh38
rs139705272 Chr.9: 92474510-92474510 on Build GRCh38
rs775038925 Chr.9: 92474862-92474862 on Build GRCh38
rs574970813 Chr.9: 92474848-92474848 on Build GRCh38
rs762235608 Chr.9: 92474757-92474758 on Build GRCh38
rs558726949 Chr.9: 92474697-92474697 on Build GRCh38
rs758586100 Chr.9: 92474811-92474811 on Build GRCh38
rs772930777 Chr.9: 92474591-92474591 on Build GRCh38
rs759099503 Chr.9: 92474502-92474502 on Build GRCh38
rs754422619 Chr.9: 92474803-92474803 on Build GRCh38
rs752632322 Chr.9: 92474798-92474803 on Build GRCh38
rs569981850 Chr.9: 92474555-92474555 on Build GRCh38
rs778441508 Chr.9: 92474672-92474672 on Build GRCh38
rs557103556 Chr.9: 92474784-92474786 on Build GRCh38
rs186840940 Chr.9: 92474873-92474873 on Build GRCh38
rs796401010 Chr.9: 92474744-92474745 on Build GRCh38
rs397838876 Chr.9: 92474782-92474784 on Build GRCh38
rs200538582 Chr.9: 92474745-92474745 on Build GRCh38
rs867716455 Chr.9: 92474770-92474770 on Build GRCh38
rs752504095 Chr.9: 92474739-92474741 on Build GRCh38
rs563486617 Chr.9: 92474717-92474717 on Build GRCh38
rs765839278 Chr.9: 92474786-92474795 on Build GRCh38
rs796984145 Chr.9: 92474740-92474741 on Build GRCh38
rs878929025 Chr.9: 92474746-92474748 on Build GRCh38
rs984130953 Chr.9: 92474594-92474594 on Build GRCh38
rs111984514 Chr.9: 92474741-92474742 on Build GRCh38
rs750729556 Chr.9: 92474899-92474899 on Build GRCh38
rs764248772 Chr.9: 92474745-92474746 on Build GRCh38
rs778526424 Chr.9: 92474697-92474697 on Build GRCh38
rs777269498 Chr.9: 92474885-92474885 on Build GRCh38
rs763948305 Chr.9: 92474688-92474688 on Build GRCh38
rs779232502 Chr.9: 92474686-92474686 on Build GRCh38
rs769182352 Chr.9: 92474750-92474750 on Build GRCh38
rs374980467 Chr.9: 92474741-92474741 on Build GRCh38
rs765974567 Chr.9: 92474608-92474608 on Build GRCh38
rs758191999 Chr.9: 92474739-92474740 on Build GRCh38
rs775007730 Chr.9: 92474791-92474791 on Build GRCh38
rs878983183 Chr.9: 92474743-92474748 on Build GRCh38
rs754977945 Chr.9: 92474682-92474682 on Build GRCh38
rs535145508 Chr.9: 92474807-92474807 on Build GRCh38
rs879047429 Chr.9: 92474748-92474749 on Build GRCh38
rs772794867 Chr.9: 92474765-92474766 on Build GRCh38
rs780311826 Chr.9: 92474747-92474748 on Build GRCh38
rs886658639 Chr.9: 92474734-92474734 on Build GRCh38
rs938973178 Chr.9: 92474642-92474642 on Build GRCh38
rs145092122 Chr.9: 92474761-92474762 on Build GRCh38
rs769651044 Chr.9: 92474722-92474722 on Build GRCh38
rs892101188 Chr.9: 92474752-92474752 on Build GRCh38
rs780235435 Chr.9: 92474700-92474700 on Build GRCh38
rs770709159 Chr.9: 92474841-92474841 on Build GRCh38
rs759299671 Chr.9: 92474775-92474775 on Build GRCh38
rs745665400 Chr.9: 92474851-92474851 on Build GRCh38
rs763749029 Chr.9: 92474875-92474875 on Build GRCh38
rs760268622 Chr.9: 92474793-92474793 on Build GRCh38
rs751247045 Chr.9: 92474615-92474615 on Build GRCh38
rs773845974 Chr.9: 92474773-92474773 on Build GRCh38
rs369738156 Chr.9: 92474644-92474644 on Build GRCh38
rs536939419 Chr.9: 92474647-92474647 on Build GRCh38
rs376433743 Chr.9: 92474742-92474742 on Build GRCh38
rs146516859 Chr.9: 92474825-92474825 on Build GRCh38
rs747069469 Chr.9: 92474703-92474703 on Build GRCh38
rs576919751 Chr.9: 92474768-92474768 on Build GRCh38
rs776230517 Chr.9: 92474787-92474798 on Build GRCh38
rs749708861 Chr.9: 92474748-92474748 on Build GRCh38
rs139405419 Chr.9: 92474763-92474763 on Build GRCh38
rs141446497 Chr.9: 92474531-92474531 on Build GRCh38
rs906292782 Chr.9: 92474619-92474619 on Build GRCh38
rs765673267 Chr.9: 92474891-92474891 on Build GRCh38

View Full Product Details