Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 85546797-85547339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGTPBP1
Entrez Gene ID
23287
Gene Name
ATP/GTP binding protein 1
Gene Aliases
CCP1, NNA1
UniGene
Hs.719980
Gene Chromosome Location
Chr.9: 85543799-85742029 on Build GRCh38
Transcripts
NM_001286715.1 NM_001286717.1 NM_015239.2 XM_005251847.3 XM_005251848.2 XM_005251849.3 XM_011518420.2 XM_017014544.1 XM_017014546.1 XM_017014548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4139001 c.3552A>T p.E1184D Chr.9: 85547238-85547238 on Build GRCh38
5580168 c.3533C>T p.S1178F Chr.9: 85547137-85547137 on Build GRCh38
287512 c.3445G>A p.A1149T Chr.9: 85547225-85547225 on Build GRCh38
4660427 c.3491_3493delGAG p.G1164delG Chr.9: 85547177-85547179 on Build GRCh38
6321468 c.3552C>A p.Y1184* Chr.9: 85547118-85547118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138412470 Chr.9: 85547151-85547151 on Build GRCh38
rs921749241 Chr.9: 85547157-85547157 on Build GRCh38
rs1054193380 Chr.9: 85547203-85547203 on Build GRCh38
rs56148123 Chr.9: 85547075-85547075 on Build GRCh38
rs761006979 Chr.9: 85547268-85547268 on Build GRCh38
rs779901525 Chr.9: 85547169-85547169 on Build GRCh38
rs1012427328 Chr.9: 85546947-85546947 on Build GRCh38
rs914335151 Chr.9: 85547258-85547258 on Build GRCh38
rs774787421 Chr.9: 85547251-85547251 on Build GRCh38
rs371574763 Chr.9: 85547078-85547078 on Build GRCh38
rs765145161 Chr.9: 85547158-85547158 on Build GRCh38
rs967892678 Chr.9: 85547247-85547247 on Build GRCh38
rs368433383 Chr.9: 85547125-85547125 on Build GRCh38
rs150752721 Chr.9: 85547274-85547274 on Build GRCh38
rs374990965 Chr.9: 85547072-85547072 on Build GRCh38
rs770161230 Chr.9: 85547107-85547107 on Build GRCh38
rs946620705 Chr.9: 85546901-85546901 on Build GRCh38
rs944792438 Chr.9: 85546855-85546855 on Build GRCh38
rs372287584 Chr.9: 85547147-85547147 on Build GRCh38
rs377366142 Chr.9: 85547086-85547086 on Build GRCh38
rs534198724 Chr.9: 85547002-85547002 on Build GRCh38
rs774901271 Chr.9: 85547143-85547143 on Build GRCh38
rs747824329 Chr.9: 85547201-85547201 on Build GRCh38
rs763549504 Chr.9: 85547131-85547131 on Build GRCh38
rs147024207 Chr.9: 85546983-85546983 on Build GRCh38
rs924113867 Chr.9: 85546855-85546855 on Build GRCh38
rs577331626 Chr.9: 85547045-85547045 on Build GRCh38
rs921778605 Chr.9: 85546896-85546896 on Build GRCh38
rs200269013 Chr.9: 85547105-85547105 on Build GRCh38
rs760053644 Chr.9: 85547252-85547252 on Build GRCh38
rs201905471 Chr.9: 85547249-85547249 on Build GRCh38
rs1043646855 Chr.9: 85546908-85546908 on Build GRCh38
rs775944173 Chr.9: 85547263-85547263 on Build GRCh38
rs560864897 Chr.9: 85547196-85547196 on Build GRCh38
rs558578947 Chr.9: 85547097-85547097 on Build GRCh38
rs148198029 Chr.9: 85547199-85547199 on Build GRCh38
rs111654098 Chr.9: 85547297-85547297 on Build GRCh38
rs796786669 Chr.9: 85546854-85546855 on Build GRCh38
rs749664353 Chr.9: 85547237-85547237 on Build GRCh38
rs746537872 Chr.9: 85547098-85547098 on Build GRCh38
rs772290733 Chr.9: 85547256-85547256 on Build GRCh38
rs376720540 Chr.9: 85547293-85547293 on Build GRCh38
rs902183690 Chr.9: 85546918-85546918 on Build GRCh38
rs769398588 Chr.9: 85547218-85547218 on Build GRCh38
rs771315001 Chr.9: 85547241-85547241 on Build GRCh38
rs757185840 Chr.9: 85547065-85547065 on Build GRCh38
rs529742922 Chr.9: 85547224-85547224 on Build GRCh38
rs184923853 Chr.9: 85547053-85547053 on Build GRCh38
rs762710629 Chr.9: 85547303-85547303 on Build GRCh38
rs896197791 Chr.9: 85546934-85546934 on Build GRCh38
rs750142860 Chr.9: 85547167-85547167 on Build GRCh38
rs997349788 Chr.9: 85546998-85546998 on Build GRCh38
rs1044569835 Chr.9: 85546975-85546975 on Build GRCh38
rs374016566 Chr.9: 85547150-85547150 on Build GRCh38
rs554075625 Chr.9: 85547019-85547019 on Build GRCh38
rs770802295 Chr.9: 85547121-85547121 on Build GRCh38
rs934376422 Chr.9: 85546859-85546859 on Build GRCh38
rs371388579 Chr.9: 85547104-85547104 on Build GRCh38
rs192876044 Chr.9: 85546924-85546924 on Build GRCh38
rs951043644 Chr.9: 85547060-85547060 on Build GRCh38
rs746947692 Chr.9: 85547181-85547181 on Build GRCh38
rs770332799 Chr.9: 85547262-85547262 on Build GRCh38
rs758218580 Chr.9: 85547094-85547094 on Build GRCh38
rs141421425 Chr.9: 85546919-85546919 on Build GRCh38
rs746058778 Chr.9: 85547200-85547200 on Build GRCh38
rs550189746 Chr.9: 85546873-85546873 on Build GRCh38
rs758178425 Chr.9: 85547168-85547168 on Build GRCh38
rs766837669 Chr.9: 85547137-85547137 on Build GRCh38
rs1038937441 Chr.9: 85546933-85546933 on Build GRCh38
rs1054628442 Chr.9: 85546872-85546872 on Build GRCh38
rs576824120 Chr.9: 85546987-85546987 on Build GRCh38
rs766864819 Chr.9: 85547047-85547047 on Build GRCh38

View Full Product Details