Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55000244-55000745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APEX2
Entrez Gene ID
27301
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
Gene Aliases
APE2, APEXL2, XTH2, ZGRF2
UniGene
Hs.659558
Gene Chromosome Location
Chr.X: 55000323-55007873 on Build GRCh38
Transcripts
NM_001271748.1 NM_014481.3

SNP Detail

SNP ID Chromosome Location
rs1010317868 Chr.X: 55000698-55000698 on Build GRCh38
rs3180303 Chr.X: 55000514-55000514 on Build GRCh38
rs940700336 Chr.X: 55000463-55000463 on Build GRCh38
rs138420763 Chr.X: 55000472-55000472 on Build GRCh38
rs374430929 Chr.X: 55000467-55000467 on Build GRCh38
rs781610550 Chr.X: 55000505-55000505 on Build GRCh38
rs377755025 Chr.X: 55000582-55000582 on Build GRCh38
rs748389498 Chr.X: 55000510-55000510 on Build GRCh38
rs755725464 Chr.X: 55000374-55000374 on Build GRCh38
rs765763585 Chr.X: 55000270-55000271 on Build GRCh38
rs748384479 Chr.X: 55000395-55000395 on Build GRCh38
rs891475723 Chr.X: 55000334-55000334 on Build GRCh38
rs1032704171 Chr.X: 55000324-55000324 on Build GRCh38
rs755351199 Chr.X: 55000496-55000496 on Build GRCh38
rs1008681 Chr.X: 55000413-55000413 on Build GRCh38
rs751875481 Chr.X: 55000352-55000352 on Build GRCh38
rs3180304 Chr.X: 55000577-55000577 on Build GRCh38
rs954551721 Chr.X: 55000338-55000338 on Build GRCh38
rs185394253 Chr.X: 55000700-55000700 on Build GRCh38
rs952157873 Chr.X: 55000403-55000403 on Build GRCh38
rs1009878058 Chr.X: 55000350-55000350 on Build GRCh38
rs867038858 Chr.X: 55000429-55000429 on Build GRCh38
rs760477098 Chr.X: 55000608-55000608 on Build GRCh38
rs759665888 Chr.X: 55000398-55000398 on Build GRCh38
rs777916232 Chr.X: 55000521-55000521 on Build GRCh38
rs956911789 Chr.X: 55000386-55000386 on Build GRCh38
rs753693216 Chr.X: 55000414-55000414 on Build GRCh38
rs370230204 Chr.X: 55000533-55000533 on Build GRCh38
rs764768861 Chr.X: 55000630-55000630 on Build GRCh38
rs747345054 Chr.X: 55000387-55000387 on Build GRCh38
rs780638793 Chr.X: 55000381-55000381 on Build GRCh38
rs750368585 Chr.X: 55000551-55000551 on Build GRCh38
rs988448378 Chr.X: 55000404-55000404 on Build GRCh38
rs45566434 Chr.X: 55000310-55000310 on Build GRCh38
rs377350221 Chr.X: 55000375-55000375 on Build GRCh38
rs1044249250 Chr.X: 55000303-55000303 on Build GRCh38
rs769012529 Chr.X: 55000389-55000389 on Build GRCh38
rs763849554 Chr.X: 55000612-55000612 on Build GRCh38
rs935798554 Chr.X: 55000558-55000558 on Build GRCh38
rs1005854446 Chr.X: 55000274-55000274 on Build GRCh38
rs776370239 Chr.X: 55000615-55000615 on Build GRCh38
rs752060931 Chr.X: 55000478-55000478 on Build GRCh38
rs747534167 Chr.X: 55000703-55000703 on Build GRCh38
rs949867579 Chr.X: 55000288-55000288 on Build GRCh38
rs760389136 Chr.X: 55000426-55000426 on Build GRCh38
rs28382675 Chr.X: 55000417-55000417 on Build GRCh38
rs781296546 Chr.X: 55000394-55000394 on Build GRCh38
rs762955444 Chr.X: 55000411-55000411 on Build GRCh38
rs868318810 Chr.X: 55000363-55000363 on Build GRCh38
rs895704106 Chr.X: 55000283-55000283 on Build GRCh38
rs752340406 Chr.X: 55000420-55000420 on Build GRCh38
rs143050863 Chr.X: 55000515-55000515 on Build GRCh38
rs1018386984 Chr.X: 55000362-55000362 on Build GRCh38
rs775349519 Chr.X: 55000603-55000603 on Build GRCh38
rs770958646 Chr.X: 55000539-55000539 on Build GRCh38
rs1054628637 Chr.X: 55000665-55000665 on Build GRCh38
rs757988525 Chr.X: 55000640-55000641 on Build GRCh38
rs762570244 Chr.X: 55000464-55000465 on Build GRCh38
rs759281654 Chr.X: 55000416-55000416 on Build GRCh38
rs763762928 Chr.X: 55000447-55000447 on Build GRCh38
rs907928140 Chr.X: 55000418-55000418 on Build GRCh38
rs773659302 Chr.X: 55000299-55000299 on Build GRCh38
rs761436446 Chr.X: 55000618-55000618 on Build GRCh38
rs780488233 Chr.X: 55000695-55000695 on Build GRCh38
rs763069640 Chr.X: 55000305-55000305 on Build GRCh38
rs11541055 Chr.X: 55000454-55000454 on Build GRCh38
rs949581968 Chr.X: 55000415-55000415 on Build GRCh38
rs745795021 Chr.X: 55000573-55000573 on Build GRCh38
rs753400276 Chr.X: 55000451-55000451 on Build GRCh38
rs758966982 Chr.X: 55000376-55000376 on Build GRCh38
rs918143644 Chr.X: 55000405-55000405 on Build GRCh38

View Full Product Details