Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14474304-14474770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB1
Entrez Gene ID
1315
Gene Name
coatomer protein complex subunit beta 1
Gene Aliases
COPB
UniGene
Hs.339278
Gene Chromosome Location
Chr.11: 14457503-14499895 on Build GRCh38
Transcripts
NM_001144061.1 NM_001144062.1 NM_016451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6068173 c.1667C>T p.S556F Chr.11: 14474565-14474565 on Build GRCh38
135998 c.1669C>T p.L557F Chr.11: 14474563-14474563 on Build GRCh38
5361044 c.1675A>G p.T559A Chr.11: 14474557-14474557 on Build GRCh38
6634030 c.1653C>A p.F551L Chr.11: 14474579-14474579 on Build GRCh38
4019608 c.1701C>T p.R567R Chr.11: 14474531-14474531 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs907731621 Chr.11: 14474651-14474651 on Build GRCh38
rs906735743 Chr.11: 14474720-14474720 on Build GRCh38
rs893789876 Chr.11: 14474748-14474748 on Build GRCh38
rs1028246345 Chr.11: 14474433-14474433 on Build GRCh38
rs747965899 Chr.11: 14474519-14474519 on Build GRCh38
rs750668881 Chr.11: 14474463-14474463 on Build GRCh38
rs932699446 Chr.11: 14474717-14474717 on Build GRCh38
rs572770517 Chr.11: 14474342-14474342 on Build GRCh38
rs541311707 Chr.11: 14474346-14474346 on Build GRCh38
rs765492249 Chr.11: 14474460-14474460 on Build GRCh38
rs764638441 Chr.11: 14474597-14474597 on Build GRCh38
rs151120842 Chr.11: 14474554-14474554 on Build GRCh38
rs901205447 Chr.11: 14474391-14474391 on Build GRCh38
rs775957818 Chr.11: 14474565-14474565 on Build GRCh38
rs765281408 Chr.11: 14474638-14474638 on Build GRCh38
rs764730703 Chr.11: 14474448-14474448 on Build GRCh38
rs202139831 Chr.11: 14474596-14474596 on Build GRCh38
rs568654874 Chr.11: 14474713-14474713 on Build GRCh38
rs906662364 Chr.11: 14474336-14474336 on Build GRCh38
rs948309993 Chr.11: 14474703-14474703 on Build GRCh38
rs770417601 Chr.11: 14474541-14474541 on Build GRCh38
rs772536589 Chr.11: 14474564-14474564 on Build GRCh38
rs781004199 Chr.11: 14474506-14474506 on Build GRCh38
rs75195112 Chr.11: 14474459-14474459 on Build GRCh38
rs745568272 Chr.11: 14474560-14474560 on Build GRCh38
rs750625763 Chr.11: 14474640-14474640 on Build GRCh38
rs550237560 Chr.11: 14474682-14474682 on Build GRCh38
rs996822155 Chr.11: 14474398-14474398 on Build GRCh38
rs142993682 Chr.11: 14474533-14474533 on Build GRCh38
rs368271760 Chr.11: 14474477-14474477 on Build GRCh38
rs915546718 Chr.11: 14474690-14474690 on Build GRCh38
rs976662584 Chr.11: 14474563-14474563 on Build GRCh38
rs370984154 Chr.11: 14474532-14474532 on Build GRCh38
rs990495419 Chr.11: 14474579-14474579 on Build GRCh38
rs140921247 Chr.11: 14474615-14474615 on Build GRCh38
rs748659224 Chr.11: 14474421-14474421 on Build GRCh38
rs374250728 Chr.11: 14474466-14474466 on Build GRCh38
rs1034784345 Chr.11: 14474537-14474537 on Build GRCh38
rs925688735 Chr.11: 14474623-14474623 on Build GRCh38
rs542882429 Chr.11: 14474437-14474437 on Build GRCh38
rs1041514198 Chr.11: 14474373-14474373 on Build GRCh38
rs139190019 Chr.11: 14474664-14474664 on Build GRCh38
rs200255955 Chr.11: 14474641-14474641 on Build GRCh38
rs1025724921 Chr.11: 14474386-14474386 on Build GRCh38
rs779335770 Chr.11: 14474350-14474350 on Build GRCh38
rs546545070 Chr.11: 14474628-14474628 on Build GRCh38
rs755994373 Chr.11: 14474529-14474529 on Build GRCh38
rs564739984 Chr.11: 14474653-14474653 on Build GRCh38
rs1049756430 Chr.11: 14474725-14474725 on Build GRCh38
rs761305456 Chr.11: 14474444-14474444 on Build GRCh38
rs940155559 Chr.11: 14474372-14474372 on Build GRCh38
rs753907266 Chr.11: 14474452-14474452 on Build GRCh38

View Full Product Details