Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140245225-140245809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PFDN1
Entrez Gene ID
5201
Gene Name
prefoldin subunit 1
Gene Aliases
PDF, PFD1
UniGene
Hs.483564
Gene Chromosome Location
Chr.5: 140245050-140303141 on Build GRCh38
Transcripts
NM_002622.4

SNP Detail

SNP ID Chromosome Location
rs187239457 Chr.5: 140245612-140245612 on Build GRCh38
rs1044646535 Chr.5: 140245645-140245645 on Build GRCh38
rs1010684518 Chr.5: 140245702-140245702 on Build GRCh38
rs182157286 Chr.5: 140245576-140245576 on Build GRCh38
rs898729366 Chr.5: 140245436-140245436 on Build GRCh38
rs1057469137 Chr.5: 140245669-140245669 on Build GRCh38
rs1037718746 Chr.5: 140245397-140245397 on Build GRCh38
rs9042 Chr.5: 140245762-140245762 on Build GRCh38
rs762430650 Chr.5: 140245396-140245396 on Build GRCh38
rs989149790 Chr.5: 140245247-140245247 on Build GRCh38
rs983970068 Chr.5: 140245470-140245470 on Build GRCh38
rs940373398 Chr.5: 140245594-140245594 on Build GRCh38
rs1044592360 Chr.5: 140245450-140245450 on Build GRCh38
rs1019971340 Chr.5: 140245260-140245260 on Build GRCh38
rs533434993 Chr.5: 140245657-140245657 on Build GRCh38
rs776209656 Chr.5: 140245533-140245533 on Build GRCh38
rs1037363807 Chr.5: 140245613-140245613 on Build GRCh38
rs1010222167 Chr.5: 140245507-140245507 on Build GRCh38
rs954573189 Chr.5: 140245779-140245780 on Build GRCh38
rs931424114 Chr.5: 140245440-140245440 on Build GRCh38
rs933077145 Chr.5: 140245283-140245283 on Build GRCh38
rs927124870 Chr.5: 140245473-140245473 on Build GRCh38
rs956265106 Chr.5: 140245507-140245507 on Build GRCh38
rs1019598896 Chr.5: 140245313-140245313 on Build GRCh38
rs58896923 Chr.5: 140245664-140245664 on Build GRCh38
rs760860868 Chr.5: 140245683-140245683 on Build GRCh38
rs191632564 Chr.5: 140245290-140245290 on Build GRCh38
rs367752486 Chr.5: 140245274-140245274 on Build GRCh38
rs114555603 Chr.5: 140245264-140245264 on Build GRCh38
rs189269036 Chr.5: 140245575-140245575 on Build GRCh38
rs141382227 Chr.5: 140245782-140245782 on Build GRCh38
rs189160340 Chr.5: 140245646-140245646 on Build GRCh38
rs112538595 Chr.5: 140245749-140245749 on Build GRCh38
rs891878252 Chr.5: 140245692-140245692 on Build GRCh38
rs548688403 Chr.5: 140245736-140245736 on Build GRCh38
rs138413860 Chr.5: 140245443-140245443 on Build GRCh38
rs1032477471 Chr.5: 140245623-140245623 on Build GRCh38
rs11558407 Chr.5: 140245639-140245639 on Build GRCh38
rs772681404 Chr.5: 140245488-140245488 on Build GRCh38
rs577615356 Chr.5: 140245589-140245589 on Build GRCh38
rs905992306 Chr.5: 140245462-140245462 on Build GRCh38
rs987193921 Chr.5: 140245582-140245582 on Build GRCh38
rs992646866 Chr.5: 140245756-140245756 on Build GRCh38
rs927123496 Chr.5: 140245747-140245747 on Build GRCh38
rs11558409 Chr.5: 140245752-140245752 on Build GRCh38
rs940339956 Chr.5: 140245310-140245310 on Build GRCh38
rs7204 Chr.5: 140245279-140245279 on Build GRCh38
rs1002977385 Chr.5: 140245463-140245463 on Build GRCh38
rs11558408 Chr.5: 140245748-140245748 on Build GRCh38
rs972753507 Chr.5: 140245392-140245392 on Build GRCh38
rs529356593 Chr.5: 140245668-140245668 on Build GRCh38
rs1025690806 Chr.5: 140245676-140245676 on Build GRCh38
rs946005559 Chr.5: 140245777-140245777 on Build GRCh38
rs984621477 Chr.5: 140245713-140245713 on Build GRCh38
rs995710008 Chr.5: 140245448-140245448 on Build GRCh38
rs777219911 Chr.5: 140245428-140245429 on Build GRCh38
rs945701601 Chr.5: 140245559-140245559 on Build GRCh38
rs772656760 Chr.5: 140245389-140245389 on Build GRCh38
rs747599070 Chr.5: 140245574-140245574 on Build GRCh38
rs771471363 Chr.5: 140245513-140245513 on Build GRCh38
rs7203 Chr.5: 140245285-140245285 on Build GRCh38
rs1056994116 Chr.5: 140245523-140245523 on Build GRCh38
rs1019861806 Chr.5: 140245647-140245647 on Build GRCh38
rs753096992 Chr.5: 140245654-140245654 on Build GRCh38
rs914484597 Chr.5: 140245263-140245263 on Build GRCh38
rs530133616 Chr.5: 140245675-140245675 on Build GRCh38
rs895758739 Chr.5: 140245552-140245552 on Build GRCh38
rs1028837136 Chr.5: 140245764-140245764 on Build GRCh38
rs907550456 Chr.5: 140245301-140245301 on Build GRCh38
rs968654991 Chr.5: 140245550-140245550 on Build GRCh38
rs184527511 Chr.5: 140245474-140245474 on Build GRCh38
rs866404347 Chr.5: 140245757-140245757 on Build GRCh38

View Full Product Details