Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80290127-80290526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_001291812.1 NM_004464.3 NM_033143.2

SNP Detail

SNP ID Chromosome Location
rs536659513 Chr.4: 80290462-80290462 on Build GRCh38
rs1047143808 Chr.4: 80290289-80290289 on Build GRCh38
rs894479719 Chr.4: 80290489-80290489 on Build GRCh38
rs757845040 Chr.4: 80290207-80290207 on Build GRCh38
rs569051087 Chr.4: 80290364-80290364 on Build GRCh38
rs893321294 Chr.4: 80290158-80290158 on Build GRCh38
rs551116032 Chr.4: 80290482-80290482 on Build GRCh38
rs1010428931 Chr.4: 80290222-80290222 on Build GRCh38
rs200665298 Chr.4: 80290337-80290338 on Build GRCh38
rs1003539862 Chr.4: 80290460-80290460 on Build GRCh38
rs906215290 Chr.4: 80290280-80290280 on Build GRCh38
rs923247903 Chr.4: 80290338-80290338 on Build GRCh38
rs907192100 Chr.4: 80290245-80290245 on Build GRCh38
rs1046299459 Chr.4: 80290261-80290261 on Build GRCh38
rs933241606 Chr.4: 80290338-80290339 on Build GRCh38
rs926078520 Chr.4: 80290193-80290193 on Build GRCh38
rs929676628 Chr.4: 80290283-80290283 on Build GRCh38
rs1002907424 Chr.4: 80290274-80290274 on Build GRCh38
rs1005221117 Chr.4: 80290301-80290301 on Build GRCh38
rs1020515656 Chr.4: 80290244-80290244 on Build GRCh38
rs927918421 Chr.4: 80290421-80290421 on Build GRCh38
rs982329682 Chr.4: 80290419-80290419 on Build GRCh38
rs142199666 Chr.4: 80290170-80290170 on Build GRCh38
rs916378247 Chr.4: 80290215-80290215 on Build GRCh38
rs1018438038 Chr.4: 80290309-80290309 on Build GRCh38
rs763329168 Chr.4: 80290351-80290351 on Build GRCh38
rs758515262 Chr.4: 80290399-80290399 on Build GRCh38
rs929850273 Chr.4: 80290431-80290431 on Build GRCh38
rs961865449 Chr.4: 80290495-80290495 on Build GRCh38
rs957443740 Chr.4: 80290385-80290385 on Build GRCh38
rs971973739 Chr.4: 80290331-80290331 on Build GRCh38
rs1006292956 Chr.4: 80290491-80290491 on Build GRCh38
rs554863214 Chr.4: 80290412-80290412 on Build GRCh38
rs565628405 Chr.4: 80290369-80290369 on Build GRCh38
rs900001661 Chr.4: 80290314-80290314 on Build GRCh38
rs936407539 Chr.4: 80290362-80290362 on Build GRCh38
rs985776901 Chr.4: 80290348-80290348 on Build GRCh38
rs552941218 Chr.4: 80290493-80290493 on Build GRCh38
rs998419932 Chr.4: 80290328-80290328 on Build GRCh38
rs888206288 Chr.4: 80290295-80290295 on Build GRCh38
rs977931525 Chr.4: 80290152-80290152 on Build GRCh38
rs936041275 Chr.4: 80290196-80290196 on Build GRCh38
rs4690150 Chr.4: 80290416-80290416 on Build GRCh38
rs968007892 Chr.4: 80290338-80290338 on Build GRCh38
rs1029893957 Chr.4: 80290332-80290332 on Build GRCh38
rs958779002 Chr.4: 80290286-80290286 on Build GRCh38
rs947987189 Chr.4: 80290217-80290217 on Build GRCh38

View Full Product Details