Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31935844-31936338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NR_104191.1

Gene Symbol
C2-AS1
Entrez Gene ID
102060414
Gene Name
C2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571112
Gene Chromosome Location
Chr.6: 31934474-31941724 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NR_104191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6106551 c.987A>G p.K329K Chr.6: 31936060-31936060 on Build GRCh38
5870227 c.877G>A p.V293I Chr.6: 31935950-31935950 on Build GRCh38
4512731 c.913C>T p.L305F Chr.6: 31935986-31935986 on Build GRCh38
3624867 c.978C>T p.A326A Chr.6: 31936051-31936051 on Build GRCh38
5870228 c.931G>A p.D311N Chr.6: 31936004-31936004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750860477 Chr.6: 31936049-31936049 on Build GRCh38
rs373410228 Chr.6: 31936067-31936067 on Build GRCh38
rs759240195 Chr.6: 31935883-31935883 on Build GRCh38
rs149815968 Chr.6: 31936282-31936282 on Build GRCh38
rs748955584 Chr.6: 31936082-31936082 on Build GRCh38
rs767907586 Chr.6: 31936033-31936033 on Build GRCh38
rs766665783 Chr.6: 31936053-31936053 on Build GRCh38
rs35299701 Chr.6: 31936016-31936017 on Build GRCh38
rs532091149 Chr.6: 31936009-31936009 on Build GRCh38
rs549144980 Chr.6: 31935950-31935950 on Build GRCh38
rs55855135 Chr.6: 31936295-31936295 on Build GRCh38
rs560405683 Chr.6: 31935898-31935898 on Build GRCh38
rs750689314 Chr.6: 31935941-31935941 on Build GRCh38
rs367795640 Chr.6: 31935949-31935949 on Build GRCh38
rs754038034 Chr.6: 31936054-31936054 on Build GRCh38
rs755202596 Chr.6: 31936057-31936057 on Build GRCh38
rs969072854 Chr.6: 31936168-31936168 on Build GRCh38
rs41267098 Chr.6: 31936182-31936182 on Build GRCh38
rs541199729 Chr.6: 31935992-31935992 on Build GRCh38
rs768119622 Chr.6: 31936085-31936085 on Build GRCh38
rs550498736 Chr.6: 31936014-31936014 on Build GRCh38
rs751970483 Chr.6: 31935873-31935873 on Build GRCh38
rs768234923 Chr.6: 31936083-31936083 on Build GRCh38
rs977363113 Chr.6: 31936178-31936178 on Build GRCh38
rs764728828 Chr.6: 31935892-31935892 on Build GRCh38
rs767887161 Chr.6: 31935925-31935925 on Build GRCh38
rs757480451 Chr.6: 31936243-31936243 on Build GRCh38
rs151097610 Chr.6: 31935916-31935916 on Build GRCh38
rs369131611 Chr.6: 31935897-31935897 on Build GRCh38
rs533764524 Chr.6: 31936202-31936202 on Build GRCh38
rs780974763 Chr.6: 31936066-31936066 on Build GRCh38
rs897977251 Chr.6: 31936061-31936061 on Build GRCh38
rs756413100 Chr.6: 31936051-31936051 on Build GRCh38
rs754004104 Chr.6: 31936219-31936219 on Build GRCh38
rs889422786 Chr.6: 31936293-31936293 on Build GRCh38
rs9332739 Chr.6: 31936027-31936027 on Build GRCh38
rs773920592 Chr.6: 31936084-31936084 on Build GRCh38
rs144201432 Chr.6: 31936002-31936002 on Build GRCh38
rs947331094 Chr.6: 31936210-31936210 on Build GRCh38
rs770933665 Chr.6: 31935993-31935993 on Build GRCh38
rs564996447 Chr.6: 31936047-31936047 on Build GRCh38
rs201677911 Chr.6: 31936065-31936065 on Build GRCh38
rs755077763 Chr.6: 31935959-31935959 on Build GRCh38
rs976558486 Chr.6: 31936209-31936209 on Build GRCh38
rs146165915 Chr.6: 31936003-31936003 on Build GRCh38
rs1007903370 Chr.6: 31936312-31936312 on Build GRCh38
rs775310958 Chr.6: 31936078-31936078 on Build GRCh38
rs9332740 Chr.6: 31936154-31936154 on Build GRCh38
rs369273342 Chr.6: 31935997-31935997 on Build GRCh38
rs924459907 Chr.6: 31935905-31935905 on Build GRCh38
rs1040463991 Chr.6: 31935946-31935946 on Build GRCh38
rs545067469 Chr.6: 31935869-31935869 on Build GRCh38
rs779097525 Chr.6: 31935969-31935969 on Build GRCh38
rs779146789 Chr.6: 31936059-31936059 on Build GRCh38
rs916590744 Chr.6: 31936101-31936101 on Build GRCh38
rs377748058 Chr.6: 31936077-31936077 on Build GRCh38
rs1052918485 Chr.6: 31936195-31936195 on Build GRCh38
rs374809864 Chr.6: 31935917-31935917 on Build GRCh38
rs746963737 Chr.6: 31935985-31935985 on Build GRCh38
rs949430602 Chr.6: 31936143-31936143 on Build GRCh38
rs756428736 Chr.6: 31935943-31935943 on Build GRCh38
rs774981560 Chr.6: 31936004-31936004 on Build GRCh38
rs113905418 Chr.6: 31935921-31935921 on Build GRCh38

View Full Product Details