Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95580454-95581050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DZIP1
Entrez Gene ID
22873
Gene Name
DAZ interacting zinc finger protein 1
Gene Aliases
DZIP, DZIPt1
UniGene
Hs.656580
Gene Chromosome Location
Chr.13: 95578202-95644706 on Build GRCh38
Transcripts
NM_014934.4 NM_198968.3

SNP Detail

SNP ID Chromosome Location
rs372480124 Chr.13: 95580894-95580894 on Build GRCh38
rs761032865 Chr.13: 95580519-95580520 on Build GRCh38
rs140220534 Chr.13: 95580869-95580869 on Build GRCh38
rs142295032 Chr.13: 95580547-95580547 on Build GRCh38
rs890666442 Chr.13: 95581014-95581014 on Build GRCh38
rs571644025 Chr.13: 95580622-95580622 on Build GRCh38
rs1026596919 Chr.13: 95580882-95580882 on Build GRCh38
rs903826229 Chr.13: 95580725-95580725 on Build GRCh38
rs929662673 Chr.13: 95580613-95580613 on Build GRCh38
rs939214330 Chr.13: 95581011-95581011 on Build GRCh38
rs980967233 Chr.13: 95580982-95580982 on Build GRCh38
rs78845384 Chr.13: 95580628-95580628 on Build GRCh38
rs778683453 Chr.13: 95580719-95580719 on Build GRCh38
rs941120534 Chr.13: 95580624-95580624 on Build GRCh38
rs968523815 Chr.13: 95580813-95580813 on Build GRCh38
rs909546846 Chr.13: 95580945-95580945 on Build GRCh38
rs993734140 Chr.13: 95580652-95580652 on Build GRCh38
rs914190066 Chr.13: 95580749-95580749 on Build GRCh38
rs924495181 Chr.13: 95580858-95580858 on Build GRCh38
rs757001673 Chr.13: 95580649-95580649 on Build GRCh38
rs954721782 Chr.13: 95580728-95580728 on Build GRCh38
rs989961138 Chr.13: 95580777-95580777 on Build GRCh38
rs114658735 Chr.13: 95580495-95580495 on Build GRCh38
rs557180224 Chr.13: 95580673-95580673 on Build GRCh38
rs966204200 Chr.13: 95580542-95580542 on Build GRCh38
rs948527324 Chr.13: 95580926-95580926 on Build GRCh38
rs375424578 Chr.13: 95580929-95580929 on Build GRCh38
rs752312357 Chr.13: 95580540-95580540 on Build GRCh38
rs894191506 Chr.13: 95580621-95580621 on Build GRCh38
rs1022737972 Chr.13: 95580708-95580708 on Build GRCh38
rs573230901 Chr.13: 95580931-95580931 on Build GRCh38
rs1018106548 Chr.13: 95580762-95580762 on Build GRCh38
rs899357272 Chr.13: 95580957-95580957 on Build GRCh38
rs150820681 Chr.13: 95580799-95580799 on Build GRCh38
rs74109004 Chr.13: 95580509-95580509 on Build GRCh38
rs938336610 Chr.13: 95580863-95580863 on Build GRCh38
rs1009559175 Chr.13: 95581027-95581027 on Build GRCh38
rs966650394 Chr.13: 95580810-95580810 on Build GRCh38
rs145685507 Chr.13: 95581009-95581009 on Build GRCh38
rs1009548992 Chr.13: 95580676-95580676 on Build GRCh38
rs762412273 Chr.13: 95580853-95580853 on Build GRCh38
rs910725686 Chr.13: 95580522-95580522 on Build GRCh38
rs181726958 Chr.13: 95580597-95580597 on Build GRCh38
rs1022366770 Chr.13: 95580483-95580483 on Build GRCh38
rs977355633 Chr.13: 95580835-95580835 on Build GRCh38
rs1003326469 Chr.13: 95580653-95580653 on Build GRCh38
rs573951808 Chr.13: 95580767-95580767 on Build GRCh38
rs1008066206 Chr.13: 95580758-95580758 on Build GRCh38
rs80307494 Chr.13: 95580620-95580620 on Build GRCh38
rs553237454 Chr.13: 95580904-95580904 on Build GRCh38
rs1056340720 Chr.13: 95580663-95580663 on Build GRCh38
rs940996281 Chr.13: 95580959-95580959 on Build GRCh38
rs566787174 Chr.13: 95580572-95580572 on Build GRCh38

View Full Product Details