Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145095641-145096166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
UniGene
Hs.480876
Gene Chromosome Location
Chr.4: 144967117-145098541 on Build GRCh38
Transcripts
NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_001318367.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865820 c.112C>T p.P38S Chr.4: 145095988-145095988 on Build GRCh38
3600627 c.76G>T p.G26W Chr.4: 145096024-145096024 on Build GRCh38
4661480 c.68G>A p.R23Q Chr.4: 145096032-145096032 on Build GRCh38
1244923 c.67C>T p.R23W Chr.4: 145096033-145096033 on Build GRCh38
3126616 c.1A>G p.M1V Chr.4: 145096099-145096099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139743809 Chr.4: 145095677-145095677 on Build GRCh38
rs563299168 Chr.4: 145095805-145095805 on Build GRCh38
rs1045457749 Chr.4: 145095712-145095712 on Build GRCh38
rs745854313 Chr.4: 145096046-145096046 on Build GRCh38
rs779611589 Chr.4: 145096039-145096039 on Build GRCh38
rs561172544 Chr.4: 145095740-145095740 on Build GRCh38
rs765638815 Chr.4: 145096118-145096118 on Build GRCh38
rs941290258 Chr.4: 145095698-145095698 on Build GRCh38
rs181961937 Chr.4: 145095725-145095725 on Build GRCh38
rs949319913 Chr.4: 145095758-145095758 on Build GRCh38
rs543558955 Chr.4: 145095792-145095792 on Build GRCh38
rs113294154 Chr.4: 145095671-145095671 on Build GRCh38
rs908489669 Chr.4: 145095745-145095745 on Build GRCh38
rs890438911 Chr.4: 145096016-145096016 on Build GRCh38
rs572178457 Chr.4: 145095686-145095686 on Build GRCh38
rs982454500 Chr.4: 145095742-145095742 on Build GRCh38
rs900710493 Chr.4: 145096114-145096114 on Build GRCh38
rs36208502 Chr.4: 145095667-145095671 on Build GRCh38
rs751664837 Chr.4: 145096143-145096143 on Build GRCh38
rs369273194 Chr.4: 145095936-145095936 on Build GRCh38
rs1033386470 Chr.4: 145095886-145095886 on Build GRCh38
rs1349990 Chr.4: 145095845-145095845 on Build GRCh38
rs769923189 Chr.4: 145095949-145095949 on Build GRCh38
rs199570289 Chr.4: 145096003-145096003 on Build GRCh38
rs758807608 Chr.4: 145095976-145095976 on Build GRCh38
rs761442053 Chr.4: 145096109-145096109 on Build GRCh38
rs765581744 Chr.4: 145095951-145095951 on Build GRCh38
rs957495206 Chr.4: 145095941-145095941 on Build GRCh38
rs1007790592 Chr.4: 145095864-145095864 on Build GRCh38
rs980688735 Chr.4: 145096138-145096138 on Build GRCh38
rs963719283 Chr.4: 145095948-145095948 on Build GRCh38
rs939365018 Chr.4: 145095748-145095748 on Build GRCh38
rs981964758 Chr.4: 145095776-145095776 on Build GRCh38
rs751544256 Chr.4: 145095998-145095998 on Build GRCh38
rs766740632 Chr.4: 145096130-145096130 on Build GRCh38
rs532382825 Chr.4: 145095810-145095810 on Build GRCh38
rs1009650539 Chr.4: 145095788-145095788 on Build GRCh38
rs937818533 Chr.4: 145095822-145095822 on Build GRCh38
rs776331064 Chr.4: 145096079-145096079 on Build GRCh38
rs941237843 Chr.4: 145096132-145096132 on Build GRCh38
rs760042262 Chr.4: 145096078-145096078 on Build GRCh38
rs755241784 Chr.4: 145096002-145096002 on Build GRCh38
rs975577061 Chr.4: 145095685-145095686 on Build GRCh38
rs893883616 Chr.4: 145095880-145095880 on Build GRCh38
rs905213890 Chr.4: 145095736-145095736 on Build GRCh38
rs909742980 Chr.4: 145096075-145096075 on Build GRCh38
rs781501474 Chr.4: 145096008-145096008 on Build GRCh38
rs982606255 Chr.4: 145096073-145096073 on Build GRCh38
rs771862321 Chr.4: 145096064-145096064 on Build GRCh38
rs746378181 Chr.4: 145096022-145096022 on Build GRCh38
rs762097496 Chr.4: 145095943-145095943 on Build GRCh38
rs775083414 Chr.4: 145096076-145096076 on Build GRCh38
rs770910770 Chr.4: 145096032-145096032 on Build GRCh38
rs750994895 Chr.4: 145095957-145095957 on Build GRCh38
rs369942751 Chr.4: 145095767-145095767 on Build GRCh38
rs866738620 Chr.4: 145096035-145096035 on Build GRCh38
rs1026731706 Chr.4: 145095685-145095685 on Build GRCh38
rs780215010 Chr.4: 145095981-145095981 on Build GRCh38
rs750713970 Chr.4: 145096124-145096124 on Build GRCh38
rs200114650 Chr.4: 145096033-145096033 on Build GRCh38
rs773227799 Chr.4: 145095966-145095969 on Build GRCh38
rs1056523121 Chr.4: 145095764-145095764 on Build GRCh38
rs976833580 Chr.4: 145096019-145096019 on Build GRCh38
rs528807783 Chr.4: 145096126-145096126 on Build GRCh38

View Full Product Details