Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7803703-7804012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
UniGene
Hs.162200
Gene Chromosome Location
Chr.1: 7784285-7845181 on Build GRCh38
Transcripts
NM_001289861.1 NM_001289862.1 NM_001289863.1 NM_001289864.1 NM_016831.2 XM_005263524.3 XM_011542385.2 XM_011542390.2 XM_017002723.1 XM_017002724.1 XM_017002725.1 XM_017002726.1 XM_017002727.1 XM_017002728.1 XM_017002729.1 XM_017002730.1 XM_017002731.1 XM_017002732.1 XM_017002733.1 XM_017002734.1 XM_017002735.1 XM_017002736.1 XM_017002737.1 XM_017002738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390401 c.1091G>T p.S364I Chr.1: 7803806-7803806 on Build GRCh38
2242675 c.1034delA p.N346fs*15 Chr.1: 7803749-7803749 on Build GRCh38
1686944 c.1016C>T p.P339L Chr.1: 7803731-7803731 on Build GRCh38
1176836 c.1103C>A p.S368Y Chr.1: 7803818-7803818 on Build GRCh38
7220378 c.1051A>T p.I351L Chr.1: 7803766-7803766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745884057 Chr.1: 7803758-7803758 on Build GRCh38
rs778909457 Chr.1: 7803749-7803749 on Build GRCh38
rs528247062 Chr.1: 7803892-7803892 on Build GRCh38
rs568252759 Chr.1: 7803919-7803919 on Build GRCh38
rs775921604 Chr.1: 7803776-7803776 on Build GRCh38
rs766000822 Chr.1: 7803810-7803810 on Build GRCh38
rs936587567 Chr.1: 7803767-7803767 on Build GRCh38
rs759521971 Chr.1: 7803887-7803887 on Build GRCh38
rs781226803 Chr.1: 7803838-7803838 on Build GRCh38
rs772950694 Chr.1: 7803808-7803808 on Build GRCh38
rs759532418 Chr.1: 7803844-7803844 on Build GRCh38
rs1025330907 Chr.1: 7803763-7803763 on Build GRCh38
rs143443665 Chr.1: 7803766-7803766 on Build GRCh38
rs756178157 Chr.1: 7803848-7803848 on Build GRCh38
rs767355809 Chr.1: 7803869-7803869 on Build GRCh38
rs890938862 Chr.1: 7803942-7803942 on Build GRCh38
rs746104252 Chr.1: 7803842-7803842 on Build GRCh38
rs765214745 Chr.1: 7803890-7803890 on Build GRCh38
rs868705028 Chr.1: 7803989-7803989 on Build GRCh38
rs774739639 Chr.1: 7803785-7803785 on Build GRCh38
rs754761909 Chr.1: 7803737-7803737 on Build GRCh38
rs191016322 Chr.1: 7803809-7803809 on Build GRCh38
rs758231609 Chr.1: 7803834-7803834 on Build GRCh38
rs780101079 Chr.1: 7803725-7803725 on Build GRCh38
rs375965833 Chr.1: 7803756-7803756 on Build GRCh38
rs758057475 Chr.1: 7803902-7803902 on Build GRCh38
rs546960560 Chr.1: 7803904-7803904 on Build GRCh38
rs374017079 Chr.1: 7803780-7803780 on Build GRCh38
rs757458058 Chr.1: 7803754-7803754 on Build GRCh38
rs200722810 Chr.1: 7803812-7803812 on Build GRCh38
rs761506646 Chr.1: 7803797-7803797 on Build GRCh38
rs767029988 Chr.1: 7803800-7803800 on Build GRCh38
rs151097620 Chr.1: 7803841-7803841 on Build GRCh38
rs774863870 Chr.1: 7803862-7803862 on Build GRCh38
rs774805653 Chr.1: 7803753-7803753 on Build GRCh38
rs755032501 Chr.1: 7803827-7803827 on Build GRCh38
rs868049631 Chr.1: 7803730-7803730 on Build GRCh38
rs373562934 Chr.1: 7803880-7803880 on Build GRCh38
rs75929074 Chr.1: 7803884-7803884 on Build GRCh38
rs780177710 Chr.1: 7803849-7803849 on Build GRCh38
rs199945897 Chr.1: 7803832-7803832 on Build GRCh38
rs960176797 Chr.1: 7803933-7803933 on Build GRCh38
rs993721124 Chr.1: 7803917-7803917 on Build GRCh38
rs753830988 Chr.1: 7803732-7803732 on Build GRCh38
rs749855218 Chr.1: 7803778-7803778 on Build GRCh38
rs752481233 Chr.1: 7803833-7803833 on Build GRCh38
rs377629053 Chr.1: 7803865-7803865 on Build GRCh38
rs12566042 Chr.1: 7803970-7803970 on Build GRCh38
rs777169668 Chr.1: 7803762-7803764 on Build GRCh38
rs752708456 Chr.1: 7803901-7803901 on Build GRCh38
rs556706543 Chr.1: 7803969-7803969 on Build GRCh38
rs749767313 Chr.1: 7803856-7803856 on Build GRCh38
rs915967086 Chr.1: 7803949-7803949 on Build GRCh38
rs182583917 Chr.1: 7803867-7803867 on Build GRCh38
rs879077399 Chr.1: 7803951-7803951 on Build GRCh38
rs765982350 Chr.1: 7803882-7803882 on Build GRCh38
rs992983900 Chr.1: 7803945-7803945 on Build GRCh38
rs760290260 Chr.1: 7803859-7803859 on Build GRCh38
rs535464221 Chr.1: 7803922-7803922 on Build GRCh38

View Full Product Details