Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46349464-46350063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

SNP Detail

SNP ID Chromosome Location
rs192730152 Chr.20: 46349600-46349600 on Build GRCh38
rs964488497 Chr.20: 46349808-46349808 on Build GRCh38
rs932343917 Chr.20: 46349992-46349992 on Build GRCh38
rs551907050 Chr.20: 46349803-46349803 on Build GRCh38
rs186547699 Chr.20: 46349509-46349509 on Build GRCh38
rs118083022 Chr.20: 46349622-46349622 on Build GRCh38
rs1033693200 Chr.20: 46349806-46349806 on Build GRCh38
rs748790791 Chr.20: 46349614-46349614 on Build GRCh38
rs984062983 Chr.20: 46349892-46349892 on Build GRCh38
rs1042671774 Chr.20: 46349577-46349577 on Build GRCh38
rs80231350 Chr.20: 46349684-46349684 on Build GRCh38
rs1055347642 Chr.20: 46349904-46349904 on Build GRCh38
rs916770109 Chr.20: 46349918-46349918 on Build GRCh38
rs559627556 Chr.20: 46349666-46349666 on Build GRCh38
rs113908366 Chr.20: 46349799-46349799 on Build GRCh38
rs914930024 Chr.20: 46349527-46349527 on Build GRCh38
rs1038482281 Chr.20: 46349952-46349952 on Build GRCh38
rs531228208 Chr.20: 46349921-46349921 on Build GRCh38
rs570038575 Chr.20: 46349828-46349828 on Build GRCh38
rs939593151 Chr.20: 46349901-46349901 on Build GRCh38
rs1017392982 Chr.20: 46349737-46349737 on Build GRCh38
rs994563525 Chr.20: 46349661-46349661 on Build GRCh38
rs567842932 Chr.20: 46349940-46349940 on Build GRCh38
rs954619951 Chr.20: 46349687-46349687 on Build GRCh38
rs914959248 Chr.20: 46349826-46349826 on Build GRCh38
rs537068397 Chr.20: 46349885-46349885 on Build GRCh38
rs908321011 Chr.20: 46349487-46349487 on Build GRCh38
rs893759835 Chr.20: 46350027-46350027 on Build GRCh38
rs978160810 Chr.20: 46349879-46349879 on Build GRCh38
rs183068226 Chr.20: 46349713-46349713 on Build GRCh38
rs989256525 Chr.20: 46349821-46349821 on Build GRCh38
rs945006222 Chr.20: 46349559-46349559 on Build GRCh38
rs879032512 Chr.20: 46349567-46349567 on Build GRCh38
rs1041108469 Chr.20: 46349529-46349529 on Build GRCh38
rs1005637555 Chr.20: 46349633-46349633 on Build GRCh38
rs147273359 Chr.20: 46349652-46349652 on Build GRCh38
rs73908692 Chr.20: 46349596-46349596 on Build GRCh38
rs1051052820 Chr.20: 46350022-46350022 on Build GRCh38
rs774873467 Chr.20: 46349608-46349608 on Build GRCh38
rs891086946 Chr.20: 46350038-46350038 on Build GRCh38
rs562454763 Chr.20: 46349736-46349736 on Build GRCh38
rs973383441 Chr.20: 46349861-46349861 on Build GRCh38
rs989594038 Chr.20: 46349483-46349483 on Build GRCh38
rs897228963 Chr.20: 46349957-46349957 on Build GRCh38
rs920874924 Chr.20: 46349874-46349874 on Build GRCh38
rs1038840216 Chr.20: 46349634-46349634 on Build GRCh38
rs943798495 Chr.20: 46349513-46349513 on Build GRCh38
rs930027286 Chr.20: 46350003-46350003 on Build GRCh38
rs920949519 Chr.20: 46349975-46349975 on Build GRCh38

View Full Product Details