Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72238595-72239194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1 NM_005169.3

Gene Symbol
PHOX2A
Entrez Gene ID
401
Gene Name
paired like homeobox 2a
Gene Aliases
ARIX, CFEOM2, FEOM2, NCAM2, PMX2A
UniGene
Hs.731115
Gene Chromosome Location
Chr.11: 72239077-72244176 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1 NM_005169.3

SNP Detail

SNP ID Chromosome Location
rs994973273 Chr.11: 72238992-72238992 on Build GRCh38
rs878898108 Chr.11: 72238775-72238775 on Build GRCh38
rs950301862 Chr.11: 72238963-72238963 on Build GRCh38
rs976975636 Chr.11: 72238844-72238844 on Build GRCh38
rs965345504 Chr.11: 72238813-72238813 on Build GRCh38
rs112117090 Chr.11: 72238823-72238823 on Build GRCh38
rs554092934 Chr.11: 72238792-72238792 on Build GRCh38
rs545989358 Chr.11: 72238904-72238904 on Build GRCh38
rs1009996960 Chr.11: 72238820-72238820 on Build GRCh38
rs371837004 Chr.11: 72238789-72238789 on Build GRCh38
rs1026683166 Chr.11: 72239019-72239019 on Build GRCh38
rs896110177 Chr.11: 72238750-72238750 on Build GRCh38
rs1020630899 Chr.11: 72239093-72239093 on Build GRCh38
rs770592288 Chr.11: 72239108-72239108 on Build GRCh38
rs913033277 Chr.11: 72238869-72238869 on Build GRCh38
rs973609888 Chr.11: 72239136-72239136 on Build GRCh38
rs1037954811 Chr.11: 72238769-72238774 on Build GRCh38
rs11548492 Chr.11: 72238902-72238902 on Build GRCh38
rs976659512 Chr.11: 72239069-72239069 on Build GRCh38
rs907876462 Chr.11: 72238705-72238705 on Build GRCh38
rs1020331670 Chr.11: 72238865-72238865 on Build GRCh38
rs538319693 Chr.11: 72238893-72238893 on Build GRCh38
rs769905725 Chr.11: 72239001-72239001 on Build GRCh38
rs911126354 Chr.11: 72238811-72238811 on Build GRCh38
rs9886 Chr.11: 72238621-72238621 on Build GRCh38
rs992637127 Chr.11: 72239002-72239002 on Build GRCh38
rs975021092 Chr.11: 72238769-72238769 on Build GRCh38
rs926009573 Chr.11: 72239166-72239166 on Build GRCh38
rs70940823 Chr.11: 72238680-72238680 on Build GRCh38
rs866844982 Chr.11: 72238993-72238993 on Build GRCh38
rs71726140 Chr.11: 72238800-72238803 on Build GRCh38
rs76160915 Chr.11: 72238656-72238656 on Build GRCh38
rs942068785 Chr.11: 72239086-72239086 on Build GRCh38
rs543154415 Chr.11: 72238979-72238979 on Build GRCh38
rs907862987 Chr.11: 72239061-72239061 on Build GRCh38
rs888233919 Chr.11: 72239038-72239038 on Build GRCh38
rs70940830 Chr.11: 72239092-72239095 on Build GRCh38
rs558748277 Chr.11: 72238636-72238636 on Build GRCh38
rs368490749 Chr.11: 72239032-72239032 on Build GRCh38
rs967197977 Chr.11: 72239133-72239133 on Build GRCh38
rs904487213 Chr.11: 72238725-72238725 on Build GRCh38
rs1015122483 Chr.11: 72238765-72238765 on Build GRCh38
rs3210036 Chr.11: 72238908-72238908 on Build GRCh38
rs1055824132 Chr.11: 72238943-72238943 on Build GRCh38
rs62625974 Chr.11: 72238662-72238662 on Build GRCh38
rs958508484 Chr.11: 72238994-72238994 on Build GRCh38
rs963925531 Chr.11: 72238761-72238761 on Build GRCh38
rs937324079 Chr.11: 72238925-72238925 on Build GRCh38
rs978632369 Chr.11: 72239150-72239150 on Build GRCh38
rs913286924 Chr.11: 72238672-72238672 on Build GRCh38
rs115908947 Chr.11: 72239041-72239041 on Build GRCh38
rs985125456 Chr.11: 72238809-72238809 on Build GRCh38
rs70940824 Chr.11: 72238968-72238968 on Build GRCh38
rs896044214 Chr.11: 72238961-72238961 on Build GRCh38
rs932250524 Chr.11: 72239156-72239158 on Build GRCh38
rs866293610 Chr.11: 72238668-72238668 on Build GRCh38
rs747186762 Chr.11: 72238683-72238683 on Build GRCh38
rs759057949 Chr.11: 72238664-72238665 on Build GRCh38
rs904228907 Chr.11: 72238917-72238917 on Build GRCh38
rs952346446 Chr.11: 72238797-72238797 on Build GRCh38
rs926063396 Chr.11: 72238922-72238922 on Build GRCh38
rs781538687 Chr.11: 72238955-72238956 on Build GRCh38
rs780890775 Chr.11: 72238737-72238737 on Build GRCh38
rs144478961 Chr.11: 72239010-72239010 on Build GRCh38
rs1017966966 Chr.11: 72239045-72239045 on Build GRCh38
rs965427802 Chr.11: 72239056-72239056 on Build GRCh38
rs3210037 Chr.11: 72238912-72238912 on Build GRCh38
rs922166885 Chr.11: 72239147-72239147 on Build GRCh38
rs777870398 Chr.11: 72238870-72238870 on Build GRCh38
rs1001545408 Chr.11: 72238735-72238735 on Build GRCh38
rs561489109 Chr.11: 72239156-72239156 on Build GRCh38
rs756180939 Chr.11: 72238825-72238825 on Build GRCh38
rs897954203 Chr.11: 72238983-72238983 on Build GRCh38
rs556476284 Chr.11: 72238696-72238696 on Build GRCh38
rs962536142 Chr.11: 72238768-72238768 on Build GRCh38
rs574736657 Chr.11: 72238777-72238777 on Build GRCh38

View Full Product Details