Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18142697-18143257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KAT14
Entrez Gene ID
57325
Gene Name
lysine acetyltransferase 14
Gene Aliases
ATAC2, CRP2BP, CSRP2BP, PRO1194, dJ717M23.1
UniGene
Hs.488051
Gene Chromosome Location
Chr.20: 18142224-18188387 on Build GRCh38
Transcripts
NM_020536.4 NM_001164811.1

Gene Symbol
PET117
Entrez Gene ID
100303755
Gene Name
PET117 homolog
Gene Aliases
CSRP2BP
UniGene
Hs.488051
Gene Chromosome Location
Chr.20: 18137855-18143169 on Build GRCh38
Transcripts
NM_020536.4 NM_001164811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7295273 c.235A>G p.K79E Chr.20: 18142895-18142895 on Build GRCh38
1249340 c.37C>T p.R13W Chr.20: 18142697-18142697 on Build GRCh38
5642243 c.136G>T p.A46S Chr.20: 18142796-18142796 on Build GRCh38
1641293 c.118G>A p.V40I Chr.20: 18142778-18142778 on Build GRCh38
1024846 c.66G>A p.S22S Chr.20: 18142726-18142726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890438601 Chr.20: 18143144-18143144 on Build GRCh38
rs756739367 Chr.20: 18142840-18142840 on Build GRCh38
rs1009968143 Chr.20: 18143226-18143226 on Build GRCh38
rs778893335 Chr.20: 18142969-18142969 on Build GRCh38
rs751152779 Chr.20: 18142837-18142837 on Build GRCh38
rs371840270 Chr.20: 18142962-18142962 on Build GRCh38
rs112718518 Chr.20: 18142931-18142931 on Build GRCh38
rs754315371 Chr.20: 18142739-18142739 on Build GRCh38
rs773537680 Chr.20: 18142945-18142945 on Build GRCh38
rs201768810 Chr.20: 18142787-18142787 on Build GRCh38
rs1020299818 Chr.20: 18143182-18143182 on Build GRCh38
rs763740306 Chr.20: 18142831-18142831 on Build GRCh38
rs972721082 Chr.20: 18143073-18143073 on Build GRCh38
rs370934662 Chr.20: 18142716-18142716 on Build GRCh38
rs773504576 Chr.20: 18142811-18142811 on Build GRCh38
rs200687917 Chr.20: 18142856-18142856 on Build GRCh38
rs150368810 Chr.20: 18142916-18142916 on Build GRCh38
rs1054619092 Chr.20: 18143181-18143188 on Build GRCh38
rs771028770 Chr.20: 18142905-18142905 on Build GRCh38
rs374168048 Chr.20: 18142768-18142768 on Build GRCh38
rs776821624 Chr.20: 18142907-18142907 on Build GRCh38
rs756717589 Chr.20: 18142780-18142780 on Build GRCh38
rs139764811 Chr.20: 18142959-18142959 on Build GRCh38
rs772493399 Chr.20: 18142799-18142799 on Build GRCh38
rs886587092 Chr.20: 18142771-18142771 on Build GRCh38
rs755322355 Chr.20: 18142753-18142753 on Build GRCh38
rs942188971 Chr.20: 18143071-18143071 on Build GRCh38
rs760131098 Chr.20: 18142817-18142817 on Build GRCh38
rs775908942 Chr.20: 18142917-18142917 on Build GRCh38
rs1050465391 Chr.20: 18143134-18143134 on Build GRCh38
rs777037743 Chr.20: 18142816-18142816 on Build GRCh38
rs1001413040 Chr.20: 18142994-18142994 on Build GRCh38
rs984575289 Chr.20: 18142750-18142750 on Build GRCh38
rs780709626 Chr.20: 18142855-18142855 on Build GRCh38
rs937930498 Chr.20: 18142954-18142954 on Build GRCh38
rs1030296583 Chr.20: 18142876-18142876 on Build GRCh38
rs375777965 Chr.20: 18142717-18142717 on Build GRCh38
rs41276414 Chr.20: 18142933-18142933 on Build GRCh38
rs765598660 Chr.20: 18142767-18142767 on Build GRCh38
rs369310182 Chr.20: 18142958-18142958 on Build GRCh38
rs572036161 Chr.20: 18142819-18142819 on Build GRCh38
rs374911798 Chr.20: 18142934-18142934 on Build GRCh38
rs766436587 Chr.20: 18142813-18142813 on Build GRCh38
rs900719164 Chr.20: 18143096-18143096 on Build GRCh38
rs759522652 Chr.20: 18142911-18142911 on Build GRCh38
rs368748280 Chr.20: 18142725-18142725 on Build GRCh38
rs969209023 Chr.20: 18143190-18143190 on Build GRCh38
rs541176370 Chr.20: 18143050-18143050 on Build GRCh38
rs535284998 Chr.20: 18143030-18143030 on Build GRCh38
rs7351173 Chr.20: 18143138-18143138 on Build GRCh38
rs75678841 Chr.20: 18143203-18143203 on Build GRCh38
rs75839727 Chr.20: 18142901-18142901 on Build GRCh38
rs146620856 Chr.20: 18143174-18143174 on Build GRCh38
rs749066519 Chr.20: 18142794-18142794 on Build GRCh38
rs767050498 Chr.20: 18142936-18142936 on Build GRCh38
rs537659680 Chr.20: 18142946-18142946 on Build GRCh38
rs11906677 Chr.20: 18143000-18143000 on Build GRCh38
rs114882072 Chr.20: 18143064-18143064 on Build GRCh38
rs745766214 Chr.20: 18142786-18142786 on Build GRCh38
rs747614022 Chr.20: 18142965-18142965 on Build GRCh38
rs954745839 Chr.20: 18142895-18142895 on Build GRCh38
rs754854036 Chr.20: 18142966-18142966 on Build GRCh38
rs747217959 Chr.20: 18142903-18142903 on Build GRCh38
rs34336937 Chr.20: 18142805-18142806 on Build GRCh38
rs952003629 Chr.20: 18143015-18143015 on Build GRCh38
rs562825031 Chr.20: 18143116-18143116 on Build GRCh38
rs760685954 Chr.20: 18142812-18142812 on Build GRCh38
rs538239335 Chr.20: 18143117-18143117 on Build GRCh38
rs766534925 Chr.20: 18142919-18142919 on Build GRCh38
rs144392833 Chr.20: 18143095-18143095 on Build GRCh38
rs984636401 Chr.20: 18143020-18143020 on Build GRCh38
rs769676065 Chr.20: 18143043-18143043 on Build GRCh38
rs781003411 Chr.20: 18142781-18142781 on Build GRCh38
rs778165809 Chr.20: 18142902-18142902 on Build GRCh38
rs760721010 Chr.20: 18143029-18143029 on Build GRCh38
rs755016705 Chr.20: 18142979-18142979 on Build GRCh38
rs746112160 Chr.20: 18142777-18142777 on Build GRCh38
rs775461065 Chr.20: 18142792-18142792 on Build GRCh38
rs756122127 Chr.20: 18142896-18142896 on Build GRCh38
rs1023951006 Chr.20: 18142957-18142957 on Build GRCh38
rs1049929323 Chr.20: 18142910-18142910 on Build GRCh38
rs546142589 Chr.20: 18143111-18143111 on Build GRCh38
rs911475101 Chr.20: 18143154-18143154 on Build GRCh38
rs746588449 Chr.20: 18142778-18142778 on Build GRCh38
rs761563935 Chr.20: 18142935-18142935 on Build GRCh38
rs990784404 Chr.20: 18142956-18142956 on Build GRCh38
rs201326150 Chr.20: 18142912-18142912 on Build GRCh38
rs780123577 Chr.20: 18142898-18142898 on Build GRCh38
rs11557574 Chr.20: 18142801-18142801 on Build GRCh38
rs760836622 Chr.20: 18142723-18142723 on Build GRCh38
rs758097005 Chr.20: 18142774-18142774 on Build GRCh38
rs749104372 Chr.20: 18142900-18142900 on Build GRCh38

View Full Product Details