Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 108894428-108894744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
UniGene
Hs.645481
Gene Chromosome Location
Chr.6: 108848410-108990738 on Build GRCh38
Transcripts
NM_001286609.1 NM_032131.5 XM_005267154.3 XM_005267155.2 XM_005267157.3 XM_006715573.2 XM_006715574.2 XM_011536166.1 XM_011536167.1 XM_011536168.2 XM_011536170.2 XM_011536172.2 XM_011536176.2 XM_017011350.1 XM_017011351.1 XM_017011352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604877 c.684G>T p.K228N Chr.6: 108894479-108894479 on Build GRCh38
3674513 c.672-1G>T p.? Chr.6: 108894466-108894466 on Build GRCh38
7157848 c.686G>T p.R229I Chr.6: 108894481-108894481 on Build GRCh38
1487099 c.692C>T p.A231V Chr.6: 108894487-108894487 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757674902 Chr.6: 108894470-108894470 on Build GRCh38
rs1055237739 Chr.6: 108894472-108894472 on Build GRCh38
rs751887172 Chr.6: 108894669-108894669 on Build GRCh38
rs909479461 Chr.6: 108894670-108894670 on Build GRCh38
rs999157350 Chr.6: 108894656-108894656 on Build GRCh38
rs1010952333 Chr.6: 108894488-108894488 on Build GRCh38
rs761877694 Chr.6: 108894500-108894500 on Build GRCh38
rs372199328 Chr.6: 108894583-108894583 on Build GRCh38
rs746816457 Chr.6: 108894489-108894489 on Build GRCh38
rs200206972 Chr.6: 108894487-108894487 on Build GRCh38
rs202205491 Chr.6: 108894542-108894542 on Build GRCh38
rs753089434 Chr.6: 108894452-108894452 on Build GRCh38
rs749952163 Chr.6: 108894571-108894571 on Build GRCh38
rs546062110 Chr.6: 108894504-108894504 on Build GRCh38
rs561061919 Chr.6: 108894640-108894640 on Build GRCh38
rs544333974 Chr.6: 108894575-108894575 on Build GRCh38
rs759795133 Chr.6: 108894558-108894559 on Build GRCh38
rs1055556634 Chr.6: 108894702-108894702 on Build GRCh38
rs745314436 Chr.6: 108894485-108894485 on Build GRCh38
rs370951932 Chr.6: 108894536-108894536 on Build GRCh38
rs112110455 Chr.6: 108894462-108894462 on Build GRCh38
rs575311737 Chr.6: 108894558-108894558 on Build GRCh38
rs779246494 Chr.6: 108894568-108894568 on Build GRCh38
rs776568087 Chr.6: 108894497-108894497 on Build GRCh38
rs533794667 Chr.6: 108894459-108894459 on Build GRCh38
rs975849273 Chr.6: 108894644-108894644 on Build GRCh38
rs768633566 Chr.6: 108894494-108894494 on Build GRCh38
rs757804284 Chr.6: 108894555-108894555 on Build GRCh38
rs1031552311 Chr.6: 108894572-108894572 on Build GRCh38
rs754651244 Chr.6: 108894589-108894589 on Build GRCh38
rs769592319 Chr.6: 108894506-108894506 on Build GRCh38
rs574445763 Chr.6: 108894707-108894707 on Build GRCh38
rs1024586337 Chr.6: 108894492-108894492 on Build GRCh38
rs942276036 Chr.6: 108894700-108894700 on Build GRCh38
rs747851752 Chr.6: 108894593-108894593 on Build GRCh38
rs767785950 Chr.6: 108894521-108894521 on Build GRCh38
rs781185368 Chr.6: 108894590-108894590 on Build GRCh38
rs756403058 Chr.6: 108894458-108894458 on Build GRCh38
rs752860322 Chr.6: 108894530-108894530 on Build GRCh38
rs964846303 Chr.6: 108894640-108894640 on Build GRCh38
rs774508939 Chr.6: 108894507-108894507 on Build GRCh38
rs1007372176 Chr.6: 108894712-108894712 on Build GRCh38
rs938222960 Chr.6: 108894463-108894463 on Build GRCh38
rs778535144 Chr.6: 108894477-108894477 on Build GRCh38
rs753568161 Chr.6: 108894511-108894511 on Build GRCh38
rs368871367 Chr.6: 108894579-108894579 on Build GRCh38

View Full Product Details