Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136487860-136488459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRS3
Entrez Gene ID
680
Gene Name
bombesin receptor subtype 3
Gene Aliases
BB3, BB3R
UniGene
Hs.121484
Gene Chromosome Location
Chr.X: 136487966-136492439 on Build GRCh38
Transcripts
NM_001727.1

SNP Detail

SNP ID Chromosome Location
rs757902588 Chr.X: 136488252-136488252 on Build GRCh38
rs374260727 Chr.X: 136488283-136488283 on Build GRCh38
rs188910675 Chr.X: 136488157-136488157 on Build GRCh38
rs371326828 Chr.X: 136488227-136488227 on Build GRCh38
rs776596971 Chr.X: 136488411-136488411 on Build GRCh38
rs749901988 Chr.X: 136488097-136488098 on Build GRCh38
rs1007299289 Chr.X: 136487890-136487890 on Build GRCh38
rs377344433 Chr.X: 136488077-136488077 on Build GRCh38
rs954364243 Chr.X: 136488024-136488024 on Build GRCh38
rs780278040 Chr.X: 136488313-136488313 on Build GRCh38
rs959105584 Chr.X: 136488017-136488017 on Build GRCh38
rs762987858 Chr.X: 136487941-136487941 on Build GRCh38
rs766182965 Chr.X: 136488091-136488091 on Build GRCh38
rs373228133 Chr.X: 136488190-136488190 on Build GRCh38
rs764845622 Chr.X: 136488433-136488433 on Build GRCh38
rs1026123540 Chr.X: 136487948-136487948 on Build GRCh38
rs866206784 Chr.X: 136488170-136488170 on Build GRCh38
rs778396417 Chr.X: 136488348-136488348 on Build GRCh38
rs193231682 Chr.X: 136488202-136488202 on Build GRCh38
rs759354340 Chr.X: 136488431-136488431 on Build GRCh38
rs773179746 Chr.X: 136488225-136488225 on Build GRCh38
rs5233 Chr.X: 136488420-136488420 on Build GRCh38
rs781457304 Chr.X: 136488177-136488177 on Build GRCh38
rs752170427 Chr.X: 136488251-136488251 on Build GRCh38
rs1006920654 Chr.X: 136488374-136488374 on Build GRCh38
rs987352373 Chr.X: 136488040-136488040 on Build GRCh38
rs746930970 Chr.X: 136488363-136488363 on Build GRCh38
rs746074637 Chr.X: 136488178-136488178 on Build GRCh38
rs141322014 Chr.X: 136488226-136488226 on Build GRCh38
rs764901364 Chr.X: 136488236-136488236 on Build GRCh38
rs990880312 Chr.X: 136488041-136488041 on Build GRCh38
rs754604236 Chr.X: 136488133-136488133 on Build GRCh38
rs138184246 Chr.X: 136488285-136488285 on Build GRCh38
rs776244488 Chr.X: 136487985-136487985 on Build GRCh38
rs752053125 Chr.X: 136488254-136488254 on Build GRCh38
rs781574380 Chr.X: 136488078-136488080 on Build GRCh38
rs757582272 Chr.X: 136488176-136488176 on Build GRCh38
rs149571479 Chr.X: 136488381-136488381 on Build GRCh38
rs752183219 Chr.X: 136488150-136488150 on Build GRCh38
rs148394265 Chr.X: 136487936-136487936 on Build GRCh38
rs771317269 Chr.X: 136487950-136487950 on Build GRCh38
rs759264397 Chr.X: 136488233-136488233 on Build GRCh38
rs5231 Chr.X: 136487965-136487965 on Build GRCh38
rs761432263 Chr.X: 136488136-136488136 on Build GRCh38
rs974911225 Chr.X: 136487973-136487973 on Build GRCh38
rs147022210 Chr.X: 136488238-136488238 on Build GRCh38
rs765250829 Chr.X: 136487910-136487910 on Build GRCh38
rs757673866 Chr.X: 136488277-136488277 on Build GRCh38
rs200304646 Chr.X: 136488201-136488201 on Build GRCh38
rs5232 Chr.X: 136488271-136488271 on Build GRCh38
rs755312035 Chr.X: 136488053-136488053 on Build GRCh38
rs183662695 Chr.X: 136488007-136488007 on Build GRCh38
rs770937346 Chr.X: 136488372-136488372 on Build GRCh38
rs768750902 Chr.X: 136488339-136488339 on Build GRCh38
rs758858613 Chr.X: 136488152-136488152 on Build GRCh38
rs1019728384 Chr.X: 136487891-136487891 on Build GRCh38
rs751766496 Chr.X: 136488052-136488052 on Build GRCh38
rs747423670 Chr.X: 136488175-136488175 on Build GRCh38
rs769943934 Chr.X: 136488184-136488184 on Build GRCh38
rs112007204 Chr.X: 136488224-136488224 on Build GRCh38
rs749381844 Chr.X: 136488315-136488315 on Build GRCh38
rs776486052 Chr.X: 136488229-136488229 on Build GRCh38
rs756324988 Chr.X: 136488287-136488287 on Build GRCh38
rs760501414 Chr.X: 136488076-136488076 on Build GRCh38

View Full Product Details