Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 57010316-57010876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB1
Entrez Gene ID
1600
Gene Name
DAB1, reelin adaptor protein
Gene Aliases
-
UniGene
Hs.477370
Gene Chromosome Location
Chr.1: 56997906-58250539 on Build GRCh38
Transcripts
NM_021080.3

Gene Symbol
LOC105378749
Entrez Gene ID
105378749
Gene Name
uncharacterized LOC105378749
Gene Aliases
-
UniGene
Hs.677235
Gene Chromosome Location
Chr.1: 56998058-57014700 on Build GRCh38
Transcripts
NM_021080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865890 c.1620C>T p.P540P Chr.1: 57010743-57010743 on Build GRCh38
4895147 c.1595C>T p.S532F Chr.1: 57010768-57010768 on Build GRCh38
3865891 c.1719C>T p.P573P Chr.1: 57010743-57010743 on Build GRCh38
3491235 c.1764C>T p.S588S Chr.1: 57010698-57010698 on Build GRCh38
3491237 c.1751C>T p.P584L Chr.1: 57010711-57010711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748631653 Chr.1: 57010785-57010785 on Build GRCh38
rs751379532 Chr.1: 57010705-57010705 on Build GRCh38
rs747682626 Chr.1: 57010718-57010720 on Build GRCh38
rs371334388 Chr.1: 57010719-57010719 on Build GRCh38
rs141647150 Chr.1: 57010704-57010704 on Build GRCh38
rs1019171521 Chr.1: 57010822-57010823 on Build GRCh38
rs1013720368 Chr.1: 57010540-57010540 on Build GRCh38
rs542456415 Chr.1: 57010484-57010484 on Build GRCh38
rs774587041 Chr.1: 57010656-57010656 on Build GRCh38
rs980043123 Chr.1: 57010336-57010336 on Build GRCh38
rs534024730 Chr.1: 57010808-57010808 on Build GRCh38
rs769421465 Chr.1: 57010815-57010815 on Build GRCh38
rs988354986 Chr.1: 57010608-57010608 on Build GRCh38
rs1050539645 Chr.1: 57010417-57010417 on Build GRCh38
rs868782174 Chr.1: 57010377-57010377 on Build GRCh38
rs189702573 Chr.1: 57010508-57010508 on Build GRCh38
rs1023378053 Chr.1: 57010541-57010541 on Build GRCh38
rs200983396 Chr.1: 57010659-57010659 on Build GRCh38
rs910662617 Chr.1: 57010825-57010825 on Build GRCh38
rs769477450 Chr.1: 57010731-57010731 on Build GRCh38
rs994096388 Chr.1: 57010479-57010479 on Build GRCh38
rs756349172 Chr.1: 57010722-57010722 on Build GRCh38
rs898400135 Chr.1: 57010473-57010473 on Build GRCh38
rs565912104 Chr.1: 57010797-57010797 on Build GRCh38
rs114941053 Chr.1: 57010703-57010703 on Build GRCh38
rs965055767 Chr.1: 57010840-57010840 on Build GRCh38
rs540098277 Chr.1: 57010400-57010400 on Build GRCh38
rs755457249 Chr.1: 57010753-57010753 on Build GRCh38
rs967362052 Chr.1: 57010558-57010558 on Build GRCh38
rs750694191 Chr.1: 57010718-57010718 on Build GRCh38
rs774875118 Chr.1: 57010816-57010816 on Build GRCh38
rs753891209 Chr.1: 57010741-57010741 on Build GRCh38
rs778265900 Chr.1: 57010802-57010802 on Build GRCh38
rs192923126 Chr.1: 57010625-57010625 on Build GRCh38
rs756130777 Chr.1: 57010544-57010544 on Build GRCh38
rs949079514 Chr.1: 57010790-57010790 on Build GRCh38
rs1001023992 Chr.1: 57010552-57010552 on Build GRCh38
rs780069039 Chr.1: 57010737-57010737 on Build GRCh38
rs182338635 Chr.1: 57010577-57010577 on Build GRCh38
rs745582673 Chr.1: 57010814-57010814 on Build GRCh38
rs761557337 Chr.1: 57010707-57010707 on Build GRCh38
rs942136750 Chr.1: 57010692-57010692 on Build GRCh38
rs770601815 Chr.1: 57010812-57010813 on Build GRCh38
rs139040792 Chr.1: 57010660-57010660 on Build GRCh38
rs754867950 Chr.1: 57010344-57010344 on Build GRCh38
rs776459757 Chr.1: 57010661-57010661 on Build GRCh38
rs551230518 Chr.1: 57010813-57010813 on Build GRCh38
rs187868016 Chr.1: 57010337-57010337 on Build GRCh38
rs899671826 Chr.1: 57010385-57010385 on Build GRCh38
rs200096242 Chr.1: 57010821-57010821 on Build GRCh38
rs767269448 Chr.1: 57010710-57010710 on Build GRCh38
rs769244253 Chr.1: 57010690-57010690 on Build GRCh38
rs1036909309 Chr.1: 57010694-57010694 on Build GRCh38
rs762474956 Chr.1: 57010817-57010817 on Build GRCh38
rs187826949 Chr.1: 57010581-57010581 on Build GRCh38
rs923293233 Chr.1: 57010390-57010390 on Build GRCh38
rs959543980 Chr.1: 57010506-57010506 on Build GRCh38
rs568313303 Chr.1: 57010649-57010649 on Build GRCh38
rs1038633522 Chr.1: 57010468-57010468 on Build GRCh38
rs576680134 Chr.1: 57010466-57010466 on Build GRCh38
rs1033700736 Chr.1: 57010493-57010493 on Build GRCh38
rs906498009 Chr.1: 57010483-57010483 on Build GRCh38
rs896439458 Chr.1: 57010776-57010776 on Build GRCh38
rs1048356650 Chr.1: 57010436-57010436 on Build GRCh38
rs777386158 Chr.1: 57010809-57010812 on Build GRCh38
rs909832421 Chr.1: 57010666-57010666 on Build GRCh38
rs759305520 Chr.1: 57010822-57010823 on Build GRCh38
rs753027354 Chr.1: 57010489-57010489 on Build GRCh38
rs779432585 Chr.1: 57010757-57010757 on Build GRCh38
rs577207324 Chr.1: 57010357-57010357 on Build GRCh38
rs150708313 Chr.1: 57010578-57010578 on Build GRCh38
rs774026265 Chr.1: 57010813-57010814 on Build GRCh38
rs768508821 Chr.1: 57010822-57010822 on Build GRCh38
rs141367079 Chr.1: 57010441-57010441 on Build GRCh38
rs369711925 Chr.1: 57010689-57010689 on Build GRCh38
rs769204679 Chr.1: 57010655-57010655 on Build GRCh38

View Full Product Details