Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42962318-42962875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Gene Symbol
GNMT
Entrez Gene ID
27232
Gene Name
glycine N-methyltransferase
Gene Aliases
HEL-S-182mP
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42960754-42963880 on Build GRCh38
Transcripts
NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1 NM_001318865.1 NM_018960.5 NR_134899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7411406 c.316C>A p.P106T Chr.6: 42962321-42962321 on Build GRCh38
1079381 c.430G>A p.A144T Chr.6: 42962857-42962857 on Build GRCh38
3079443 c.431C>T p.A144V Chr.6: 42962858-42962858 on Build GRCh38
6672419 c.339C>T p.I113I Chr.6: 42962766-42962766 on Build GRCh38
5097581 c.334+3A>G p.? Chr.6: 42962342-42962342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs997449610 Chr.6: 42962534-42962535 on Build GRCh38
rs572545021 Chr.6: 42962378-42962378 on Build GRCh38
rs190613711 Chr.6: 42962838-42962838 on Build GRCh38
rs1028072024 Chr.6: 42962553-42962553 on Build GRCh38
rs143043221 Chr.6: 42962800-42962800 on Build GRCh38
rs748566577 Chr.6: 42962724-42962724 on Build GRCh38
rs181830984 Chr.6: 42962856-42962856 on Build GRCh38
rs977270384 Chr.6: 42962707-42962707 on Build GRCh38
rs888783198 Chr.6: 42962631-42962631 on Build GRCh38
rs561305071 Chr.6: 42962649-42962649 on Build GRCh38
rs529994595 Chr.6: 42962639-42962639 on Build GRCh38
rs777594419 Chr.6: 42962365-42962365 on Build GRCh38
rs763777365 Chr.6: 42962778-42962778 on Build GRCh38
rs751060218 Chr.6: 42962369-42962369 on Build GRCh38
rs543707887 Chr.6: 42962446-42962446 on Build GRCh38
rs966386321 Chr.6: 42962589-42962589 on Build GRCh38
rs759363288 Chr.6: 42962766-42962766 on Build GRCh38
rs1021391829 Chr.6: 42962650-42962650 on Build GRCh38
rs113082620 Chr.6: 42962540-42962540 on Build GRCh38
rs776738631 Chr.6: 42962765-42962765 on Build GRCh38
rs747209151 Chr.6: 42962852-42962852 on Build GRCh38
rs772126379 Chr.6: 42962730-42962730 on Build GRCh38
rs758814048 Chr.6: 42962849-42962850 on Build GRCh38
rs370786846 Chr.6: 42962767-42962767 on Build GRCh38
rs558770242 Chr.6: 42962364-42962364 on Build GRCh38
rs1037176478 Chr.6: 42962837-42962837 on Build GRCh38
rs985880457 Chr.6: 42962421-42962421 on Build GRCh38
rs749442227 Chr.6: 42962554-42962554 on Build GRCh38
rs760980193 Chr.6: 42962511-42962511 on Build GRCh38
rs993250212 Chr.6: 42962461-42962461 on Build GRCh38
rs541524958 Chr.6: 42962601-42962601 on Build GRCh38
rs374221964 Chr.6: 42962382-42962382 on Build GRCh38
rs770984841 Chr.6: 42962744-42962744 on Build GRCh38
rs115749165 Chr.6: 42962721-42962721 on Build GRCh38
rs3800292 Chr.6: 42962571-42962571 on Build GRCh38
rs864321678 Chr.6: 42962849-42962849 on Build GRCh38
rs148199736 Chr.6: 42962828-42962828 on Build GRCh38
rs557243794 Chr.6: 42962468-42962468 on Build GRCh38
rs141132997 Chr.6: 42962846-42962846 on Build GRCh38
rs376412328 Chr.6: 42962795-42962795 on Build GRCh38
rs751482570 Chr.6: 42962595-42962595 on Build GRCh38
rs576938593 Chr.6: 42962482-42962482 on Build GRCh38
rs554957004 Chr.6: 42962430-42962430 on Build GRCh38
rs749747704 Chr.6: 42962383-42962383 on Build GRCh38
rs1030434474 Chr.6: 42962592-42962592 on Build GRCh38
rs755468965 Chr.6: 42962361-42962361 on Build GRCh38
rs570363037 Chr.6: 42962696-42962696 on Build GRCh38
rs752579906 Chr.6: 42962358-42962358 on Build GRCh38
rs769257752 Chr.6: 42962719-42962719 on Build GRCh38
rs574896156 Chr.6: 42962445-42962445 on Build GRCh38
rs950102375 Chr.6: 42962677-42962677 on Build GRCh38
rs1048462684 Chr.6: 42962509-42962509 on Build GRCh38
rs1007989413 Chr.6: 42962560-42962560 on Build GRCh38
rs1037122954 Chr.6: 42962513-42962513 on Build GRCh38
rs938587124 Chr.6: 42962450-42962450 on Build GRCh38
rs748483425 Chr.6: 42962739-42962740 on Build GRCh38
rs908525566 Chr.6: 42962705-42962705 on Build GRCh38
rs754989144 Chr.6: 42962566-42962566 on Build GRCh38
rs530437991 Chr.6: 42962662-42962662 on Build GRCh38
rs116551815 Chr.6: 42962512-42962512 on Build GRCh38
rs751256417 Chr.6: 42962794-42962794 on Build GRCh38
rs9381227 Chr.6: 42962673-42962673 on Build GRCh38
rs979343661 Chr.6: 42962621-42962621 on Build GRCh38
rs924523011 Chr.6: 42962447-42962447 on Build GRCh38
rs550187950 Chr.6: 42962692-42962692 on Build GRCh38
rs370454543 Chr.6: 42962817-42962817 on Build GRCh38
rs546013977 Chr.6: 42962502-42962502 on Build GRCh38
rs779466342 Chr.6: 42962379-42962379 on Build GRCh38
rs915903013 Chr.6: 42962494-42962494 on Build GRCh38
rs764916813 Chr.6: 42962343-42962343 on Build GRCh38
rs968416888 Chr.6: 42962694-42962694 on Build GRCh38
rs997019650 Chr.6: 42962741-42962741 on Build GRCh38
rs375296683 Chr.6: 42962774-42962774 on Build GRCh38
rs749978652 Chr.6: 42962810-42962810 on Build GRCh38
rs745660734 Chr.6: 42962732-42962732 on Build GRCh38
rs535014756 Chr.6: 42962420-42962420 on Build GRCh38
rs984054622 Chr.6: 42962791-42962791 on Build GRCh38
rs747297365 Chr.6: 42962734-42962734 on Build GRCh38
rs753965293 Chr.6: 42962354-42962354 on Build GRCh38
rs369254135 Chr.6: 42962607-42962607 on Build GRCh38

View Full Product Details