Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31936907-31937457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2
Entrez Gene ID
717
Gene Name
complement component 2
Gene Aliases
ARMD14, CO2
UniGene
Hs.408903
Gene Chromosome Location
Chr.6: 31897785-31945674 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NR_104191.1

Gene Symbol
C2-AS1
Entrez Gene ID
102060414
Gene Name
C2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571112
Gene Chromosome Location
Chr.6: 31934474-31941724 on Build GRCh38
Transcripts
NM_000063.5 NM_001145903.2 NM_001178063.2 NM_001282457.1 NM_001282458.1 NR_104191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362452 c.1017C>A p.T339T Chr.6: 31937347-31937347 on Build GRCh38
3624869 c.994G>A p.E332K Chr.6: 31937324-31937324 on Build GRCh38
5870229 c.995delA p.N333fs*11 Chr.6: 31937325-31937325 on Build GRCh38
3624868 c.991C>T p.H331Y Chr.6: 31937321-31937321 on Build GRCh38
5915497 c.989-3C>T p.? Chr.6: 31937316-31937316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200964591 Chr.6: 31937199-31937200 on Build GRCh38
rs781227209 Chr.6: 31937382-31937382 on Build GRCh38
rs9279478 Chr.6: 31937222-31937222 on Build GRCh38
rs1003940786 Chr.6: 31937128-31937128 on Build GRCh38
rs761204218 Chr.6: 31937304-31937304 on Build GRCh38
rs866130958 Chr.6: 31937432-31937432 on Build GRCh38
rs754195921 Chr.6: 31937349-31937349 on Build GRCh38
rs915333334 Chr.6: 31937244-31937244 on Build GRCh38
rs556020050 Chr.6: 31937260-31937260 on Build GRCh38
rs971042718 Chr.6: 31937125-31937125 on Build GRCh38
rs145799939 Chr.6: 31937394-31937394 on Build GRCh38
rs192297029 Chr.6: 31937172-31937172 on Build GRCh38
rs58719596 Chr.6: 31937207-31937207 on Build GRCh38
rs894644101 Chr.6: 31936946-31936946 on Build GRCh38
rs773563930 Chr.6: 31937279-31937279 on Build GRCh38
rs968915982 Chr.6: 31937253-31937253 on Build GRCh38
rs140693719 Chr.6: 31937144-31937144 on Build GRCh38
rs886061293 Chr.6: 31937396-31937396 on Build GRCh38
rs201511739 Chr.6: 31936957-31936959 on Build GRCh38
rs984334693 Chr.6: 31937119-31937119 on Build GRCh38
rs369051082 Chr.6: 31936956-31936958 on Build GRCh38
rs747697695 Chr.6: 31937417-31937417 on Build GRCh38
rs878910475 Chr.6: 31937207-31937208 on Build GRCh38
rs985348112 Chr.6: 31937312-31937312 on Build GRCh38
rs965324161 Chr.6: 31937066-31937066 on Build GRCh38
rs1042798818 Chr.6: 31936987-31936987 on Build GRCh38
rs145877701 Chr.6: 31936953-31936953 on Build GRCh38
rs758382884 Chr.6: 31937370-31937370 on Build GRCh38
rs956723296 Chr.6: 31937186-31937187 on Build GRCh38
rs888588098 Chr.6: 31937081-31937081 on Build GRCh38
rs28986182 Chr.6: 31937208-31937209 on Build GRCh38
rs1040752112 Chr.6: 31936943-31936943 on Build GRCh38
rs1031721233 Chr.6: 31937020-31937020 on Build GRCh38
rs61730574 Chr.6: 31937412-31937412 on Build GRCh38
rs963751568 Chr.6: 31937166-31937166 on Build GRCh38
rs537472753 Chr.6: 31937227-31937227 on Build GRCh38
rs538818827 Chr.6: 31937393-31937393 on Build GRCh38
rs879085755 Chr.6: 31937206-31937207 on Build GRCh38
rs904279699 Chr.6: 31937352-31937352 on Build GRCh38
rs561080660 Chr.6: 31936932-31936932 on Build GRCh38
rs761770385 Chr.6: 31937372-31937372 on Build GRCh38
rs771513144 Chr.6: 31937418-31937418 on Build GRCh38
rs993021352 Chr.6: 31937248-31937248 on Build GRCh38
rs989493407 Chr.6: 31937216-31937216 on Build GRCh38
rs998534815 Chr.6: 31937004-31937004 on Build GRCh38
rs878930285 Chr.6: 31937206-31937206 on Build GRCh38
rs766677286 Chr.6: 31937321-31937321 on Build GRCh38
rs761100503 Chr.6: 31937433-31937433 on Build GRCh38
rs1028185530 Chr.6: 31937106-31937106 on Build GRCh38
rs1042663 Chr.6: 31937353-31937353 on Build GRCh38
rs765550448 Chr.6: 31937355-31937355 on Build GRCh38
rs866292178 Chr.6: 31937258-31937258 on Build GRCh38
rs761391221 Chr.6: 31937207-31937207 on Build GRCh38
rs532729243 Chr.6: 31937111-31937111 on Build GRCh38
rs57937265 Chr.6: 31937208-31937208 on Build GRCh38
rs139113913 Chr.6: 31937426-31937426 on Build GRCh38
rs752881306 Chr.6: 31937361-31937361 on Build GRCh38
rs1052827440 Chr.6: 31937377-31937377 on Build GRCh38
rs565597972 Chr.6: 31937109-31937109 on Build GRCh38
rs778517883 Chr.6: 31937403-31937403 on Build GRCh38
rs998100859 Chr.6: 31937075-31937075 on Build GRCh38
rs768357652 Chr.6: 31937401-31937401 on Build GRCh38
rs972097151 Chr.6: 31937195-31937195 on Build GRCh38
rs144611083 Chr.6: 31937160-31937160 on Build GRCh38
rs948928258 Chr.6: 31937278-31937278 on Build GRCh38
rs9332721 Chr.6: 31936953-31936955 on Build GRCh38
rs187383698 Chr.6: 31936988-31936988 on Build GRCh38

View Full Product Details