Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45609626-45610121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML2
Entrez Gene ID
24139
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
ELP70, EMAP-2, EMAP2
UniGene
Hs.24178
Gene Chromosome Location
Chr.19: 45609396-45645601 on Build GRCh38
Transcripts
NM_001193268.1 NM_001193269.1 NM_012155.2 NR_034098.1 XM_006723125.2 XM_006723126.2 XM_011526694.2 XM_011526695.2 XM_011526698.2 XM_011526700.2 XM_011526701.2 XM_017026550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274774 c.1890C>A p.D630E Chr.19: 45609723-45609723 on Build GRCh38
6287570 c.1893C>A p.S631R Chr.19: 45609720-45609720 on Build GRCh38
4127714 c.1859A>G p.H620R Chr.19: 45609754-45609754 on Build GRCh38
4913542 c.2320T>C p.L774L Chr.19: 45609734-45609734 on Build GRCh38
4680882 c.2354_2355insG p.K786fs*>12 Chr.19: 45609699-45609700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757185768 Chr.19: 45610025-45610025 on Build GRCh38
rs893439575 Chr.19: 45610082-45610082 on Build GRCh38
rs756751133 Chr.19: 45609714-45609714 on Build GRCh38
rs944964067 Chr.19: 45609790-45609790 on Build GRCh38
rs544510778 Chr.19: 45610060-45610060 on Build GRCh38
rs536338716 Chr.19: 45610048-45610048 on Build GRCh38
rs116310579 Chr.19: 45610034-45610034 on Build GRCh38
rs764353231 Chr.19: 45609716-45609716 on Build GRCh38
rs779017141 Chr.19: 45609648-45609648 on Build GRCh38
rs767129308 Chr.19: 45609812-45609812 on Build GRCh38
rs999267706 Chr.19: 45610031-45610031 on Build GRCh38
rs769550501 Chr.19: 45609664-45609664 on Build GRCh38
rs531252928 Chr.19: 45609958-45609958 on Build GRCh38
rs746114004 Chr.19: 45609764-45609764 on Build GRCh38
rs149432784 Chr.19: 45609703-45609703 on Build GRCh38
rs897924231 Chr.19: 45609838-45609838 on Build GRCh38
rs748912148 Chr.19: 45609700-45609727 on Build GRCh38
rs749067314 Chr.19: 45609781-45609781 on Build GRCh38
rs764596009 Chr.19: 45609840-45609840 on Build GRCh38
rs762665531 Chr.19: 45609670-45609670 on Build GRCh38
rs910789124 Chr.19: 45609787-45609787 on Build GRCh38
rs934731074 Chr.19: 45609866-45609866 on Build GRCh38
rs568858107 Chr.19: 45609951-45609951 on Build GRCh38
rs764062379 Chr.19: 45610022-45610022 on Build GRCh38
rs201508885 Chr.19: 45609660-45609660 on Build GRCh38
rs555319672 Chr.19: 45609722-45609722 on Build GRCh38
rs529589308 Chr.19: 45609895-45609895 on Build GRCh38
rs897475832 Chr.19: 45609983-45609983 on Build GRCh38
rs770650644 Chr.19: 45609786-45609786 on Build GRCh38
rs774122975 Chr.19: 45609791-45609791 on Build GRCh38
rs924925693 Chr.19: 45610081-45610081 on Build GRCh38
rs540437081 Chr.19: 45609885-45609885 on Build GRCh38
rs376150890 Chr.19: 45609922-45609922 on Build GRCh38
rs73050333 Chr.19: 45609890-45609890 on Build GRCh38
rs773023936 Chr.19: 45609777-45609777 on Build GRCh38
rs746540115 Chr.19: 45609709-45609709 on Build GRCh38
rs201166343 Chr.19: 45609904-45609904 on Build GRCh38
rs533474367 Chr.19: 45609867-45609867 on Build GRCh38
rs1044948915 Chr.19: 45609913-45609913 on Build GRCh38
rs199957663 Chr.19: 45609729-45609729 on Build GRCh38
rs993160382 Chr.19: 45610003-45610003 on Build GRCh38
rs905084701 Chr.19: 45609914-45609914 on Build GRCh38
rs1046265622 Chr.19: 45610021-45610021 on Build GRCh38
rs924610521 Chr.19: 45609841-45609841 on Build GRCh38
rs758453328 Chr.19: 45609770-45609770 on Build GRCh38
rs374687287 Chr.19: 45609659-45609659 on Build GRCh38
rs544684948 Chr.19: 45609834-45609834 on Build GRCh38
rs930687930 Chr.19: 45609920-45609920 on Build GRCh38
rs1010585099 Chr.19: 45610098-45610098 on Build GRCh38
rs1027339376 Chr.19: 45610019-45610019 on Build GRCh38
rs532755028 Chr.19: 45610023-45610023 on Build GRCh38
rs748076682 Chr.19: 45609774-45609774 on Build GRCh38
rs765986079 Chr.19: 45609671-45609671 on Build GRCh38
rs956232287 Chr.19: 45610094-45610094 on Build GRCh38
rs758671956 Chr.19: 45610009-45610009 on Build GRCh38
rs769833074 Chr.19: 45609775-45609775 on Build GRCh38
rs377253305 Chr.19: 45609719-45609719 on Build GRCh38
rs976086864 Chr.19: 45609796-45609796 on Build GRCh38
rs3745816 Chr.19: 45609649-45609649 on Build GRCh38
rs368671855 Chr.19: 45609888-45609888 on Build GRCh38
rs199538741 Chr.19: 45609889-45609890 on Build GRCh38
rs773130735 Chr.19: 45609667-45609667 on Build GRCh38
rs551358226 Chr.19: 45609961-45609961 on Build GRCh38
rs190700619 Chr.19: 45609673-45609673 on Build GRCh38
rs200762252 Chr.19: 45609741-45609741 on Build GRCh38
rs768054324 Chr.19: 45609804-45609807 on Build GRCh38
rs760276339 Chr.19: 45609836-45609836 on Build GRCh38
rs759300949 Chr.19: 45609806-45609806 on Build GRCh38
rs1048762826 Chr.19: 45609931-45609931 on Build GRCh38
rs946515171 Chr.19: 45609899-45609899 on Build GRCh38
rs976750323 Chr.19: 45610065-45610065 on Build GRCh38
rs1036441462 Chr.19: 45609864-45609864 on Build GRCh38
rs182889975 Chr.19: 45609845-45609845 on Build GRCh38
rs780151012 Chr.19: 45609771-45609771 on Build GRCh38
rs547163582 Chr.19: 45610085-45610085 on Build GRCh38
rs542537075 Chr.19: 45609674-45609674 on Build GRCh38

View Full Product Details