Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126208974-126209483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1L1-AS2
Entrez Gene ID
100862662
Gene Name
ALDH1L1 antisense RNA 2
Gene Aliases
-
UniGene
Hs.633362
Gene Chromosome Location
Chr.3: 126180065-126210169 on Build GRCh38
Transcripts
NR_046383.1

SNP Detail

SNP ID Chromosome Location
rs956436741 Chr.3: 126209194-126209194 on Build GRCh38
rs1057255572 Chr.3: 126209033-126209033 on Build GRCh38
rs377114583 Chr.3: 126209228-126209228 on Build GRCh38
rs572536397 Chr.3: 126209313-126209313 on Build GRCh38
rs138492540 Chr.3: 126209397-126209397 on Build GRCh38
rs577913127 Chr.3: 126209399-126209399 on Build GRCh38
rs895412885 Chr.3: 126209045-126209045 on Build GRCh38
rs147684908 Chr.3: 126209073-126209073 on Build GRCh38
rs918463336 Chr.3: 126209192-126209192 on Build GRCh38
rs140467389 Chr.3: 126209269-126209269 on Build GRCh38
rs761643503 Chr.3: 126209270-126209270 on Build GRCh38
rs890687048 Chr.3: 126209392-126209392 on Build GRCh38
rs938643038 Chr.3: 126209027-126209027 on Build GRCh38
rs915088195 Chr.3: 126209212-126209212 on Build GRCh38
rs771007386 Chr.3: 126209007-126209007 on Build GRCh38
rs899303644 Chr.3: 126209092-126209092 on Build GRCh38
rs1055023643 Chr.3: 126209037-126209037 on Build GRCh38
rs549638513 Chr.3: 126209053-126209053 on Build GRCh38
rs151247343 Chr.3: 126209218-126209218 on Build GRCh38
rs1037756642 Chr.3: 126209335-126209335 on Build GRCh38
rs150478638 Chr.3: 126209338-126209338 on Build GRCh38
rs773029729 Chr.3: 126209405-126209405 on Build GRCh38
rs886510203 Chr.3: 126209051-126209051 on Build GRCh38
rs769423743 Chr.3: 126209341-126209341 on Build GRCh38
rs1029284031 Chr.3: 126209357-126209357 on Build GRCh38
rs965293823 Chr.3: 126209129-126209129 on Build GRCh38
rs768632326 Chr.3: 126209120-126209120 on Build GRCh38
rs976727860 Chr.3: 126209404-126209404 on Build GRCh38
rs143213913 Chr.3: 126209123-126209123 on Build GRCh38
rs895110561 Chr.3: 126209234-126209234 on Build GRCh38
rs977073377 Chr.3: 126209132-126209132 on Build GRCh38
rs765190853 Chr.3: 126209221-126209221 on Build GRCh38
rs529149680 Chr.3: 126209433-126209433 on Build GRCh38
rs989165315 Chr.3: 126209202-126209202 on Build GRCh38
rs909683105 Chr.3: 126209454-126209454 on Build GRCh38
rs927926530 Chr.3: 126209455-126209455 on Build GRCh38
rs533273535 Chr.3: 126209024-126209024 on Build GRCh38
rs999316208 Chr.3: 126209077-126209077 on Build GRCh38
rs987536790 Chr.3: 126209133-126209133 on Build GRCh38
rs940769857 Chr.3: 126209327-126209327 on Build GRCh38
rs142444192 Chr.3: 126209109-126209109 on Build GRCh38
rs969287851 Chr.3: 126209443-126209443 on Build GRCh38
rs534401013 Chr.3: 126209106-126209106 on Build GRCh38
rs984372068 Chr.3: 126209428-126209428 on Build GRCh38
rs1030734774 Chr.3: 126209152-126209152 on Build GRCh38
rs938725715 Chr.3: 126209240-126209240 on Build GRCh38
rs13100771 Chr.3: 126209213-126209213 on Build GRCh38
rs556852889 Chr.3: 126209176-126209176 on Build GRCh38
rs543323657 Chr.3: 126209421-126209421 on Build GRCh38
rs112528060 Chr.3: 126209460-126209460 on Build GRCh38
rs11917873 Chr.3: 126209103-126209103 on Build GRCh38
rs949676345 Chr.3: 126209264-126209264 on Build GRCh38
rs905870765 Chr.3: 126209023-126209023 on Build GRCh38
rs529014714 Chr.3: 126209099-126209099 on Build GRCh38
rs142926280 Chr.3: 126209440-126209440 on Build GRCh38
rs942088188 Chr.3: 126209224-126209224 on Build GRCh38
rs113413988 Chr.3: 126209049-126209049 on Build GRCh38
rs1006607307 Chr.3: 126209347-126209347 on Build GRCh38
rs967276969 Chr.3: 126209074-126209074 on Build GRCh38
rs981005040 Chr.3: 126209445-126209445 on Build GRCh38
rs540126659 Chr.3: 126209127-126209127 on Build GRCh38
rs116795371 Chr.3: 126209424-126209424 on Build GRCh38
rs1016460739 Chr.3: 126209087-126209087 on Build GRCh38

View Full Product Details